Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27368509-27369089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093560 c.424C>T p.Q142* Chr.2: 27369060-27369060 on Build GRCh38
5666590 c.399A>T p.T133T Chr.2: 27369025-27369025 on Build GRCh38
2939907 c.440C>G p.P147R Chr.2: 27369044-27369044 on Build GRCh38
4983035 c.505C>G p.Q169E Chr.2: 27368707-27368707 on Build GRCh38
4093558 c.469A>C p.T157P Chr.2: 27369015-27369015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978812492 Chr.2: 27368888-27368888 on Build GRCh38
rs1036621304 Chr.2: 27368967-27368967 on Build GRCh38
rs752260211 Chr.2: 27368745-27368745 on Build GRCh38
rs776773132 Chr.2: 27368956-27368956 on Build GRCh38
rs201042063 Chr.2: 27368640-27368640 on Build GRCh38
rs754559917 Chr.2: 27368599-27368599 on Build GRCh38
rs374526553 Chr.2: 27368766-27368766 on Build GRCh38
rs758627794 Chr.2: 27368582-27368582 on Build GRCh38
rs1049908754 Chr.2: 27368924-27368924 on Build GRCh38
rs923612148 Chr.2: 27368819-27368819 on Build GRCh38
rs759358033 Chr.2: 27368828-27368828 on Build GRCh38
rs76522591 Chr.2: 27368806-27368806 on Build GRCh38
rs947539380 Chr.2: 27368570-27368570 on Build GRCh38
rs751135430 Chr.2: 27368588-27368588 on Build GRCh38
rs758641876 Chr.2: 27368535-27368535 on Build GRCh38
rs747320250 Chr.2: 27368767-27368767 on Build GRCh38
rs761810644 Chr.2: 27368711-27368711 on Build GRCh38
rs770591078 Chr.2: 27368661-27368661 on Build GRCh38
rs914750494 Chr.2: 27368562-27368562 on Build GRCh38
rs371511880 Chr.2: 27368748-27368748 on Build GRCh38
rs1027298754 Chr.2: 27369009-27369009 on Build GRCh38
rs368507949 Chr.2: 27368978-27368978 on Build GRCh38
rs527417427 Chr.2: 27368830-27368830 on Build GRCh38
rs149121402 Chr.2: 27368723-27368723 on Build GRCh38
rs988113567 Chr.2: 27368839-27368839 on Build GRCh38
rs779782574 Chr.2: 27368583-27368583 on Build GRCh38
rs371253140 Chr.2: 27369032-27369032 on Build GRCh38
rs748155307 Chr.2: 27368634-27368634 on Build GRCh38
rs773154770 Chr.2: 27369064-27369064 on Build GRCh38
rs985907495 Chr.2: 27368802-27368802 on Build GRCh38
rs201503266 Chr.2: 27368666-27368666 on Build GRCh38
rs569664227 Chr.2: 27368697-27368697 on Build GRCh38
rs886038382 Chr.2: 27368617-27368617 on Build GRCh38
rs769575268 Chr.2: 27369018-27369026 on Build GRCh38
rs757134177 Chr.2: 27369018-27369018 on Build GRCh38
rs755723756 Chr.2: 27369010-27369010 on Build GRCh38
rs542746816 Chr.2: 27368842-27368842 on Build GRCh38
rs149520370 Chr.2: 27369038-27369038 on Build GRCh38
rs897166165 Chr.2: 27368936-27368936 on Build GRCh38
rs377054241 Chr.2: 27368822-27368822 on Build GRCh38
rs570777761 Chr.2: 27368740-27368740 on Build GRCh38
rs961728137 Chr.2: 27368747-27368747 on Build GRCh38
rs765743112 Chr.2: 27368714-27368714 on Build GRCh38
rs774576919 Chr.2: 27368984-27368984 on Build GRCh38
rs879039338 Chr.2: 27368798-27368798 on Build GRCh38
rs754665816 Chr.2: 27368725-27368725 on Build GRCh38
rs368243788 Chr.2: 27368724-27368724 on Build GRCh38
rs937433718 Chr.2: 27368904-27368904 on Build GRCh38
rs753868838 Chr.2: 27369014-27369014 on Build GRCh38
rs1053334796 Chr.2: 27368540-27368540 on Build GRCh38
rs913265804 Chr.2: 27368538-27368538 on Build GRCh38
rs914826534 Chr.2: 27368869-27368869 on Build GRCh38
rs748102093 Chr.2: 27369052-27369053 on Build GRCh38
rs763488648 Chr.2: 27369012-27369012 on Build GRCh38
rs866357253 Chr.2: 27368670-27368670 on Build GRCh38
rs567344773 Chr.2: 27368897-27368897 on Build GRCh38
rs767290808 Chr.2: 27368996-27368996 on Build GRCh38
rs559965006 Chr.2: 27368763-27368763 on Build GRCh38
rs367810151 Chr.2: 27369000-27369000 on Build GRCh38
rs1056356918 Chr.2: 27368926-27368926 on Build GRCh38
rs141872864 Chr.2: 27369052-27369052 on Build GRCh38
rs926045766 Chr.2: 27368901-27368901 on Build GRCh38
rs770699036 Chr.2: 27368746-27368746 on Build GRCh38
rs775819795 Chr.2: 27368662-27368662 on Build GRCh38
rs779235094 Chr.2: 27368744-27368746 on Build GRCh38
rs529735415 Chr.2: 27368639-27368639 on Build GRCh38
rs773813808 Chr.2: 27368971-27368971 on Build GRCh38
rs768867189 Chr.2: 27368780-27368780 on Build GRCh38
rs1048334665 Chr.2: 27368946-27368946 on Build GRCh38
rs199827769 Chr.2: 27368993-27368993 on Build GRCh38
rs146732803 Chr.2: 27369039-27369039 on Build GRCh38
rs147672203 Chr.2: 27368669-27368669 on Build GRCh38
rs946111331 Chr.2: 27368552-27368552 on Build GRCh38
rs911319154 Chr.2: 27368925-27368925 on Build GRCh38
rs778839851 Chr.2: 27369023-27369023 on Build GRCh38
rs750788095 Chr.2: 27368579-27368579 on Build GRCh38
rs768779136 Chr.2: 27368685-27368685 on Build GRCh38
rs554691151 Chr.2: 27368859-27368859 on Build GRCh38
rs966346571 Chr.2: 27368824-27368824 on Build GRCh38
rs776685186 Chr.2: 27368695-27368695 on Build GRCh38
rs754878418 Chr.2: 27368970-27368990 on Build GRCh38
rs1008673121 Chr.2: 27368647-27368647 on Build GRCh38
rs777786550 Chr.2: 27368644-27368644 on Build GRCh38
rs750850634 Chr.2: 27368719-27368719 on Build GRCh38
rs1041075414 Chr.2: 27368935-27368935 on Build GRCh38
rs200581892 Chr.2: 27368960-27368960 on Build GRCh38
rs80051818 Chr.2: 27368937-27368937 on Build GRCh38
rs749149001 Chr.2: 27368752-27368752 on Build GRCh38
rs374686395 Chr.2: 27369045-27369045 on Build GRCh38
rs779565150 Chr.2: 27368588-27368589 on Build GRCh38
rs534790807 Chr.2: 27368750-27368750 on Build GRCh38
rs56402945 Chr.2: 27368841-27368841 on Build GRCh38
rs370413772 Chr.2: 27368650-27368650 on Build GRCh38
rs750669369 Chr.2: 27368962-27368967 on Build GRCh38
rs377518092 Chr.2: 27368620-27368620 on Build GRCh38
rs766827254 Chr.2: 27368982-27368982 on Build GRCh38
rs770808088 Chr.2: 27368765-27368765 on Build GRCh38
rs944140079 Chr.2: 27368929-27368929 on Build GRCh38

View Full Product Details