Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36552478-36553069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ2
Entrez Gene ID
9637
Gene Name
fasciculation and elongation protein zeta 2
Gene Aliases
HUM3CL
UniGene
Hs.258563
Gene Chromosome Location
Chr.2: 36552239-36598190 on Build GRCh38
Transcripts
NM_001042548.1 NM_005102.2 XM_006712153.2

SNP Detail

SNP ID Chromosome Location
rs918978915 Chr.2: 36552518-36552518 on Build GRCh38
rs1011700091 Chr.2: 36552689-36552690 on Build GRCh38
rs769091020 Chr.2: 36552776-36552776 on Build GRCh38
rs901776860 Chr.2: 36552617-36552617 on Build GRCh38
rs745923284 Chr.2: 36552705-36552705 on Build GRCh38
rs1047296402 Chr.2: 36552837-36552837 on Build GRCh38
rs922872593 Chr.2: 36552784-36552784 on Build GRCh38
rs1023073673 Chr.2: 36552915-36552916 on Build GRCh38
rs1045170356 Chr.2: 36552682-36552682 on Build GRCh38
rs961570052 Chr.2: 36552993-36552993 on Build GRCh38
rs1005295059 Chr.2: 36552667-36552667 on Build GRCh38
rs72868427 Chr.2: 36552906-36552906 on Build GRCh38
rs910049221 Chr.2: 36552839-36552839 on Build GRCh38
rs772136699 Chr.2: 36552738-36552738 on Build GRCh38
rs537912698 Chr.2: 36552896-36552896 on Build GRCh38
rs878943856 Chr.2: 36552946-36552946 on Build GRCh38
rs930419792 Chr.2: 36552519-36552519 on Build GRCh38
rs1031515452 Chr.2: 36552652-36552652 on Build GRCh38
rs747479386 Chr.2: 36552976-36552976 on Build GRCh38
rs1035880822 Chr.2: 36552753-36552754 on Build GRCh38
rs1041267320 Chr.2: 36552569-36552569 on Build GRCh38
rs1047244460 Chr.2: 36552642-36552642 on Build GRCh38
rs1049167040 Chr.2: 36552525-36552525 on Build GRCh38
rs150362693 Chr.2: 36552855-36552855 on Build GRCh38
rs930388385 Chr.2: 36552817-36552817 on Build GRCh38
rs1003877071 Chr.2: 36552697-36552697 on Build GRCh38
rs386389934 Chr.2: 36552514-36552515 on Build GRCh38
rs888226608 Chr.2: 36552663-36552663 on Build GRCh38
rs964051048 Chr.2: 36552762-36552762 on Build GRCh38
rs893003672 Chr.2: 36552908-36552909 on Build GRCh38
rs956556591 Chr.2: 36552710-36552710 on Build GRCh38
rs5830428 Chr.2: 36552512-36552513 on Build GRCh38
rs184409641 Chr.2: 36552550-36552550 on Build GRCh38
rs577998064 Chr.2: 36552706-36552706 on Build GRCh38
rs778244358 Chr.2: 36552541-36552541 on Build GRCh38
rs1009429725 Chr.2: 36552985-36552985 on Build GRCh38
rs3076528 Chr.2: 36552513-36552514 on Build GRCh38
rs374906288 Chr.2: 36552565-36552565 on Build GRCh38
rs532723871 Chr.2: 36553023-36553023 on Build GRCh38
rs540675904 Chr.2: 36552899-36552899 on Build GRCh38
rs1003186265 Chr.2: 36552874-36552874 on Build GRCh38
rs904816601 Chr.2: 36552691-36552691 on Build GRCh38
rs397774857 Chr.2: 36552517-36552518 on Build GRCh38
rs148282508 Chr.2: 36552712-36552712 on Build GRCh38
rs1022121439 Chr.2: 36553031-36553031 on Build GRCh38
rs972842283 Chr.2: 36552756-36552756 on Build GRCh38
rs575479914 Chr.2: 36552753-36552753 on Build GRCh38
rs181966332 Chr.2: 36552683-36552683 on Build GRCh38
rs1012061860 Chr.2: 36552910-36552910 on Build GRCh38
rs543159095 Chr.2: 36552798-36552798 on Build GRCh38
rs185851546 Chr.2: 36552832-36552832 on Build GRCh38
rs140289170 Chr.2: 36552739-36552739 on Build GRCh38
rs998659268 Chr.2: 36552639-36552639 on Build GRCh38
rs930107711 Chr.2: 36552609-36552609 on Build GRCh38
rs573896953 Chr.2: 36552819-36552819 on Build GRCh38
rs760419412 Chr.2: 36552940-36552942 on Build GRCh38
rs1002801412 Chr.2: 36552934-36552934 on Build GRCh38
rs904870057 Chr.2: 36552868-36552868 on Build GRCh38
rs1022068975 Chr.2: 36552737-36552737 on Build GRCh38
rs967875241 Chr.2: 36553033-36553033 on Build GRCh38
rs1000703157 Chr.2: 36553039-36553039 on Build GRCh38
rs905961966 Chr.2: 36552925-36552926 on Build GRCh38
rs35775049 Chr.2: 36552516-36552517 on Build GRCh38
rs11885432 Chr.2: 36552607-36552607 on Build GRCh38
rs941109054 Chr.2: 36552841-36552841 on Build GRCh38

View Full Product Details