Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108923224-108923821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDAR
Entrez Gene ID
10913
Gene Name
ectodysplasin A receptor
Gene Aliases
DL, ECTD10A, ECTD10B, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, HRM1
UniGene
Hs.171971
Gene Chromosome Location
Chr.2: 108894471-108989372 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680154 c.429delC p.N144fs*45 Chr.2: 108923381-108923381 on Build GRCh38
3708806 c.357-3T>C p.? Chr.2: 108923456-108923456 on Build GRCh38
714790 c.380C>T p.P127L Chr.2: 108923430-108923430 on Build GRCh38
1613281 c.357-3T>C p.? Chr.2: 108923456-108923456 on Build GRCh38
6378511 c.363C>A p.Y121* Chr.2: 108923447-108923447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766028277 Chr.2: 108923404-108923404 on Build GRCh38
rs745501063 Chr.2: 108923356-108923357 on Build GRCh38
rs778267481 Chr.2: 108923323-108923323 on Build GRCh38
rs780312834 Chr.2: 108923664-108923664 on Build GRCh38
rs771832164 Chr.2: 108923382-108923382 on Build GRCh38
rs540520804 Chr.2: 108923458-108923458 on Build GRCh38
rs757595565 Chr.2: 108923430-108923430 on Build GRCh38
rs533786849 Chr.2: 108923288-108923288 on Build GRCh38
rs930043367 Chr.2: 108923397-108923397 on Build GRCh38
rs544647280 Chr.2: 108923793-108923793 on Build GRCh38
rs534458369 Chr.2: 108923632-108923632 on Build GRCh38
rs746726414 Chr.2: 108923466-108923466 on Build GRCh38
rs1023295434 Chr.2: 108923583-108923583 on Build GRCh38
rs990289058 Chr.2: 108923802-108923802 on Build GRCh38
rs1034559980 Chr.2: 108923678-108923678 on Build GRCh38
rs749313377 Chr.2: 108923449-108923449 on Build GRCh38
rs748224 Chr.2: 108923580-108923580 on Build GRCh38
rs971457524 Chr.2: 108923675-108923675 on Build GRCh38
rs577303846 Chr.2: 108923775-108923775 on Build GRCh38
rs546904176 Chr.2: 108923569-108923569 on Build GRCh38
rs376168791 Chr.2: 108923462-108923462 on Build GRCh38
rs752040374 Chr.2: 108923423-108923423 on Build GRCh38
rs934194350 Chr.2: 108923769-108923769 on Build GRCh38
rs367605908 Chr.2: 108923471-108923471 on Build GRCh38
rs113809781 Chr.2: 108923286-108923286 on Build GRCh38
rs767913764 Chr.2: 108923504-108923504 on Build GRCh38
rs774744688 Chr.2: 108923488-108923488 on Build GRCh38
rs774257841 Chr.2: 108923369-108923369 on Build GRCh38
rs1050996962 Chr.2: 108923637-108923637 on Build GRCh38
rs994286974 Chr.2: 108923668-108923668 on Build GRCh38
rs927762012 Chr.2: 108923656-108923656 on Build GRCh38
rs1054652018 Chr.2: 108923797-108923797 on Build GRCh38
rs368841777 Chr.2: 108923367-108923367 on Build GRCh38
rs555333048 Chr.2: 108923635-108923635 on Build GRCh38
rs149933447 Chr.2: 108923553-108923553 on Build GRCh38
rs773029710 Chr.2: 108923394-108923394 on Build GRCh38
rs544304309 Chr.2: 108923324-108923324 on Build GRCh38
rs566699731 Chr.2: 108923588-108923588 on Build GRCh38
rs77401860 Chr.2: 108923676-108923676 on Build GRCh38
rs760178861 Chr.2: 108923302-108923302 on Build GRCh38
rs374404022 Chr.2: 108923354-108923354 on Build GRCh38
rs902397631 Chr.2: 108923374-108923374 on Build GRCh38
rs758801305 Chr.2: 108923321-108923321 on Build GRCh38
rs972038870 Chr.2: 108923500-108923500 on Build GRCh38
rs558009004 Chr.2: 108923651-108923651 on Build GRCh38
rs761525478 Chr.2: 108923375-108923375 on Build GRCh38
rs941370719 Chr.2: 108923459-108923459 on Build GRCh38
rs903355718 Chr.2: 108923529-108923529 on Build GRCh38
rs551050118 Chr.2: 108923246-108923246 on Build GRCh38
rs139905358 Chr.2: 108923287-108923287 on Build GRCh38
rs201033204 Chr.2: 108923339-108923339 on Build GRCh38
rs564477144 Chr.2: 108923396-108923396 on Build GRCh38
rs1047591578 Chr.2: 108923409-108923409 on Build GRCh38
rs144922479 Chr.2: 108923639-108923639 on Build GRCh38
rs562313577 Chr.2: 108923467-108923467 on Build GRCh38
rs1042431518 Chr.2: 108923603-108923603 on Build GRCh38
rs780171221 Chr.2: 108923446-108923446 on Build GRCh38
rs764524798 Chr.2: 108923421-108923421 on Build GRCh38
rs113124398 Chr.2: 108923480-108923480 on Build GRCh38
rs3749107 Chr.2: 108923318-108923318 on Build GRCh38
rs763575900 Chr.2: 108923415-108923415 on Build GRCh38
rs998140625 Chr.2: 108923401-108923401 on Build GRCh38
rs950345090 Chr.2: 108923301-108923301 on Build GRCh38
rs750696392 Chr.2: 108923431-108923431 on Build GRCh38
rs533398401 Chr.2: 108923574-108923574 on Build GRCh38
rs1051233793 Chr.2: 108923514-108923514 on Build GRCh38
rs982857935 Chr.2: 108923665-108923665 on Build GRCh38
rs144492072 Chr.2: 108923340-108923340 on Build GRCh38
rs569288463 Chr.2: 108923261-108923261 on Build GRCh38
rs768720529 Chr.2: 108923451-108923451 on Build GRCh38
rs74690043 Chr.2: 108923309-108923309 on Build GRCh38
rs946500101 Chr.2: 108923589-108923589 on Build GRCh38
rs954481157 Chr.2: 108923281-108923281 on Build GRCh38
rs753394117 Chr.2: 108923413-108923413 on Build GRCh38
rs539905986 Chr.2: 108923650-108923650 on Build GRCh38
rs370198195 Chr.2: 108923327-108923327 on Build GRCh38
rs778718816 Chr.2: 108923452-108923452 on Build GRCh38
rs182230412 Chr.2: 108923561-108923561 on Build GRCh38
rs958961191 Chr.2: 108923680-108923680 on Build GRCh38
rs748225 Chr.2: 108923457-108923457 on Build GRCh38
rs756294692 Chr.2: 108923441-108923441 on Build GRCh38
rs376733198 Chr.2: 108923486-108923486 on Build GRCh38
rs762364356 Chr.2: 108923499-108923499 on Build GRCh38
rs921273845 Chr.2: 108923742-108923742 on Build GRCh38
rs999262153 Chr.2: 108923560-108923560 on Build GRCh38
rs184890557 Chr.2: 108923669-108923669 on Build GRCh38
rs903324300 Chr.2: 108923792-108923793 on Build GRCh38
rs539813982 Chr.2: 108923268-108923268 on Build GRCh38

View Full Product Details