Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134838433-134838814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACMSD
Entrez Gene ID
130013
Gene Name
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases
-
UniGene
Hs.655728
Gene Chromosome Location
Chr.2: 134838319-134902906 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_011510592.2 XM_017003325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1006680 c.37G>T p.E13* Chr.2: 134838719-134838719 on Build GRCh38
6333907 c.3G>T p.M1I Chr.2: 134838685-134838685 on Build GRCh38
1006679 c.8T>G p.I3S Chr.2: 134838690-134838690 on Build GRCh38
6592195 c.50_51insA p.R20fs*42 Chr.2: 134838732-134838733 on Build GRCh38
6592192 c.19A>G p.S7G Chr.2: 134838701-134838701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189203003 Chr.2: 134838629-134838629 on Build GRCh38
rs551015512 Chr.2: 134838581-134838581 on Build GRCh38
rs762462403 Chr.2: 134838655-134838655 on Build GRCh38
rs199786838 Chr.2: 134838696-134838696 on Build GRCh38
rs781161029 Chr.2: 134838634-134838634 on Build GRCh38
rs745433402 Chr.2: 134838784-134838784 on Build GRCh38
rs780469282 Chr.2: 134838753-134838753 on Build GRCh38
rs556160917 Chr.2: 134838766-134838766 on Build GRCh38
rs551174841 Chr.2: 134838458-134838458 on Build GRCh38
rs564731298 Chr.2: 134838471-134838471 on Build GRCh38
rs1007265141 Chr.2: 134838548-134838548 on Build GRCh38
rs755139468 Chr.2: 134838708-134838710 on Build GRCh38
rs758513803 Chr.2: 134838740-134838740 on Build GRCh38
rs961960842 Chr.2: 134838617-134838617 on Build GRCh38
rs200456967 Chr.2: 134838697-134838697 on Build GRCh38
rs893556699 Chr.2: 134838544-134838544 on Build GRCh38
rs770364386 Chr.2: 134838656-134838656 on Build GRCh38
rs748204970 Chr.2: 134838565-134838565 on Build GRCh38
rs754345616 Chr.2: 134838706-134838706 on Build GRCh38
rs1034904670 Chr.2: 134838591-134838591 on Build GRCh38
rs368027343 Chr.2: 134838783-134838783 on Build GRCh38
rs754244393 Chr.2: 134838510-134838510 on Build GRCh38
rs370081851 Chr.2: 134838712-134838712 on Build GRCh38
rs755979686 Chr.2: 134838763-134838763 on Build GRCh38
rs750615358 Chr.2: 134838720-134838720 on Build GRCh38
rs41320845 Chr.2: 134838715-134838715 on Build GRCh38
rs150617858 Chr.2: 134838545-134838545 on Build GRCh38
rs185091564 Chr.2: 134838595-134838595 on Build GRCh38
rs771700602 Chr.2: 134838786-134838786 on Build GRCh38
rs1040361179 Chr.2: 134838732-134838732 on Build GRCh38
rs747940524 Chr.2: 134838756-134838756 on Build GRCh38
rs1049451995 Chr.2: 134838507-134838507 on Build GRCh38
rs919097972 Chr.2: 134838494-134838494 on Build GRCh38
rs992144520 Chr.2: 134838741-134838741 on Build GRCh38
rs773352992 Chr.2: 134838654-134838654 on Build GRCh38
rs767697301 Chr.2: 134838766-134838766 on Build GRCh38
rs547358221 Chr.2: 134838497-134838497 on Build GRCh38
rs770438490 Chr.2: 134838778-134838778 on Build GRCh38
rs193088385 Chr.2: 134838644-134838644 on Build GRCh38
rs901614766 Chr.2: 134838607-134838607 on Build GRCh38
rs759142404 Chr.2: 134838670-134838670 on Build GRCh38
rs193050701 Chr.2: 134838457-134838457 on Build GRCh38
rs969485446 Chr.2: 134838579-134838579 on Build GRCh38
rs940082896 Chr.2: 134838475-134838475 on Build GRCh38
rs749209264 Chr.2: 134838774-134838774 on Build GRCh38
rs749426315 Chr.2: 134838725-134838725 on Build GRCh38
rs528675929 Chr.2: 134838480-134838480 on Build GRCh38
rs753040063 Chr.2: 134838694-134838694 on Build GRCh38
rs533725889 Chr.2: 134838663-134838663 on Build GRCh38
rs775192990 Chr.2: 134838790-134838790 on Build GRCh38
rs747993721 Chr.2: 134838635-134838635 on Build GRCh38
rs866657720 Chr.2: 134838651-134838651 on Build GRCh38
rs529564731 Chr.2: 134838555-134838555 on Build GRCh38
rs569122064 Chr.2: 134838583-134838583 on Build GRCh38
rs767240117 Chr.2: 134838677-134838677 on Build GRCh38
rs968627004 Chr.2: 134838667-134838667 on Build GRCh38

View Full Product Details