Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169537031-169537337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD1
Entrez Gene ID
79675
Gene Name
FAST kinase domains 1
Gene Aliases
-
UniGene
Hs.529276
Gene Chromosome Location
Chr.2: 169527879-169573921 on Build GRCh38
Transcripts
NM_001281476.2 NM_001322046.1 NM_001322048.1 NM_001322049.1 NM_024622.5 NR_104020.2 XM_006712750.3 XM_006712751.3 XM_017004911.1 XM_017004912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79837 c.2160G>A p.S720S Chr.2: 169537255-169537255 on Build GRCh38
74616 c.2125G>A p.E709K Chr.2: 169537290-169537290 on Build GRCh38
6858441 c.2100A>G p.Q700Q Chr.2: 169537315-169537315 on Build GRCh38
7112723 c.2180A>G p.H727R Chr.2: 169537235-169537235 on Build GRCh38
6660699 c.2168C>T p.T723M Chr.2: 169537247-169537247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959729849 Chr.2: 169537207-169537207 on Build GRCh38
rs781765852 Chr.2: 169537256-169537256 on Build GRCh38
rs889022195 Chr.2: 169537098-169537098 on Build GRCh38
rs759453718 Chr.2: 169537216-169537216 on Build GRCh38
rs756527039 Chr.2: 169537270-169537270 on Build GRCh38
rs557434091 Chr.2: 169537054-169537054 on Build GRCh38
rs748594138 Chr.2: 169537226-169537226 on Build GRCh38
rs1003400363 Chr.2: 169537202-169537202 on Build GRCh38
rs762324826 Chr.2: 169537189-169537189 on Build GRCh38
rs962162885 Chr.2: 169537149-169537149 on Build GRCh38
rs1021978977 Chr.2: 169537246-169537246 on Build GRCh38
rs929231476 Chr.2: 169537088-169537088 on Build GRCh38
rs773790032 Chr.2: 169537288-169537288 on Build GRCh38
rs779481627 Chr.2: 169537115-169537115 on Build GRCh38
rs34833763 Chr.2: 169537140-169537141 on Build GRCh38
rs758586669 Chr.2: 169537240-169537240 on Build GRCh38
rs1050334735 Chr.2: 169537095-169537095 on Build GRCh38
rs997436151 Chr.2: 169537080-169537080 on Build GRCh38
rs372027328 Chr.2: 169537255-169537255 on Build GRCh38
rs906354358 Chr.2: 169537151-169537151 on Build GRCh38
rs980064055 Chr.2: 169537310-169537310 on Build GRCh38
rs764599638 Chr.2: 169537290-169537292 on Build GRCh38
rs772952916 Chr.2: 169537179-169537179 on Build GRCh38
rs773894227 Chr.2: 169537210-169537210 on Build GRCh38
rs770454797 Chr.2: 169537192-169537192 on Build GRCh38
rs201465952 Chr.2: 169537247-169537247 on Build GRCh38
rs745383724 Chr.2: 169537291-169537291 on Build GRCh38
rs376288447 Chr.2: 169537281-169537281 on Build GRCh38
rs761248058 Chr.2: 169537236-169537236 on Build GRCh38
rs767453569 Chr.2: 169537219-169537219 on Build GRCh38
rs1006498361 Chr.2: 169537106-169537111 on Build GRCh38
rs748942515 Chr.2: 169537284-169537284 on Build GRCh38
rs750636088 Chr.2: 169537228-169537228 on Build GRCh38
rs752418771 Chr.2: 169537223-169537223 on Build GRCh38
rs139576803 Chr.2: 169537227-169537227 on Build GRCh38
rs746323442 Chr.2: 169537279-169537279 on Build GRCh38
rs748422410 Chr.2: 169537264-169537264 on Build GRCh38
rs1055267661 Chr.2: 169537222-169537222 on Build GRCh38
rs770639864 Chr.2: 169537287-169537287 on Build GRCh38
rs16857023 Chr.2: 169537055-169537055 on Build GRCh38

View Full Product Details