Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64226646-64227216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESR2
Entrez Gene ID
2100
Gene Name
estrogen receptor 2
Gene Aliases
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
UniGene
Hs.660607
Gene Chromosome Location
Chr.14: 64226712-64338631 on Build GRCh38
Transcripts
NM_001040275.1 NM_001214902.1 NM_001291712.1 NM_001291723.1 NR_073496.1

SNP Detail

SNP ID Chromosome Location
rs1009505921 Chr.14: 64226956-64226956 on Build GRCh38
rs534231123 Chr.14: 64226980-64226980 on Build GRCh38
rs1033472750 Chr.14: 64226991-64226991 on Build GRCh38
rs991697065 Chr.14: 64226877-64226877 on Build GRCh38
rs970860068 Chr.14: 64226795-64226795 on Build GRCh38
rs1055381716 Chr.14: 64227011-64227011 on Build GRCh38
rs887220923 Chr.14: 64226693-64226693 on Build GRCh38
rs558593421 Chr.14: 64227176-64227176 on Build GRCh38
rs1014407023 Chr.14: 64227070-64227071 on Build GRCh38
rs899846127 Chr.14: 64226729-64226729 on Build GRCh38
rs940175318 Chr.14: 64226977-64226977 on Build GRCh38
rs780094124 Chr.14: 64226873-64226873 on Build GRCh38
rs2772163 Chr.14: 64226667-64226667 on Build GRCh38
rs996888501 Chr.14: 64227115-64227115 on Build GRCh38
rs932679572 Chr.14: 64226927-64226927 on Build GRCh38
rs868843829 Chr.14: 64226707-64226707 on Build GRCh38
rs529944593 Chr.14: 64226720-64226720 on Build GRCh38
rs998647850 Chr.14: 64227089-64227089 on Build GRCh38
rs11622991 Chr.14: 64226748-64226748 on Build GRCh38
rs898849107 Chr.14: 64226715-64226718 on Build GRCh38
rs1032264733 Chr.14: 64226746-64226746 on Build GRCh38
rs745857611 Chr.14: 64226892-64226892 on Build GRCh38
rs932620064 Chr.14: 64227188-64227188 on Build GRCh38
rs575740643 Chr.14: 64226887-64226887 on Build GRCh38
rs528003706 Chr.14: 64226760-64226760 on Build GRCh38
rs908691300 Chr.14: 64226972-64226972 on Build GRCh38
rs944092251 Chr.14: 64226903-64226903 on Build GRCh38
rs955514083 Chr.14: 64227145-64227145 on Build GRCh38
rs908731182 Chr.14: 64226896-64226896 on Build GRCh38
rs1042584484 Chr.14: 64226970-64226970 on Build GRCh38
rs770930430 Chr.14: 64227141-64227141 on Build GRCh38
rs531587542 Chr.14: 64226745-64226745 on Build GRCh38
rs921392265 Chr.14: 64226932-64226932 on Build GRCh38
rs967347970 Chr.14: 64226863-64226863 on Build GRCh38
rs1032902691 Chr.14: 64226885-64226889 on Build GRCh38
rs1024539036 Chr.14: 64226810-64226810 on Build GRCh38
rs866540144 Chr.14: 64226708-64226708 on Build GRCh38
rs1055492217 Chr.14: 64227182-64227182 on Build GRCh38
rs1036016902 Chr.14: 64226710-64226711 on Build GRCh38
rs1045454732 Chr.14: 64227081-64227081 on Build GRCh38
rs116164997 Chr.14: 64227030-64227030 on Build GRCh38
rs1020683771 Chr.14: 64226782-64226782 on Build GRCh38
rs145278696 Chr.14: 64226735-64226735 on Build GRCh38
rs139004885 Chr.14: 64227083-64227083 on Build GRCh38
rs1014124161 Chr.14: 64226805-64226805 on Build GRCh38
rs547730274 Chr.14: 64226981-64226981 on Build GRCh38
rs1255998 Chr.14: 64227153-64227153 on Build GRCh38
rs974180368 Chr.14: 64227162-64227162 on Build GRCh38
rs1053689868 Chr.14: 64226942-64226942 on Build GRCh38
rs952870350 Chr.14: 64226888-64226888 on Build GRCh38
rs961529993 Chr.14: 64227159-64227159 on Build GRCh38
rs994441756 Chr.14: 64226755-64226755 on Build GRCh38
rs567510489 Chr.14: 64227009-64227009 on Build GRCh38
rs77943947 Chr.14: 64226802-64226802 on Build GRCh38
rs938264794 Chr.14: 64227178-64227178 on Build GRCh38
rs58262369 Chr.14: 64227194-64227194 on Build GRCh38
rs1027177805 Chr.14: 64227059-64227059 on Build GRCh38
rs576107158 Chr.14: 64227091-64227091 on Build GRCh38
rs770676995 Chr.14: 64226692-64226692 on Build GRCh38
rs188970860 Chr.14: 64226943-64226943 on Build GRCh38
rs893635211 Chr.14: 64226747-64226747 on Build GRCh38
rs2094914 Chr.14: 64226960-64226960 on Build GRCh38
rs921386014 Chr.14: 64227181-64227181 on Build GRCh38
rs887405573 Chr.14: 64226693-64226694 on Build GRCh38
rs977239999 Chr.14: 64227169-64227169 on Build GRCh38
rs987011502 Chr.14: 64226940-64226940 on Build GRCh38
rs551776512 Chr.14: 64226868-64226868 on Build GRCh38
rs931519102 Chr.14: 64226673-64226673 on Build GRCh38
rs1033726834 Chr.14: 64226845-64226845 on Build GRCh38
rs952943954 Chr.14: 64227084-64227084 on Build GRCh38
rs778874474 Chr.14: 64226804-64226804 on Build GRCh38
rs934065764 Chr.14: 64226933-64226933 on Build GRCh38
rs3842327 Chr.14: 64226698-64226699 on Build GRCh38
rs754892930 Chr.14: 64226904-64226904 on Build GRCh38
rs921226570 Chr.14: 64226919-64226919 on Build GRCh38
rs753837061 Chr.14: 64226876-64226876 on Build GRCh38
rs75492434 Chr.14: 64226696-64226696 on Build GRCh38
rs981895437 Chr.14: 64226828-64226828 on Build GRCh38
rs957266133 Chr.14: 64227110-64227110 on Build GRCh38
rs747171118 Chr.14: 64227133-64227133 on Build GRCh38
rs76801570 Chr.14: 64226698-64226698 on Build GRCh38
rs1047130316 Chr.14: 64226679-64226679 on Build GRCh38
rs549198121 Chr.14: 64226721-64226721 on Build GRCh38
rs758694599 Chr.14: 64227003-64227003 on Build GRCh38

View Full Product Details