Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69234568-69235104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
UniGene
Hs.533878
Gene Chromosome Location
Chr.14: 69191477-69244020 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_005267817.3 XM_011536908.2 XM_017021421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6335107 c.978G>T p.V326V Chr.14: 69234960-69234960 on Build GRCh38
6842328 c.638A>G p.H213R Chr.14: 69234995-69234995 on Build GRCh38
3497343 c.502C>T p.R168* Chr.14: 69234859-69234859 on Build GRCh38
5417423 c.574A>C p.N192H Chr.14: 69234931-69234931 on Build GRCh38
5417422 c.949A>C p.N317H Chr.14: 69234931-69234931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777777365 Chr.14: 69234934-69234934 on Build GRCh38
rs2139542 Chr.14: 69234734-69234734 on Build GRCh38
rs535766399 Chr.14: 69234712-69234712 on Build GRCh38
rs770383229 Chr.14: 69234859-69234859 on Build GRCh38
rs758339231 Chr.14: 69235015-69235015 on Build GRCh38
rs979844763 Chr.14: 69234624-69234624 on Build GRCh38
rs371679710 Chr.14: 69234922-69234922 on Build GRCh38
rs766930628 Chr.14: 69234739-69234739 on Build GRCh38
rs763324106 Chr.14: 69234875-69234875 on Build GRCh38
rs572634242 Chr.14: 69234763-69234763 on Build GRCh38
rs997479585 Chr.14: 69235057-69235057 on Build GRCh38
rs756516458 Chr.14: 69235072-69235072 on Build GRCh38
rs764241699 Chr.14: 69235034-69235034 on Build GRCh38
rs370954963 Chr.14: 69235057-69235058 on Build GRCh38
rs372445263 Chr.14: 69234995-69234995 on Build GRCh38
rs368212336 Chr.14: 69234970-69234970 on Build GRCh38
rs767011683 Chr.14: 69234864-69234864 on Build GRCh38
rs946451722 Chr.14: 69234602-69234602 on Build GRCh38
rs750146788 Chr.14: 69234787-69234787 on Build GRCh38
rs748285549 Chr.14: 69234700-69234700 on Build GRCh38
rs775769920 Chr.14: 69234846-69234846 on Build GRCh38
rs375032366 Chr.14: 69234956-69234956 on Build GRCh38
rs763427338 Chr.14: 69234980-69234980 on Build GRCh38
rs755996596 Chr.14: 69234668-69234668 on Build GRCh38
rs368972120 Chr.14: 69234982-69234982 on Build GRCh38
rs373683852 Chr.14: 69234806-69234806 on Build GRCh38
rs754820477 Chr.14: 69234916-69234916 on Build GRCh38
rs768709431 Chr.14: 69234679-69234679 on Build GRCh38
rs777295774 Chr.14: 69234745-69234745 on Build GRCh38
rs778935942 Chr.14: 69234919-69234919 on Build GRCh38
rs752648901 Chr.14: 69234920-69234920 on Build GRCh38
rs757506560 Chr.14: 69235042-69235042 on Build GRCh38
rs546218193 Chr.14: 69234770-69234770 on Build GRCh38
rs1006806862 Chr.14: 69235022-69235022 on Build GRCh38
rs765023615 Chr.14: 69235003-69235003 on Build GRCh38
rs761351759 Chr.14: 69234782-69234782 on Build GRCh38
rs747323333 Chr.14: 69234839-69234839 on Build GRCh38
rs766038338 Chr.14: 69234990-69234990 on Build GRCh38
rs745431467 Chr.14: 69235070-69235070 on Build GRCh38
rs537498635 Chr.14: 69234636-69234636 on Build GRCh38
rs766129898 Chr.14: 69234904-69234904 on Build GRCh38
rs776070848 Chr.14: 69234978-69234978 on Build GRCh38
rs568278424 Chr.14: 69234676-69234676 on Build GRCh38
rs747233095 Chr.14: 69234936-69234936 on Build GRCh38
rs988365882 Chr.14: 69234588-69234588 on Build GRCh38
rs759375341 Chr.14: 69234996-69234996 on Build GRCh38
rs367788970 Chr.14: 69234871-69234871 on Build GRCh38
rs752669990 Chr.14: 69235005-69235005 on Build GRCh38
rs373883869 Chr.14: 69234618-69234618 on Build GRCh38
rs764503211 Chr.14: 69234910-69234910 on Build GRCh38
rs530622642 Chr.14: 69234591-69234591 on Build GRCh38
rs887983719 Chr.14: 69234958-69234958 on Build GRCh38
rs772747829 Chr.14: 69234989-69234989 on Build GRCh38
rs753724220 Chr.14: 69234985-69234985 on Build GRCh38
rs896731318 Chr.14: 69234825-69234825 on Build GRCh38
rs771413672 Chr.14: 69234844-69234844 on Build GRCh38
rs370120407 Chr.14: 69234783-69234783 on Build GRCh38
rs556344811 Chr.14: 69234666-69234666 on Build GRCh38
rs770774040 Chr.14: 69234796-69234798 on Build GRCh38
rs751741372 Chr.14: 69235037-69235037 on Build GRCh38
rs760180688 Chr.14: 69234762-69234762 on Build GRCh38
rs772140674 Chr.14: 69234834-69234834 on Build GRCh38
rs774555740 Chr.14: 69234682-69234682 on Build GRCh38
rs554474914 Chr.14: 69234733-69234733 on Build GRCh38
rs760387735 Chr.14: 69234897-69234897 on Build GRCh38
rs778057014 Chr.14: 69234837-69234837 on Build GRCh38
rs913107411 Chr.14: 69234795-69234795 on Build GRCh38
rs74518015 Chr.14: 69235066-69235066 on Build GRCh38
rs576613897 Chr.14: 69234860-69234860 on Build GRCh38
rs746258479 Chr.14: 69234967-69234967 on Build GRCh38
rs780349133 Chr.14: 69235074-69235074 on Build GRCh38
rs557864570 Chr.14: 69234788-69234788 on Build GRCh38
rs754947748 Chr.14: 69234803-69234803 on Build GRCh38
rs868470727 Chr.14: 69234826-69234826 on Build GRCh38
rs748280203 Chr.14: 69234816-69234816 on Build GRCh38
rs543897449 Chr.14: 69234955-69234955 on Build GRCh38
rs746169695 Chr.14: 69235068-69235068 on Build GRCh38
rs779998314 Chr.14: 69234673-69234673 on Build GRCh38
rs927063226 Chr.14: 69234634-69234634 on Build GRCh38
rs773959450 Chr.14: 69234946-69234946 on Build GRCh38
rs753680327 Chr.14: 69234908-69234908 on Build GRCh38
rs753769783 Chr.14: 69234796-69234796 on Build GRCh38
rs761149650 Chr.14: 69234737-69234737 on Build GRCh38

View Full Product Details