Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34789947-34790544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTC1
Entrez Gene ID
70
Gene Name
actin, alpha, cardiac muscle 1
Gene Aliases
ACTC, ASD5, CMD1R, CMH11, LVNC4
UniGene
Hs.118127
Gene Chromosome Location
Chr.15: 34788096-34795726 on Build GRCh38
Transcripts
NM_005159.4 NR_120329.1

Gene Symbol
LOC101928174
Entrez Gene ID
101928174
Gene Name
uncharacterized LOC101928174
Gene Aliases
-
UniGene
Hs.680670
Gene Chromosome Location
Chr.15: 34755084-34812923 on Build GRCh38
Transcripts
NM_005159.4 NR_120329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4608194 c.1102G>A p.G368S Chr.15: 34790444-34790444 on Build GRCh38
7292779 c.1016A>G p.Y339C Chr.15: 34790530-34790530 on Build GRCh38
5546197 c.1110C>T p.S370S Chr.15: 34790436-34790436 on Build GRCh38
6481825 c.1026G>T p.W342C Chr.15: 34790520-34790520 on Build GRCh38
4054170 c.1109C>G p.S370C Chr.15: 34790437-34790437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748053539 Chr.15: 34790377-34790377 on Build GRCh38
rs886051084 Chr.15: 34789990-34789990 on Build GRCh38
rs373997876 Chr.15: 34790386-34790386 on Build GRCh38
rs928551623 Chr.15: 34789996-34789996 on Build GRCh38
rs866993141 Chr.15: 34790401-34790401 on Build GRCh38
rs540240407 Chr.15: 34790198-34790198 on Build GRCh38
rs727504415 Chr.15: 34790409-34790409 on Build GRCh38
rs28730667 Chr.15: 34790390-34790390 on Build GRCh38
rs1034845815 Chr.15: 34789976-34789976 on Build GRCh38
rs955752034 Chr.15: 34790256-34790256 on Build GRCh38
rs769780832 Chr.15: 34790382-34790382 on Build GRCh38
rs753985896 Chr.15: 34790204-34790204 on Build GRCh38
rs942411801 Chr.15: 34790318-34790318 on Build GRCh38
rs370919129 Chr.15: 34790366-34790366 on Build GRCh38
rs766325228 Chr.15: 34790199-34790199 on Build GRCh38
rs121912674 Chr.15: 34790458-34790458 on Build GRCh38
rs753155881 Chr.15: 34790400-34790400 on Build GRCh38
rs886051087 Chr.15: 34790128-34790128 on Build GRCh38
rs559979638 Chr.15: 34790101-34790101 on Build GRCh38
rs547800928 Chr.15: 34790076-34790076 on Build GRCh38
rs975556139 Chr.15: 34790044-34790044 on Build GRCh38
rs762711685 Chr.15: 34790078-34790078 on Build GRCh38
rs730880407 Chr.15: 34790434-34790434 on Build GRCh38
rs918355027 Chr.15: 34790038-34790038 on Build GRCh38
rs553652706 Chr.15: 34790296-34790296 on Build GRCh38
rs74009720 Chr.15: 34790143-34790143 on Build GRCh38
rs758951219 Chr.15: 34790215-34790215 on Build GRCh38
rs748062476 Chr.15: 34790508-34790508 on Build GRCh38
rs143407466 Chr.15: 34790392-34790392 on Build GRCh38
rs1003523293 Chr.15: 34790451-34790451 on Build GRCh38
rs910956441 Chr.15: 34790317-34790317 on Build GRCh38
rs756594449 Chr.15: 34790407-34790407 on Build GRCh38
rs374252209 Chr.15: 34790367-34790367 on Build GRCh38
rs759733343 Chr.15: 34790217-34790217 on Build GRCh38
rs868273758 Chr.15: 34790218-34790218 on Build GRCh38
rs1017185318 Chr.15: 34790224-34790224 on Build GRCh38
rs537551929 Chr.15: 34790252-34790252 on Build GRCh38
rs201258118 Chr.15: 34790396-34790396 on Build GRCh38
rs886051088 Chr.15: 34790165-34790165 on Build GRCh38
rs886051085 Chr.15: 34790019-34790019 on Build GRCh38
rs377546862 Chr.15: 34790372-34790372 on Build GRCh38
rs905147662 Chr.15: 34790098-34790098 on Build GRCh38
rs895745627 Chr.15: 34790496-34790496 on Build GRCh38
rs758173419 Chr.15: 34790448-34790448 on Build GRCh38
rs960487568 Chr.15: 34789981-34789981 on Build GRCh38
rs113835687 Chr.15: 34790514-34790514 on Build GRCh38
rs773231264 Chr.15: 34790383-34790383 on Build GRCh38
rs964271501 Chr.15: 34790227-34790227 on Build GRCh38
rs3729758 Chr.15: 34790373-34790373 on Build GRCh38
rs976399940 Chr.15: 34790333-34790333 on Build GRCh38
rs931049224 Chr.15: 34790057-34790057 on Build GRCh38
rs370256882 Chr.15: 34790447-34790447 on Build GRCh38
rs778272368 Chr.15: 34790326-34790326 on Build GRCh38
rs968143994 Chr.15: 34790009-34790009 on Build GRCh38
rs886051086 Chr.15: 34790105-34790105 on Build GRCh38
rs367884939 Chr.15: 34790297-34790297 on Build GRCh38
rs1370154 Chr.15: 34790024-34790024 on Build GRCh38
rs754736799 Chr.15: 34790463-34790463 on Build GRCh38
rs763198919 Chr.15: 34790384-34790384 on Build GRCh38
rs1038370444 Chr.15: 34790096-34790096 on Build GRCh38
rs934104142 Chr.15: 34790073-34790073 on Build GRCh38
rs1000527685 Chr.15: 34790221-34790221 on Build GRCh38
rs764683976 Chr.15: 34790415-34790415 on Build GRCh38
rs893906281 Chr.15: 34790174-34790174 on Build GRCh38
rs910850745 Chr.15: 34790088-34790088 on Build GRCh38
rs975607015 Chr.15: 34790229-34790229 on Build GRCh38
rs876657738 Chr.15: 34790462-34790462 on Build GRCh38
rs869025355 Chr.15: 34790422-34790422 on Build GRCh38
rs972554259 Chr.15: 34790246-34790246 on Build GRCh38
rs549446281 Chr.15: 34790264-34790264 on Build GRCh38
rs551820554 Chr.15: 34790188-34790188 on Build GRCh38
rs375435815 Chr.15: 34790461-34790461 on Build GRCh38
rs562187785 Chr.15: 34789985-34789985 on Build GRCh38
rs776430979 Chr.15: 34790376-34790376 on Build GRCh38
rs901415027 Chr.15: 34790111-34790111 on Build GRCh38
rs1009705105 Chr.15: 34789983-34789983 on Build GRCh38
rs959898660 Chr.15: 34790019-34790019 on Build GRCh38
rs1009386317 Chr.15: 34790150-34790150 on Build GRCh38
rs535622574 Chr.15: 34790291-34790291 on Build GRCh38
rs778288220 Chr.15: 34790469-34790469 on Build GRCh38
rs964261556 Chr.15: 34790027-34790027 on Build GRCh38
rs151321743 Chr.15: 34790493-34790493 on Build GRCh38
rs727505192 Chr.15: 34790507-34790507 on Build GRCh38
rs1056315352 Chr.15: 34790116-34790116 on Build GRCh38
rs550947264 Chr.15: 34790216-34790216 on Build GRCh38
rs140261885 Chr.15: 34790454-34790454 on Build GRCh38
rs149031785 Chr.15: 34790275-34790275 on Build GRCh38
rs1025398990 Chr.15: 34790260-34790260 on Build GRCh38
rs1002080271 Chr.15: 34790112-34790112 on Build GRCh38
rs984202027 Chr.15: 34790081-34790081 on Build GRCh38
rs1041356078 Chr.15: 34790108-34790108 on Build GRCh38
rs959926169 Chr.15: 34790242-34790242 on Build GRCh38
rs572031368 Chr.15: 34790301-34790301 on Build GRCh38
rs1035599172 Chr.15: 34790236-34790236 on Build GRCh38
rs773298484 Chr.15: 34790152-34790152 on Build GRCh38

View Full Product Details