Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67184688-67185255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L1
Entrez Gene ID
283849
Gene Name
exocyst complex component 3 like 1
Gene Aliases
EXOC3L
UniGene
Hs.647356
Gene Chromosome Location
Chr.16: 67184379-67190204 on Build GRCh38
Transcripts
NM_178516.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7418713 c.1823G>T p.C608F Chr.16: 67184984-67184984 on Build GRCh38
5740684 c.1901G>A p.S634N Chr.16: 67184906-67184906 on Build GRCh38
6657118 c.1935G>A p.P645P Chr.16: 67184781-67184781 on Build GRCh38
3691097 c.1900A>G p.S634G Chr.16: 67184907-67184907 on Build GRCh38
972323 c.1665G>C p.S555S Chr.16: 67185220-67185220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143965297 Chr.16: 67184962-67184962 on Build GRCh38
rs377657887 Chr.16: 67184729-67184729 on Build GRCh38
rs1025413712 Chr.16: 67185094-67185094 on Build GRCh38
rs770515857 Chr.16: 67184907-67184908 on Build GRCh38
rs1012410742 Chr.16: 67185013-67185013 on Build GRCh38
rs772023184 Chr.16: 67184879-67184879 on Build GRCh38
rs149002457 Chr.16: 67185188-67185188 on Build GRCh38
rs773713271 Chr.16: 67185036-67185044 on Build GRCh38
rs372580618 Chr.16: 67184806-67184806 on Build GRCh38
rs752107674 Chr.16: 67184851-67184851 on Build GRCh38
rs773653756 Chr.16: 67185153-67185153 on Build GRCh38
rs746848602 Chr.16: 67184978-67184978 on Build GRCh38
rs764938588 Chr.16: 67184846-67184846 on Build GRCh38
rs755106466 Chr.16: 67185085-67185085 on Build GRCh38
rs1022364177 Chr.16: 67184741-67184741 on Build GRCh38
rs778420591 Chr.16: 67185229-67185229 on Build GRCh38
rs111513602 Chr.16: 67185093-67185093 on Build GRCh38
rs774633963 Chr.16: 67184952-67184952 on Build GRCh38
rs747769813 Chr.16: 67185236-67185236 on Build GRCh38
rs770834369 Chr.16: 67184901-67184901 on Build GRCh38
rs754032003 Chr.16: 67185071-67185071 on Build GRCh38
rs761198789 Chr.16: 67185063-67185063 on Build GRCh38
rs773224804 Chr.16: 67184830-67184830 on Build GRCh38
rs747995921 Chr.16: 67184818-67184818 on Build GRCh38
rs555234021 Chr.16: 67184776-67184776 on Build GRCh38
rs903477219 Chr.16: 67184957-67184957 on Build GRCh38
rs201786873 Chr.16: 67184816-67184816 on Build GRCh38
rs780471406 Chr.16: 67184875-67184875 on Build GRCh38
rs370913149 Chr.16: 67185026-67185026 on Build GRCh38
rs752945742 Chr.16: 67185098-67185098 on Build GRCh38
rs375655845 Chr.16: 67185227-67185227 on Build GRCh38
rs371556682 Chr.16: 67184869-67184869 on Build GRCh38
rs756098820 Chr.16: 67185117-67185117 on Build GRCh38
rs781044046 Chr.16: 67184994-67184994 on Build GRCh38
rs546290851 Chr.16: 67184860-67184860 on Build GRCh38
rs775947137 Chr.16: 67184837-67184837 on Build GRCh38
rs769294128 Chr.16: 67185017-67185017 on Build GRCh38
rs747795615 Chr.16: 67185145-67185145 on Build GRCh38
rs896125966 Chr.16: 67184980-67184980 on Build GRCh38
rs367672691 Chr.16: 67184864-67184864 on Build GRCh38
rs947779710 Chr.16: 67185205-67185205 on Build GRCh38
rs73585101 Chr.16: 67184858-67184858 on Build GRCh38
rs756284974 Chr.16: 67184968-67184968 on Build GRCh38
rs757384868 Chr.16: 67184802-67184802 on Build GRCh38
rs16957212 Chr.16: 67184907-67184907 on Build GRCh38
rs996963313 Chr.16: 67184763-67184763 on Build GRCh38
rs779184172 Chr.16: 67184977-67184977 on Build GRCh38
rs763173796 Chr.16: 67185185-67185185 on Build GRCh38
rs759045894 Chr.16: 67184747-67184747 on Build GRCh38
rs899601213 Chr.16: 67184755-67184755 on Build GRCh38
rs745866906 Chr.16: 67185048-67185087 on Build GRCh38
rs781064445 Chr.16: 67184804-67184804 on Build GRCh38
rs757048997 Chr.16: 67184990-67184990 on Build GRCh38
rs899464688 Chr.16: 67185001-67185001 on Build GRCh38
rs762522248 Chr.16: 67184949-67184949 on Build GRCh38
rs200200061 Chr.16: 67184853-67184853 on Build GRCh38
rs775150481 Chr.16: 67184937-67184937 on Build GRCh38
rs968788367 Chr.16: 67185173-67185173 on Build GRCh38
rs201742331 Chr.16: 67184897-67184897 on Build GRCh38
rs764398849 Chr.16: 67184849-67184849 on Build GRCh38
rs376511701 Chr.16: 67185035-67185035 on Build GRCh38
rs962477857 Chr.16: 67184782-67184782 on Build GRCh38
rs1057362266 Chr.16: 67185203-67185203 on Build GRCh38
rs896250736 Chr.16: 67184761-67184761 on Build GRCh38
rs557233303 Chr.16: 67184834-67184834 on Build GRCh38
rs947504073 Chr.16: 67184929-67184929 on Build GRCh38
rs532889499 Chr.16: 67185088-67185088 on Build GRCh38
rs1025548072 Chr.16: 67184840-67184840 on Build GRCh38
rs986705197 Chr.16: 67185114-67185114 on Build GRCh38
rs9939768 Chr.16: 67185204-67185204 on Build GRCh38
rs748834819 Chr.16: 67185125-67185125 on Build GRCh38
rs568605749 Chr.16: 67184813-67184813 on Build GRCh38
rs1023439509 Chr.16: 67184770-67184770 on Build GRCh38
rs995337026 Chr.16: 67185060-67185060 on Build GRCh38
rs762132461 Chr.16: 67185052-67185052 on Build GRCh38
rs951295516 Chr.16: 67184842-67184842 on Build GRCh38
rs999633132 Chr.16: 67184742-67184742 on Build GRCh38
rs748779711 Chr.16: 67185029-67185029 on Build GRCh38
rs776629063 Chr.16: 67184905-67184905 on Build GRCh38
rs760497086 Chr.16: 67185225-67185239 on Build GRCh38
rs772202784 Chr.16: 67184707-67184707 on Build GRCh38
rs145474504 Chr.16: 67185149-67185149 on Build GRCh38
rs755779413 Chr.16: 67184789-67184797 on Build GRCh38
rs773886781 Chr.16: 67185062-67185062 on Build GRCh38
rs764453867 Chr.16: 67184950-67184950 on Build GRCh38
rs745426514 Chr.16: 67184906-67184906 on Build GRCh38
rs753725027 Chr.16: 67185213-67185213 on Build GRCh38
rs1010593199 Chr.16: 67184716-67184716 on Build GRCh38
rs374354242 Chr.16: 67184876-67184876 on Build GRCh38
rs750919533 Chr.16: 67184966-67184966 on Build GRCh38
rs761108975 Chr.16: 67185159-67185159 on Build GRCh38
rs766527114 Chr.16: 67184971-67184971 on Build GRCh38
rs369614815 Chr.16: 67184974-67184974 on Build GRCh38
rs751958003 Chr.16: 67184793-67184793 on Build GRCh38
rs766582125 Chr.16: 67185065-67185065 on Build GRCh38
rs62057953 Chr.16: 67185061-67185061 on Build GRCh38
rs745760978 Chr.16: 67185009-67185009 on Build GRCh38
rs142110646 Chr.16: 67184748-67184748 on Build GRCh38
rs748446668 Chr.16: 67184780-67184780 on Build GRCh38
rs541801424 Chr.16: 67184883-67184883 on Build GRCh38
rs757029621 Chr.16: 67185101-67185101 on Build GRCh38
rs371574544 Chr.16: 67185184-67185184 on Build GRCh38
rs774919316 Chr.16: 67185027-67185027 on Build GRCh38
rs752320613 Chr.16: 67184847-67184847 on Build GRCh38
rs753120174 Chr.16: 67184757-67184757 on Build GRCh38
rs763361020 Chr.16: 67184848-67184848 on Build GRCh38
rs772067952 Chr.16: 67185123-67185124 on Build GRCh38
rs928715461 Chr.16: 67184933-67184933 on Build GRCh38
rs762231652 Chr.16: 67184960-67184960 on Build GRCh38
rs148629602 Chr.16: 67185223-67185223 on Build GRCh38
rs964039669 Chr.16: 67185126-67185126 on Build GRCh38
rs201658978 Chr.16: 67185143-67185143 on Build GRCh38
rs562013805 Chr.16: 67184900-67184900 on Build GRCh38
rs777435483 Chr.16: 67184821-67184822 on Build GRCh38
rs201243773 Chr.16: 67185132-67185132 on Build GRCh38
rs1043403885 Chr.16: 67185000-67185000 on Build GRCh38
rs368539550 Chr.16: 67184877-67184877 on Build GRCh38
rs765757293 Chr.16: 67184714-67184714 on Build GRCh38
rs200521022 Chr.16: 67185166-67185166 on Build GRCh38
rs777248961 Chr.16: 67185180-67185180 on Build GRCh38
rs374156443 Chr.16: 67184708-67184708 on Build GRCh38
rs755869767 Chr.16: 67185198-67185198 on Build GRCh38
rs996521015 Chr.16: 67185019-67185019 on Build GRCh38
rs750370383 Chr.16: 67185102-67185102 on Build GRCh38
rs745542811 Chr.16: 67185156-67185156 on Build GRCh38
rs771969854 Chr.16: 67184825-67184825 on Build GRCh38
rs768257010 Chr.16: 67185030-67185030 on Build GRCh38
rs775436622 Chr.16: 67185171-67185171 on Build GRCh38
rs759076250 Chr.16: 67184843-67184843 on Build GRCh38
rs890861988 Chr.16: 67185032-67185032 on Build GRCh38
rs760489811 Chr.16: 67184832-67184832 on Build GRCh38
rs748782020 Chr.16: 67184901-67184901 on Build GRCh38
rs746111806 Chr.16: 67184810-67184810 on Build GRCh38
rs528877746 Chr.16: 67185187-67185187 on Build GRCh38
rs758761121 Chr.16: 67184762-67184762 on Build GRCh38

View Full Product Details