Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80975961-80976470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
UniGene
Hs.55028
Gene Chromosome Location
Chr.16: 80975800-81007046 on Build GRCh38
Transcripts
NM_020188.3 XM_017023461.1 XM_017023462.1 XM_017023464.1 XM_017023465.1 XM_017023466.1 XM_017023467.1 XM_017023468.1 XM_017023469.1 XM_017023470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426523 c.199C>T p.R67* Chr.16: 80976134-80976134 on Build GRCh38
4672996 c.204G>A p.K68K Chr.16: 80976129-80976129 on Build GRCh38
2692790 c.232G>A p.E78K Chr.16: 80976101-80976101 on Build GRCh38
6295411 c.165C>A p.N55K Chr.16: 80976168-80976168 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1042872463 Chr.16: 80975987-80975987 on Build GRCh38
rs1127390 Chr.16: 80976176-80976176 on Build GRCh38
rs141572999 Chr.16: 80976204-80976204 on Build GRCh38
rs1047263121 Chr.16: 80975997-80975997 on Build GRCh38
rs866043362 Chr.16: 80976135-80976135 on Build GRCh38
rs765082851 Chr.16: 80976195-80976196 on Build GRCh38
rs1032815242 Chr.16: 80976032-80976032 on Build GRCh38
rs750308084 Chr.16: 80976196-80976196 on Build GRCh38
rs746556209 Chr.16: 80976048-80976048 on Build GRCh38
rs1000984462 Chr.16: 80976234-80976234 on Build GRCh38
rs917883115 Chr.16: 80976300-80976300 on Build GRCh38
rs757550201 Chr.16: 80976224-80976224 on Build GRCh38
rs756225013 Chr.16: 80976089-80976089 on Build GRCh38
rs758141126 Chr.16: 80976228-80976228 on Build GRCh38
rs12446410 Chr.16: 80976218-80976218 on Build GRCh38
rs374458830 Chr.16: 80976198-80976198 on Build GRCh38
rs770537683 Chr.16: 80976314-80976314 on Build GRCh38
rs140656923 Chr.16: 80976103-80976103 on Build GRCh38
rs1000589395 Chr.16: 80976024-80976024 on Build GRCh38
rs999111472 Chr.16: 80976026-80976026 on Build GRCh38
rs554855954 Chr.16: 80976102-80976102 on Build GRCh38
rs776310897 Chr.16: 80976387-80976387 on Build GRCh38
rs775312252 Chr.16: 80976078-80976078 on Build GRCh38
rs541435706 Chr.16: 80976129-80976129 on Build GRCh38
rs868158782 Chr.16: 80976160-80976160 on Build GRCh38
rs745359055 Chr.16: 80976116-80976116 on Build GRCh38
rs780422536 Chr.16: 80976225-80976225 on Build GRCh38
rs964106909 Chr.16: 80976132-80976132 on Build GRCh38
rs765409124 Chr.16: 80976202-80976202 on Build GRCh38
rs527462893 Chr.16: 80976080-80976080 on Build GRCh38
rs758905230 Chr.16: 80976177-80976177 on Build GRCh38
rs569319798 Chr.16: 80976412-80976412 on Build GRCh38
rs751820411 Chr.16: 80976188-80976188 on Build GRCh38
rs369812890 Chr.16: 80976143-80976143 on Build GRCh38
rs375780601 Chr.16: 80976037-80976037 on Build GRCh38
rs7193361 Chr.16: 80976446-80976446 on Build GRCh38
rs72813721 Chr.16: 80976052-80976052 on Build GRCh38
rs761870147 Chr.16: 80976120-80976121 on Build GRCh38
rs746677657 Chr.16: 80976110-80976110 on Build GRCh38
rs769791284 Chr.16: 80976120-80976120 on Build GRCh38
rs1015230669 Chr.16: 80976376-80976376 on Build GRCh38
rs556077510 Chr.16: 80976291-80976291 on Build GRCh38
rs745442520 Chr.16: 80976195-80976195 on Build GRCh38
rs773980950 Chr.16: 80976144-80976144 on Build GRCh38
rs776226623 Chr.16: 80976115-80976115 on Build GRCh38
rs769056259 Chr.16: 80976251-80976251 on Build GRCh38
rs60961984 Chr.16: 80976305-80976306 on Build GRCh38
rs1045624033 Chr.16: 80976013-80976014 on Build GRCh38
rs759546954 Chr.16: 80976058-80976058 on Build GRCh38
rs200441699 Chr.16: 80976182-80976182 on Build GRCh38
rs563542917 Chr.16: 80976107-80976107 on Build GRCh38
rs1013786363 Chr.16: 80976007-80976007 on Build GRCh38
rs1027060298 Chr.16: 80976274-80976274 on Build GRCh38
rs143780675 Chr.16: 80976111-80976111 on Build GRCh38
rs200941646 Chr.16: 80976172-80976172 on Build GRCh38
rs955236394 Chr.16: 80976328-80976328 on Build GRCh38
rs772990731 Chr.16: 80976046-80976046 on Build GRCh38
rs961321460 Chr.16: 80976420-80976420 on Build GRCh38
rs764486190 Chr.16: 80976222-80976222 on Build GRCh38
rs759681083 Chr.16: 80976169-80976169 on Build GRCh38
rs576950576 Chr.16: 80975992-80975992 on Build GRCh38
rs1027142919 Chr.16: 80976448-80976448 on Build GRCh38
rs986430937 Chr.16: 80976137-80976137 on Build GRCh38
rs766514366 Chr.16: 80976091-80976091 on Build GRCh38
rs140972336 Chr.16: 80976168-80976168 on Build GRCh38
rs138381032 Chr.16: 80976199-80976199 on Build GRCh38
rs904960323 Chr.16: 80976015-80976015 on Build GRCh38
rs759772130 Chr.16: 80976200-80976200 on Build GRCh38
rs376325635 Chr.16: 80976101-80976101 on Build GRCh38
rs150075723 Chr.16: 80976019-80976019 on Build GRCh38
rs963142942 Chr.16: 80976259-80976259 on Build GRCh38
rs377531723 Chr.16: 80976192-80976192 on Build GRCh38
rs910351469 Chr.16: 80976105-80976105 on Build GRCh38
rs552966787 Chr.16: 80976309-80976309 on Build GRCh38
rs775393956 Chr.16: 80976133-80976133 on Build GRCh38
rs2303216 Chr.16: 80976273-80976273 on Build GRCh38
rs201310266 Chr.16: 80976125-80976125 on Build GRCh38
rs1007917468 Chr.16: 80976369-80976369 on Build GRCh38
rs2303215 Chr.16: 80976240-80976240 on Build GRCh38
rs748542413 Chr.16: 80976290-80976290 on Build GRCh38
rs200012173 Chr.16: 80976147-80976147 on Build GRCh38
rs753814170 Chr.16: 80976095-80976095 on Build GRCh38
rs763575438 Chr.16: 80976209-80976209 on Build GRCh38
rs13609 Chr.16: 80976138-80976138 on Build GRCh38
rs186471151 Chr.16: 80976086-80976086 on Build GRCh38
rs370096810 Chr.16: 80976197-80976197 on Build GRCh38
rs772781212 Chr.16: 80976284-80976284 on Build GRCh38
rs765110940 Chr.16: 80976068-80976068 on Build GRCh38
rs954773300 Chr.16: 80976079-80976079 on Build GRCh38
rs376834541 Chr.16: 80976053-80976053 on Build GRCh38
rs2303214 Chr.16: 80976002-80976002 on Build GRCh38
rs751993831 Chr.16: 80976223-80976223 on Build GRCh38
rs756277382 Chr.16: 80976196-80976196 on Build GRCh38
rs762821797 Chr.16: 80976134-80976134 on Build GRCh38

View Full Product Details