Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3880140-3880708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKK1
Entrez Gene ID
84254
Gene Name
calcium/calmodulin dependent protein kinase kinase 1
Gene Aliases
CAMKKA
UniGene
Hs.8417
Gene Chromosome Location
Chr.17: 3860315-3894891 on Build GRCh38
Transcripts
NM_032294.2 NM_172206.1 NM_172207.2 XM_005256817.1 XM_006721588.2 XM_011524031.2 XM_011524032.2 XM_011524033.1 XM_011524035.2 XM_017025219.1 XM_017025220.1 XM_017025221.1 XM_017025222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3937366 c.903C>T p.L301L Chr.17: 3880353-3880353 on Build GRCh38
4668962 c.711C>T p.P237P Chr.17: 3880431-3880431 on Build GRCh38
1588860 c.904G>A p.E302K Chr.17: 3880352-3880352 on Build GRCh38
6505822 c.832G>A p.E278K Chr.17: 3880424-3880424 on Build GRCh38
6436538 c.748G>T p.E250* Chr.17: 3880394-3880394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033163704 Chr.17: 3880479-3880480 on Build GRCh38
rs908377722 Chr.17: 3880357-3880357 on Build GRCh38
rs756188864 Chr.17: 3880361-3880361 on Build GRCh38
rs1013393920 Chr.17: 3880180-3880180 on Build GRCh38
rs545046675 Chr.17: 3880233-3880233 on Build GRCh38
rs150870972 Chr.17: 3880431-3880431 on Build GRCh38
rs1015736558 Chr.17: 3880524-3880524 on Build GRCh38
rs186035554 Chr.17: 3880338-3880338 on Build GRCh38
rs551437101 Chr.17: 3880456-3880456 on Build GRCh38
rs201476330 Chr.17: 3880423-3880423 on Build GRCh38
rs538754654 Chr.17: 3880638-3880638 on Build GRCh38
rs1024823556 Chr.17: 3880197-3880197 on Build GRCh38
rs761829053 Chr.17: 3880397-3880397 on Build GRCh38
rs548689848 Chr.17: 3880168-3880168 on Build GRCh38
rs773590248 Chr.17: 3880563-3880563 on Build GRCh38
rs563467705 Chr.17: 3880321-3880321 on Build GRCh38
rs769375322 Chr.17: 3880296-3880296 on Build GRCh38
rs886264434 Chr.17: 3880484-3880484 on Build GRCh38
rs189793103 Chr.17: 3880667-3880667 on Build GRCh38
rs761162696 Chr.17: 3880384-3880384 on Build GRCh38
rs769959072 Chr.17: 3880499-3880499 on Build GRCh38
rs572879316 Chr.17: 3880187-3880187 on Build GRCh38
rs769028318 Chr.17: 3880441-3880441 on Build GRCh38
rs973906117 Chr.17: 3880566-3880566 on Build GRCh38
rs531189416 Chr.17: 3880510-3880510 on Build GRCh38
rs374754501 Chr.17: 3880451-3880451 on Build GRCh38
rs752475218 Chr.17: 3880413-3880413 on Build GRCh38
rs1050483211 Chr.17: 3880274-3880274 on Build GRCh38
rs754737840 Chr.17: 3880353-3880353 on Build GRCh38
rs963024025 Chr.17: 3880257-3880257 on Build GRCh38
rs985309491 Chr.17: 3880583-3880583 on Build GRCh38
rs760015698 Chr.17: 3880617-3880617 on Build GRCh38
rs11651131 Chr.17: 3880578-3880578 on Build GRCh38
rs140888315 Chr.17: 3880293-3880293 on Build GRCh38
rs141585801 Chr.17: 3880398-3880398 on Build GRCh38
rs1013173759 Chr.17: 3880486-3880486 on Build GRCh38
rs939986205 Chr.17: 3880193-3880193 on Build GRCh38
rs752669927 Chr.17: 3880356-3880356 on Build GRCh38
rs747419940 Chr.17: 3880440-3880440 on Build GRCh38
rs1054671464 Chr.17: 3880646-3880646 on Build GRCh38
rs978248388 Chr.17: 3880666-3880666 on Build GRCh38
rs375338007 Chr.17: 3880483-3880483 on Build GRCh38
rs998993189 Chr.17: 3880468-3880468 on Build GRCh38
rs753443637 Chr.17: 3880167-3880167 on Build GRCh38
rs775999357 Chr.17: 3880318-3880318 on Build GRCh38
rs771424249 Chr.17: 3880605-3880605 on Build GRCh38
rs775775222 Chr.17: 3880379-3880379 on Build GRCh38
rs756101063 Chr.17: 3880415-3880415 on Build GRCh38
rs370720296 Chr.17: 3880337-3880337 on Build GRCh38
rs1046387698 Chr.17: 3880481-3880481 on Build GRCh38
rs903318131 Chr.17: 3880300-3880300 on Build GRCh38
rs769187818 Chr.17: 3880385-3880385 on Build GRCh38
rs910021143 Chr.17: 3880241-3880241 on Build GRCh38
rs777484107 Chr.17: 3880304-3880304 on Build GRCh38
rs889229968 Chr.17: 3880280-3880280 on Build GRCh38
rs148366995 Chr.17: 3880394-3880394 on Build GRCh38
rs770749532 Chr.17: 3880308-3880308 on Build GRCh38
rs753981187 Chr.17: 3880327-3880327 on Build GRCh38
rs759894076 Chr.17: 3880312-3880312 on Build GRCh38
rs973979355 Chr.17: 3880380-3880380 on Build GRCh38
rs748229139 Chr.17: 3880457-3880457 on Build GRCh38
rs368864344 Chr.17: 3880458-3880458 on Build GRCh38
rs577826400 Chr.17: 3880279-3880279 on Build GRCh38
rs773236375 Chr.17: 3880453-3880453 on Build GRCh38
rs994772403 Chr.17: 3880234-3880234 on Build GRCh38
rs766558958 Chr.17: 3880477-3880477 on Build GRCh38
rs138395664 Chr.17: 3880570-3880570 on Build GRCh38
rs750605303 Chr.17: 3880399-3880399 on Build GRCh38
rs761990301 Chr.17: 3880330-3880330 on Build GRCh38
rs777059654 Chr.17: 3880447-3880447 on Build GRCh38
rs778586310 Chr.17: 3880371-3880371 on Build GRCh38
rs879279060 Chr.17: 3880576-3880576 on Build GRCh38
rs75354830 Chr.17: 3880175-3880175 on Build GRCh38
rs763146758 Chr.17: 3880400-3880400 on Build GRCh38
rs971438314 Chr.17: 3880504-3880504 on Build GRCh38
rs772423159 Chr.17: 3880376-3880376 on Build GRCh38
rs1016054041 Chr.17: 3880255-3880255 on Build GRCh38
rs566663063 Chr.17: 3880614-3880614 on Build GRCh38
rs767340323 Chr.17: 3880406-3880406 on Build GRCh38
rs777386537 Chr.17: 3880364-3880364 on Build GRCh38
rs950409710 Chr.17: 3880243-3880243 on Build GRCh38
rs770588197 Chr.17: 3880367-3880367 on Build GRCh38
rs903293628 Chr.17: 3880630-3880630 on Build GRCh38
rs745519128 Chr.17: 3880430-3880430 on Build GRCh38
rs745510115 Chr.17: 3880373-3880373 on Build GRCh38
rs753857143 Chr.17: 3880427-3880427 on Build GRCh38
rs1019853117 Chr.17: 3880625-3880625 on Build GRCh38
rs748858266 Chr.17: 3880365-3880365 on Build GRCh38
rs111961237 Chr.17: 3880517-3880517 on Build GRCh38
rs774796201 Chr.17: 3880608-3880608 on Build GRCh38
rs150158165 Chr.17: 3880450-3880450 on Build GRCh38
rs772352954 Chr.17: 3880317-3880317 on Build GRCh38
rs764141524 Chr.17: 3880323-3880323 on Build GRCh38
rs780431300 Chr.17: 3880438-3880438 on Build GRCh38
rs910554585 Chr.17: 3880595-3880595 on Build GRCh38
rs774558597 Chr.17: 3880311-3880311 on Build GRCh38
rs778299550 Chr.17: 3880429-3880429 on Build GRCh38
rs567832324 Chr.17: 3880567-3880567 on Build GRCh38
rs980275078 Chr.17: 3880191-3880191 on Build GRCh38
rs557618641 Chr.17: 3880273-3880273 on Build GRCh38
rs757079141 Chr.17: 3880292-3880292 on Build GRCh38
rs567767725 Chr.17: 3880163-3880163 on Build GRCh38
rs1038215509 Chr.17: 3880193-3880194 on Build GRCh38
rs1004631079 Chr.17: 3880244-3880244 on Build GRCh38

View Full Product Details