Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 31305183-31305684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI2B
Entrez Gene ID
2124
Gene Name
ecotropic viral integration site 2B
Gene Aliases
CD361, D17S376, EVDB
UniGene
Hs.5509
Gene Chromosome Location
Chr.17: 31303766-31314149 on Build GRCh38
Transcripts
NM_006495.3 XM_005257946.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
UniGene
Hs.113577
Gene Chromosome Location
Chr.17: 31094927-31377677 on Build GRCh38
Transcripts
NM_006495.3 XM_005257946.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1316086 c.413C>A p.P138Q Chr.17: 31305197-31305197 on Build GRCh38
6656883 c.189C>T p.S63S Chr.17: 31305421-31305421 on Build GRCh38
4544515 c.358G>A p.A120T Chr.17: 31305252-31305252 on Build GRCh38
4782084 c.465G>T p.K155N Chr.17: 31305190-31305190 on Build GRCh38
4172206 c.53C>A p.T18K Chr.17: 31305557-31305557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201399165 Chr.17: 31305334-31305334 on Build GRCh38
rs776156138 Chr.17: 31305287-31305287 on Build GRCh38
rs941071081 Chr.17: 31305558-31305558 on Build GRCh38
rs777799482 Chr.17: 31305638-31305638 on Build GRCh38
rs151096117 Chr.17: 31305421-31305421 on Build GRCh38
rs1023260730 Chr.17: 31305278-31305278 on Build GRCh38
rs929602758 Chr.17: 31305410-31305410 on Build GRCh38
rs756983686 Chr.17: 31305268-31305268 on Build GRCh38
rs368216496 Chr.17: 31305474-31305474 on Build GRCh38
rs767214948 Chr.17: 31305461-31305461 on Build GRCh38
rs766073949 Chr.17: 31305216-31305216 on Build GRCh38
rs764127894 Chr.17: 31305447-31305447 on Build GRCh38
rs754101275 Chr.17: 31305625-31305625 on Build GRCh38
rs185652683 Chr.17: 31305401-31305401 on Build GRCh38
rs921186585 Chr.17: 31305622-31305622 on Build GRCh38
rs750146352 Chr.17: 31305590-31305591 on Build GRCh38
rs144364775 Chr.17: 31305253-31305253 on Build GRCh38
rs760956267 Chr.17: 31305440-31305440 on Build GRCh38
rs778827046 Chr.17: 31305417-31305417 on Build GRCh38
rs780540629 Chr.17: 31305491-31305491 on Build GRCh38
rs748117738 Chr.17: 31305536-31305536 on Build GRCh38
rs753236143 Chr.17: 31305637-31305637 on Build GRCh38
rs765186864 Chr.17: 31305238-31305238 on Build GRCh38
rs757911020 Chr.17: 31305249-31305249 on Build GRCh38
rs748756060 Chr.17: 31305309-31305309 on Build GRCh38
rs569234254 Chr.17: 31305317-31305317 on Build GRCh38
rs564349361 Chr.17: 31305554-31305554 on Build GRCh38
rs780923966 Chr.17: 31305377-31305377 on Build GRCh38
rs750989438 Chr.17: 31305449-31305449 on Build GRCh38
rs746629693 Chr.17: 31305255-31305255 on Build GRCh38
rs576969125 Chr.17: 31305557-31305557 on Build GRCh38
rs374306730 Chr.17: 31305533-31305533 on Build GRCh38
rs138554367 Chr.17: 31305223-31305223 on Build GRCh38
rs755399921 Chr.17: 31305506-31305506 on Build GRCh38
rs774439090 Chr.17: 31305211-31305211 on Build GRCh38
rs572332677 Chr.17: 31305215-31305215 on Build GRCh38
rs1029616866 Chr.17: 31305289-31305289 on Build GRCh38
rs903105392 Chr.17: 31305581-31305581 on Build GRCh38
rs751442776 Chr.17: 31305411-31305412 on Build GRCh38
rs766161104 Chr.17: 31305623-31305623 on Build GRCh38
rs374849042 Chr.17: 31305633-31305633 on Build GRCh38
rs531300031 Chr.17: 31305636-31305636 on Build GRCh38
rs747123373 Chr.17: 31305641-31305641 on Build GRCh38
rs762833164 Chr.17: 31305565-31305565 on Build GRCh38
rs774353882 Chr.17: 31305567-31305567 on Build GRCh38
rs9903564 Chr.17: 31305453-31305453 on Build GRCh38
rs762769586 Chr.17: 31305444-31305444 on Build GRCh38
rs752151189 Chr.17: 31305241-31305241 on Build GRCh38
rs766333180 Chr.17: 31305260-31305261 on Build GRCh38
rs16972176 Chr.17: 31305646-31305646 on Build GRCh38
rs991622786 Chr.17: 31305337-31305337 on Build GRCh38
rs77954806 Chr.17: 31305647-31305647 on Build GRCh38
rs777889615 Chr.17: 31305556-31305556 on Build GRCh38
rs754289943 Chr.17: 31305492-31305492 on Build GRCh38
rs999235860 Chr.17: 31305237-31305237 on Build GRCh38
rs375405082 Chr.17: 31305489-31305489 on Build GRCh38
rs768610622 Chr.17: 31305292-31305292 on Build GRCh38
rs144434599 Chr.17: 31305404-31305404 on Build GRCh38
rs774543831 Chr.17: 31305296-31305296 on Build GRCh38
rs771183458 Chr.17: 31305559-31305559 on Build GRCh38
rs764811466 Chr.17: 31305356-31305356 on Build GRCh38
rs373866701 Chr.17: 31305399-31305399 on Build GRCh38
rs566803471 Chr.17: 31305420-31305420 on Build GRCh38
rs775314294 Chr.17: 31305441-31305441 on Build GRCh38
rs760368575 Chr.17: 31305620-31305620 on Build GRCh38
rs781384375 Chr.17: 31305272-31305272 on Build GRCh38
rs180817786 Chr.17: 31305291-31305291 on Build GRCh38
rs763490665 Chr.17: 31305352-31305352 on Build GRCh38
rs772378094 Chr.17: 31305570-31305570 on Build GRCh38
rs532544954 Chr.17: 31305275-31305275 on Build GRCh38
rs866847971 Chr.17: 31305390-31305390 on Build GRCh38
rs371540689 Chr.17: 31305385-31305385 on Build GRCh38
rs749998773 Chr.17: 31305618-31305618 on Build GRCh38
rs768424280 Chr.17: 31305566-31305566 on Build GRCh38
rs772195960 Chr.17: 31305540-31305540 on Build GRCh38
rs767820560 Chr.17: 31305300-31305300 on Build GRCh38
rs773434316 Chr.17: 31305431-31305431 on Build GRCh38
rs770379408 Chr.17: 31305281-31305281 on Build GRCh38
rs1050900648 Chr.17: 31305628-31305628 on Build GRCh38
rs751120621 Chr.17: 31305360-31305360 on Build GRCh38
rs749796377 Chr.17: 31305412-31305412 on Build GRCh38
rs769251597 Chr.17: 31305415-31305415 on Build GRCh38
rs373785777 Chr.17: 31305318-31305318 on Build GRCh38
rs761914010 Chr.17: 31305297-31305297 on Build GRCh38
rs868808379 Chr.17: 31305323-31305323 on Build GRCh38
rs141059622 Chr.17: 31305626-31305626 on Build GRCh38
rs370014149 Chr.17: 31305597-31305597 on Build GRCh38
rs200921973 Chr.17: 31305562-31305562 on Build GRCh38
rs756823393 Chr.17: 31305363-31305363 on Build GRCh38
rs190016033 Chr.17: 31305658-31305658 on Build GRCh38
rs371465687 Chr.17: 31305445-31305445 on Build GRCh38

View Full Product Details