Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60679294-60679701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140395 c.3G>T p.M1I Chr.17: 60679460-60679460 on Build GRCh38
4269592 c.35G>A p.R12K Chr.17: 60679492-60679492 on Build GRCh38
4269593 c.83+1G>A p.? Chr.17: 60679541-60679541 on Build GRCh38
4665353 c.64G>A p.V22I Chr.17: 60679521-60679521 on Build GRCh38
7140394 c.3G>T p.M1I Chr.17: 60679460-60679460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577397534 Chr.17: 60679565-60679565 on Build GRCh38
rs148962787 Chr.17: 60679610-60679610 on Build GRCh38
rs555003474 Chr.17: 60679365-60679365 on Build GRCh38
rs766607946 Chr.17: 60679501-60679503 on Build GRCh38
rs528509134 Chr.17: 60679416-60679416 on Build GRCh38
rs780307553 Chr.17: 60679411-60679411 on Build GRCh38
rs775011218 Chr.17: 60679444-60679444 on Build GRCh38
rs192677054 Chr.17: 60679590-60679590 on Build GRCh38
rs779478430 Chr.17: 60679579-60679579 on Build GRCh38
rs370101729 Chr.17: 60679540-60679540 on Build GRCh38
rs760252824 Chr.17: 60679459-60679459 on Build GRCh38
rs765556399 Chr.17: 60679501-60679501 on Build GRCh38
rs371277552 Chr.17: 60679523-60679523 on Build GRCh38
rs1002048867 Chr.17: 60679611-60679611 on Build GRCh38
rs768113757 Chr.17: 60679591-60679591 on Build GRCh38
rs772000827 Chr.17: 60679567-60679567 on Build GRCh38
rs758533729 Chr.17: 60679408-60679408 on Build GRCh38
rs1021297327 Chr.17: 60679677-60679677 on Build GRCh38
rs762229492 Chr.17: 60679483-60679483 on Build GRCh38
rs770699470 Chr.17: 60679429-60679429 on Build GRCh38
rs997333231 Chr.17: 60679602-60679602 on Build GRCh38
rs778701813 Chr.17: 60679559-60679559 on Build GRCh38
rs955829791 Chr.17: 60679622-60679622 on Build GRCh38
rs970153444 Chr.17: 60679596-60679596 on Build GRCh38
rs55807727 Chr.17: 60679638-60679638 on Build GRCh38
rs748359271 Chr.17: 60679527-60679527 on Build GRCh38
rs1007480337 Chr.17: 60679462-60679462 on Build GRCh38
rs781473256 Chr.17: 60679526-60679526 on Build GRCh38
rs1031447772 Chr.17: 60679605-60679605 on Build GRCh38
rs376050639 Chr.17: 60679443-60679443 on Build GRCh38
rs145551547 Chr.17: 60679425-60679425 on Build GRCh38
rs943127753 Chr.17: 60679379-60679379 on Build GRCh38
rs751704466 Chr.17: 60679524-60679524 on Build GRCh38
rs778314330 Chr.17: 60679560-60679560 on Build GRCh38
rs887513530 Chr.17: 60679366-60679366 on Build GRCh38
rs562085506 Chr.17: 60679663-60679663 on Build GRCh38
rs763390916 Chr.17: 60679436-60679437 on Build GRCh38
rs766502815 Chr.17: 60679521-60679521 on Build GRCh38
rs751033594 Chr.17: 60679407-60679407 on Build GRCh38
rs77869658 Chr.17: 60679426-60679426 on Build GRCh38
rs1049371766 Chr.17: 60679361-60679361 on Build GRCh38
rs763818286 Chr.17: 60679469-60679469 on Build GRCh38
rs919367157 Chr.17: 60679390-60679390 on Build GRCh38
rs773898879 Chr.17: 60679425-60679428 on Build GRCh38
rs767093331 Chr.17: 60679344-60679344 on Build GRCh38
rs553962227 Chr.17: 60679428-60679428 on Build GRCh38
rs776202684 Chr.17: 60679477-60679477 on Build GRCh38
rs750914642 Chr.17: 60679509-60679509 on Build GRCh38
rs755073133 Chr.17: 60679525-60679525 on Build GRCh38
rs759009633 Chr.17: 60679516-60679516 on Build GRCh38
rs747196760 Chr.17: 60679418-60679418 on Build GRCh38
rs774315088 Chr.17: 60679441-60679441 on Build GRCh38

View Full Product Details