Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68536772-68537330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20A
Entrez Gene ID
54757
Gene Name
family with sequence similarity 20 member A
Gene Aliases
AI1G, AIGFS, FP2747
UniGene
Hs.268874
Gene Chromosome Location
Chr.17: 68534561-68600954 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Gene Symbol
PRKAR1A
Entrez Gene ID
5573
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Gene Aliases
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1
UniGene
Hs.280342
Gene Chromosome Location
Chr.17: 68413623-68551316 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

SNP Detail

SNP ID Chromosome Location
rs145648532 Chr.17: 68537043-68537043 on Build GRCh38
rs1030380326 Chr.17: 68537258-68537258 on Build GRCh38
rs967512113 Chr.17: 68536881-68536881 on Build GRCh38
rs992338334 Chr.17: 68537023-68537023 on Build GRCh38
rs754267824 Chr.17: 68537029-68537029 on Build GRCh38
rs1030538740 Chr.17: 68536953-68536953 on Build GRCh38
rs765464156 Chr.17: 68537140-68537140 on Build GRCh38
rs770857622 Chr.17: 68537309-68537309 on Build GRCh38
rs576982595 Chr.17: 68536814-68536814 on Build GRCh38
rs775273366 Chr.17: 68537293-68537293 on Build GRCh38
rs372549540 Chr.17: 68537107-68537107 on Build GRCh38
rs1052571732 Chr.17: 68537100-68537100 on Build GRCh38
rs747312936 Chr.17: 68537213-68537213 on Build GRCh38
rs763764612 Chr.17: 68537307-68537307 on Build GRCh38
rs761191418 Chr.17: 68536906-68536906 on Build GRCh38
rs559260402 Chr.17: 68536861-68536861 on Build GRCh38
rs2286554 Chr.17: 68537267-68537267 on Build GRCh38
rs903174373 Chr.17: 68537215-68537215 on Build GRCh38
rs925523760 Chr.17: 68537084-68537084 on Build GRCh38
rs944107686 Chr.17: 68537227-68537227 on Build GRCh38
rs753365633 Chr.17: 68537062-68537062 on Build GRCh38
rs1002086090 Chr.17: 68536818-68536818 on Build GRCh38
rs894484512 Chr.17: 68536838-68536838 on Build GRCh38
rs1009574930 Chr.17: 68537244-68537244 on Build GRCh38
rs149978140 Chr.17: 68536833-68536833 on Build GRCh38
rs530634351 Chr.17: 68537091-68537091 on Build GRCh38
rs1029305933 Chr.17: 68536885-68536885 on Build GRCh38
rs760322499 Chr.17: 68537047-68537047 on Build GRCh38
rs929093989 Chr.17: 68537061-68537061 on Build GRCh38
rs1021666581 Chr.17: 68537209-68537209 on Build GRCh38
rs996885884 Chr.17: 68536882-68536882 on Build GRCh38
rs758933279 Chr.17: 68537143-68537143 on Build GRCh38
rs768997260 Chr.17: 68536887-68536887 on Build GRCh38
rs767272967 Chr.17: 68537012-68537012 on Build GRCh38
rs774195501 Chr.17: 68536999-68536999 on Build GRCh38
rs781515996 Chr.17: 68537231-68537231 on Build GRCh38
rs535802048 Chr.17: 68537281-68537281 on Build GRCh38
rs1022028962 Chr.17: 68536877-68536877 on Build GRCh38
rs954824427 Chr.17: 68536965-68536965 on Build GRCh38
rs188485434 Chr.17: 68537122-68537122 on Build GRCh38
rs773694730 Chr.17: 68536807-68536807 on Build GRCh38
rs563178726 Chr.17: 68537085-68537085 on Build GRCh38
rs535079025 Chr.17: 68537208-68537208 on Build GRCh38
rs554981211 Chr.17: 68536966-68536966 on Build GRCh38
rs142622721 Chr.17: 68537280-68537280 on Build GRCh38
rs762912027 Chr.17: 68537231-68537233 on Build GRCh38
rs1046099326 Chr.17: 68537066-68537066 on Build GRCh38
rs150972449 Chr.17: 68537284-68537284 on Build GRCh38
rs557669191 Chr.17: 68537285-68537285 on Build GRCh38
rs747296601 Chr.17: 68536853-68536853 on Build GRCh38
rs986327576 Chr.17: 68537283-68537283 on Build GRCh38
rs952001488 Chr.17: 68536903-68536903 on Build GRCh38
rs369794249 Chr.17: 68537049-68537049 on Build GRCh38
rs751494832 Chr.17: 68536827-68536827 on Build GRCh38
rs748349083 Chr.17: 68537242-68537242 on Build GRCh38
rs1019587313 Chr.17: 68536859-68536859 on Build GRCh38
rs752878094 Chr.17: 68536914-68536914 on Build GRCh38
rs1054977677 Chr.17: 68537102-68537102 on Build GRCh38
rs769013630 Chr.17: 68537230-68537230 on Build GRCh38
rs778459468 Chr.17: 68537090-68537090 on Build GRCh38
rs918917241 Chr.17: 68537045-68537045 on Build GRCh38
rs764554217 Chr.17: 68537011-68537011 on Build GRCh38
rs984579561 Chr.17: 68536980-68536980 on Build GRCh38
rs770924714 Chr.17: 68536991-68536991 on Build GRCh38
rs998947356 Chr.17: 68537254-68537254 on Build GRCh38
rs756686151 Chr.17: 68537069-68537069 on Build GRCh38

View Full Product Details