Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 66211877-66212160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOH
Entrez Gene ID
350
Gene Name
apolipoprotein H
Gene Aliases
B2G1, B2GP1, BG
UniGene
Hs.445358
Gene Chromosome Location
Chr.17: 66212029-66229438 on Build GRCh38
Transcripts
NM_000042.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6532535 c.1035C>G p.C345W Chr.17: 66212136-66212136 on Build GRCh38
6147558 c.1011T>G p.T337T Chr.17: 66212160-66212160 on Build GRCh38
3521077 c.1021G>A p.D341N Chr.17: 66212150-66212150 on Build GRCh38
4580000 c.1032A>G p.P344P Chr.17: 66212139-66212139 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932841813 Chr.17: 66212042-66212042 on Build GRCh38
rs541084675 Chr.17: 66212087-66212087 on Build GRCh38
rs113041144 Chr.17: 66212031-66212031 on Build GRCh38
rs982233919 Chr.17: 66211958-66211958 on Build GRCh38
rs780480286 Chr.17: 66212105-66212105 on Build GRCh38
rs750997155 Chr.17: 66212091-66212091 on Build GRCh38
rs562833065 Chr.17: 66212133-66212133 on Build GRCh38
rs769938315 Chr.17: 66211915-66211915 on Build GRCh38
rs775417167 Chr.17: 66212139-66212139 on Build GRCh38
rs139819858 Chr.17: 66211900-66211900 on Build GRCh38
rs916755176 Chr.17: 66211946-66211946 on Build GRCh38
rs774235022 Chr.17: 66212130-66212130 on Build GRCh38
rs748272877 Chr.17: 66212114-66212114 on Build GRCh38
rs1015672775 Chr.17: 66212106-66212106 on Build GRCh38
rs570681722 Chr.17: 66211935-66211935 on Build GRCh38
rs113813266 Chr.17: 66212028-66212028 on Build GRCh38
rs369364551 Chr.17: 66212016-66212016 on Build GRCh38
rs920547287 Chr.17: 66212061-66212061 on Build GRCh38
rs112890054 Chr.17: 66212032-66212032 on Build GRCh38
rs369004865 Chr.17: 66211954-66211954 on Build GRCh38
rs771800586 Chr.17: 66212136-66212136 on Build GRCh38
rs373238657 Chr.17: 66211982-66211982 on Build GRCh38
rs190137879 Chr.17: 66212111-66212111 on Build GRCh38
rs749081636 Chr.17: 66211957-66211957 on Build GRCh38
rs8178951 Chr.17: 66212071-66212071 on Build GRCh38
rs902827652 Chr.17: 66212132-66212132 on Build GRCh38
rs199553389 Chr.17: 66212120-66212120 on Build GRCh38
rs752992912 Chr.17: 66212036-66212036 on Build GRCh38
rs758767603 Chr.17: 66212095-66212095 on Build GRCh38
rs940726390 Chr.17: 66212002-66212002 on Build GRCh38
rs1041496447 Chr.17: 66212126-66212126 on Build GRCh38
rs561622404 Chr.17: 66212030-66212030 on Build GRCh38
rs6933 Chr.17: 66212112-66212112 on Build GRCh38
rs991018771 Chr.17: 66211919-66211919 on Build GRCh38
rs769883181 Chr.17: 66212128-66212128 on Build GRCh38
rs572178041 Chr.17: 66212076-66212076 on Build GRCh38

View Full Product Details