Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64502205-64502709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX5
Entrez Gene ID
1655
Gene Name
DEAD-box helicase 5
Gene Aliases
G17P1, HLR1, HUMP68, p68
UniGene
Hs.279806
Gene Chromosome Location
Chr.17: 64498254-64507038 on Build GRCh38
Transcripts
NM_001320595.1 NM_001320596.1 NM_001320597.1 NM_004396.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2796188 c.1028A>C p.K343T Chr.17: 64502505-64502505 on Build GRCh38
5449025 c.991C>T p.R331C Chr.17: 64502542-64502542 on Build GRCh38
7349705 c.1076G>C p.R359T Chr.17: 64502457-64502457 on Build GRCh38
6641889 c.1010T>C p.M337T Chr.17: 64502523-64502523 on Build GRCh38
3742389 c.1105A>G p.M369V Chr.17: 64502213-64502213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782008166 Chr.17: 64502580-64502580 on Build GRCh38
rs781929574 Chr.17: 64502241-64502241 on Build GRCh38
rs781897802 Chr.17: 64502553-64502553 on Build GRCh38
rs544762345 Chr.17: 64502519-64502519 on Build GRCh38
rs782326949 Chr.17: 64502454-64502454 on Build GRCh38
rs782213689 Chr.17: 64502560-64502560 on Build GRCh38
rs573496167 Chr.17: 64502408-64502408 on Build GRCh38
rs782418919 Chr.17: 64502566-64502566 on Build GRCh38
rs1026829610 Chr.17: 64502336-64502336 on Build GRCh38
rs782647646 Chr.17: 64502559-64502559 on Build GRCh38
rs782721045 Chr.17: 64502262-64502262 on Build GRCh38
rs1002450072 Chr.17: 64502274-64502274 on Build GRCh38
rs782016494 Chr.17: 64502456-64502456 on Build GRCh38
rs540494151 Chr.17: 64502579-64502579 on Build GRCh38
rs782064342 Chr.17: 64502388-64502388 on Build GRCh38
rs781820615 Chr.17: 64502404-64502404 on Build GRCh38
rs782093380 Chr.17: 64502435-64502435 on Build GRCh38
rs782744419 Chr.17: 64502592-64502592 on Build GRCh38
rs781953419 Chr.17: 64502424-64502424 on Build GRCh38
rs781790471 Chr.17: 64502593-64502593 on Build GRCh38
rs782125498 Chr.17: 64502259-64502259 on Build GRCh38
rs782251343 Chr.17: 64502287-64502287 on Build GRCh38
rs782760916 Chr.17: 64502403-64502404 on Build GRCh38
rs782185827 Chr.17: 64502407-64502407 on Build GRCh38
rs782801888 Chr.17: 64502545-64502545 on Build GRCh38
rs782274176 Chr.17: 64502564-64502564 on Build GRCh38
rs140855073 Chr.17: 64502303-64502303 on Build GRCh38
rs782440358 Chr.17: 64502544-64502544 on Build GRCh38
rs781795658 Chr.17: 64502541-64502541 on Build GRCh38
rs769020 Chr.17: 64502448-64502448 on Build GRCh38
rs782488375 Chr.17: 64502562-64502562 on Build GRCh38
rs376858379 Chr.17: 64502405-64502405 on Build GRCh38
rs896706310 Chr.17: 64502623-64502623 on Build GRCh38
rs782163985 Chr.17: 64502236-64502236 on Build GRCh38
rs782387257 Chr.17: 64502420-64502420 on Build GRCh38
rs149936634 Chr.17: 64502567-64502568 on Build GRCh38
rs113053030 Chr.17: 64502436-64502436 on Build GRCh38
rs1043447663 Chr.17: 64502604-64502604 on Build GRCh38
rs782157394 Chr.17: 64502581-64502581 on Build GRCh38
rs782090087 Chr.17: 64502529-64502529 on Build GRCh38
rs781791351 Chr.17: 64502387-64502387 on Build GRCh38
rs373951221 Chr.17: 64502258-64502258 on Build GRCh38
rs782347414 Chr.17: 64502576-64502576 on Build GRCh38
rs782767557 Chr.17: 64502510-64502510 on Build GRCh38
rs371960655 Chr.17: 64502599-64502599 on Build GRCh38
rs187153242 Chr.17: 64502607-64502607 on Build GRCh38
rs112043743 Chr.17: 64502351-64502351 on Build GRCh38
rs781828510 Chr.17: 64502402-64502402 on Build GRCh38
rs113314395 Chr.17: 64502242-64502242 on Build GRCh38
rs985196446 Chr.17: 64502372-64502372 on Build GRCh38
rs782617455 Chr.17: 64502234-64502234 on Build GRCh38
rs782283153 Chr.17: 64502326-64502326 on Build GRCh38
rs531211772 Chr.17: 64502662-64502662 on Build GRCh38
rs994090132 Chr.17: 64502646-64502646 on Build GRCh38
rs782470159 Chr.17: 64502588-64502588 on Build GRCh38
rs781868095 Chr.17: 64502583-64502583 on Build GRCh38
rs559023447 Chr.17: 64502371-64502371 on Build GRCh38
rs547744637 Chr.17: 64502664-64502664 on Build GRCh38
rs552443191 Chr.17: 64502639-64502639 on Build GRCh38
rs368650652 Chr.17: 64502558-64502558 on Build GRCh38
rs782245917 Chr.17: 64502417-64502417 on Build GRCh38
rs577084333 Chr.17: 64502353-64502353 on Build GRCh38
rs782625600 Chr.17: 64502557-64502557 on Build GRCh38
rs1035257091 Chr.17: 64502616-64502616 on Build GRCh38
rs782170028 Chr.17: 64502507-64502507 on Build GRCh38
rs910516987 Chr.17: 64502397-64502397 on Build GRCh38
rs782678125 Chr.17: 64502412-64502412 on Build GRCh38
rs782230372 Chr.17: 64502237-64502237 on Build GRCh38
rs782684539 Chr.17: 64502321-64502321 on Build GRCh38
rs1052254271 Chr.17: 64502571-64502571 on Build GRCh38
rs782268130 Chr.17: 64502570-64502570 on Build GRCh38
rs764832780 Chr.17: 64502560-64502563 on Build GRCh38
rs782755413 Chr.17: 64502537-64502537 on Build GRCh38
rs964670744 Chr.17: 64502672-64502672 on Build GRCh38
rs781910026 Chr.17: 64502393-64502393 on Build GRCh38
rs139129097 Chr.17: 64502248-64502251 on Build GRCh38
rs782510105 Chr.17: 64502398-64502398 on Build GRCh38
rs972202100 Chr.17: 64502327-64502327 on Build GRCh38
rs781988355 Chr.17: 64502569-64502569 on Build GRCh38
rs782503163 Chr.17: 64502404-64502404 on Build GRCh38
rs904858179 Chr.17: 64502605-64502605 on Build GRCh38
rs1026440541 Chr.17: 64502647-64502647 on Build GRCh38
rs782656768 Chr.17: 64502400-64502400 on Build GRCh38
rs782307670 Chr.17: 64502240-64502240 on Build GRCh38
rs182117980 Chr.17: 64502370-64502370 on Build GRCh38
rs782391627 Chr.17: 64502685-64502685 on Build GRCh38
rs561370578 Chr.17: 64502563-64502563 on Build GRCh38
rs562194842 Chr.17: 64502658-64502658 on Build GRCh38
rs782457336 Chr.17: 64502379-64502379 on Build GRCh38
rs922543187 Chr.17: 64502688-64502688 on Build GRCh38
rs374155557 Chr.17: 64502578-64502578 on Build GRCh38

View Full Product Details