Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5415932-5416526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709323 c.1686G>T p.G562G Chr.18: 5416199-5416199 on Build GRCh38
363785 c.1684G>A p.G562R Chr.18: 5416201-5416201 on Build GRCh38
1640997 c.1622G>C p.G541A Chr.18: 5416263-5416263 on Build GRCh38
4897171 c.1794C>G p.S598S Chr.18: 5416091-5416091 on Build GRCh38
7216099 c.1724A>G p.Y575C Chr.18: 5416161-5416161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748081713 Chr.18: 5416328-5416328 on Build GRCh38
rs111274175 Chr.18: 5416319-5416319 on Build GRCh38
rs778849719 Chr.18: 5416322-5416322 on Build GRCh38
rs36014038 Chr.18: 5416088-5416088 on Build GRCh38
rs756935985 Chr.18: 5416202-5416202 on Build GRCh38
rs745854933 Chr.18: 5416002-5416002 on Build GRCh38
rs201925314 Chr.18: 5416253-5416253 on Build GRCh38
rs772432794 Chr.18: 5416258-5416258 on Build GRCh38
rs745342533 Chr.18: 5416129-5416129 on Build GRCh38
rs140780596 Chr.18: 5415992-5415992 on Build GRCh38
rs768620034 Chr.18: 5416225-5416225 on Build GRCh38
rs760271032 Chr.18: 5416360-5416360 on Build GRCh38
rs543040062 Chr.18: 5416422-5416422 on Build GRCh38
rs770492712 Chr.18: 5415995-5415995 on Build GRCh38
rs769729986 Chr.18: 5416132-5416132 on Build GRCh38
rs757425312 Chr.18: 5416390-5416390 on Build GRCh38
rs746659318 Chr.18: 5415975-5415975 on Build GRCh38
rs753008817 Chr.18: 5416287-5416287 on Build GRCh38
rs764484780 Chr.18: 5416355-5416355 on Build GRCh38
rs774497427 Chr.18: 5416022-5416022 on Build GRCh38
rs777230071 Chr.18: 5415970-5415970 on Build GRCh38
rs745397874 Chr.18: 5416218-5416218 on Build GRCh38
rs150101312 Chr.18: 5416003-5416003 on Build GRCh38
rs1043635866 Chr.18: 5416482-5416482 on Build GRCh38
rs147018763 Chr.18: 5416113-5416113 on Build GRCh38
rs80084120 Chr.18: 5416432-5416432 on Build GRCh38
rs762092790 Chr.18: 5416039-5416039 on Build GRCh38
rs764431403 Chr.18: 5416296-5416296 on Build GRCh38
rs762944082 Chr.18: 5416399-5416400 on Build GRCh38
rs138184132 Chr.18: 5416146-5416146 on Build GRCh38
rs200875404 Chr.18: 5416223-5416223 on Build GRCh38
rs754570387 Chr.18: 5416379-5416379 on Build GRCh38
rs1011280555 Chr.18: 5416478-5416478 on Build GRCh38
rs150384501 Chr.18: 5416300-5416300 on Build GRCh38
rs763448311 Chr.18: 5416343-5416343 on Build GRCh38
rs534850711 Chr.18: 5416175-5416175 on Build GRCh38
rs777044444 Chr.18: 5416139-5416139 on Build GRCh38
rs893129638 Chr.18: 5416462-5416462 on Build GRCh38
rs969174603 Chr.18: 5416217-5416217 on Build GRCh38
rs759982893 Chr.18: 5416282-5416282 on Build GRCh38
rs765570304 Chr.18: 5416200-5416200 on Build GRCh38
rs777731525 Chr.18: 5416384-5416384 on Build GRCh38
rs528974171 Chr.18: 5416475-5416475 on Build GRCh38
rs375555518 Chr.18: 5416174-5416174 on Build GRCh38
rs1051165485 Chr.18: 5416044-5416044 on Build GRCh38
rs374487187 Chr.18: 5416332-5416332 on Build GRCh38
rs781418280 Chr.18: 5416394-5416394 on Build GRCh38
rs746760968 Chr.18: 5416389-5416389 on Build GRCh38
rs753296952 Chr.18: 5416378-5416378 on Build GRCh38
rs756880758 Chr.18: 5416111-5416111 on Build GRCh38
rs755355008 Chr.18: 5416088-5416088 on Build GRCh38
rs780756917 Chr.18: 5416210-5416210 on Build GRCh38
rs921456771 Chr.18: 5416358-5416358 on Build GRCh38
rs539490874 Chr.18: 5416201-5416201 on Build GRCh38
rs9966357 Chr.18: 5416222-5416222 on Build GRCh38
rs1053495390 Chr.18: 5416446-5416446 on Build GRCh38
rs749227489 Chr.18: 5416224-5416224 on Build GRCh38
rs548784436 Chr.18: 5415996-5415996 on Build GRCh38
rs778279169 Chr.18: 5416380-5416380 on Build GRCh38
rs771135161 Chr.18: 5416335-5416335 on Build GRCh38
rs191005310 Chr.18: 5416178-5416178 on Build GRCh38
rs754239715 Chr.18: 5416070-5416070 on Build GRCh38
rs1019070520 Chr.18: 5416177-5416177 on Build GRCh38
rs535925015 Chr.18: 5416299-5416299 on Build GRCh38
rs771758610 Chr.18: 5415961-5415961 on Build GRCh38
rs754368851 Chr.18: 5416190-5416190 on Build GRCh38
rs368126962 Chr.18: 5416010-5416010 on Build GRCh38
rs769058901 Chr.18: 5416386-5416395 on Build GRCh38
rs753344299 Chr.18: 5416318-5416318 on Build GRCh38
rs148074485 Chr.18: 5416290-5416290 on Build GRCh38
rs776949469 Chr.18: 5416386-5416389 on Build GRCh38
rs369506575 Chr.18: 5416340-5416340 on Build GRCh38
rs950152813 Chr.18: 5416021-5416021 on Build GRCh38
rs773624800 Chr.18: 5416263-5416263 on Build GRCh38
rs765736013 Chr.18: 5416285-5416285 on Build GRCh38
rs1054956720 Chr.18: 5416078-5416078 on Build GRCh38
rs779329304 Chr.18: 5416094-5416094 on Build GRCh38
rs199822438 Chr.18: 5416311-5416311 on Build GRCh38
rs8082898 Chr.18: 5416161-5416161 on Build GRCh38
rs369688255 Chr.18: 5416254-5416254 on Build GRCh38
rs750553653 Chr.18: 5416116-5416116 on Build GRCh38
rs773423711 Chr.18: 5416395-5416398 on Build GRCh38
rs529981219 Chr.18: 5416056-5416056 on Build GRCh38
rs777678156 Chr.18: 5416331-5416331 on Build GRCh38
rs770217031 Chr.18: 5416342-5416342 on Build GRCh38
rs750512041 Chr.18: 5416043-5416043 on Build GRCh38
rs564665638 Chr.18: 5416356-5416356 on Build GRCh38
rs746680748 Chr.18: 5416095-5416095 on Build GRCh38
rs145588627 Chr.18: 5416007-5416007 on Build GRCh38
rs749572023 Chr.18: 5415958-5415958 on Build GRCh38
rs899989694 Chr.18: 5416037-5416037 on Build GRCh38
rs772057472 Chr.18: 5416329-5416329 on Build GRCh38
rs983067488 Chr.18: 5415999-5415999 on Build GRCh38
rs775518887 Chr.18: 5416423-5416423 on Build GRCh38
rs763247088 Chr.18: 5416292-5416292 on Build GRCh38
rs776604249 Chr.18: 5416337-5416337 on Build GRCh38
rs779482228 Chr.18: 5416316-5416316 on Build GRCh38
rs766019164 Chr.18: 5416369-5416369 on Build GRCh38
rs766551459 Chr.18: 5416182-5416182 on Build GRCh38
rs768993946 Chr.18: 5416383-5416383 on Build GRCh38
rs765361578 Chr.18: 5416204-5416204 on Build GRCh38
rs746065810 Chr.18: 5416396-5416396 on Build GRCh38
rs868864994 Chr.18: 5416074-5416074 on Build GRCh38
rs755477005 Chr.18: 5416196-5416196 on Build GRCh38
rs375525801 Chr.18: 5416276-5416276 on Build GRCh38
rs141321874 Chr.18: 5416008-5416008 on Build GRCh38
rs768404272 Chr.18: 5416144-5416144 on Build GRCh38
rs776750033 Chr.18: 5416281-5416281 on Build GRCh38
rs765430265 Chr.18: 5416391-5416394 on Build GRCh38
rs372345879 Chr.18: 5416264-5416264 on Build GRCh38
rs578183073 Chr.18: 5416385-5416385 on Build GRCh38
rs762089110 Chr.18: 5416385-5416386 on Build GRCh38
rs775234996 Chr.18: 5416136-5416136 on Build GRCh38

View Full Product Details