Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31129533-31130092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC1
Entrez Gene ID
1823
Gene Name
desmocollin 1
Gene Aliases
CDHF1, DG2/DG3
UniGene
Hs.567260
Gene Chromosome Location
Chr.18: 31129236-31162856 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Gene Symbol
DSCAS
Entrez Gene ID
101927698
Gene Name
DSC1/DSC2 antisense RNA
Gene Aliases
-
UniGene
Hs.579538
Gene Chromosome Location
Chr.18: 31101588-31162789 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

SNP Detail

SNP ID Chromosome Location
rs1008636264 Chr.18: 31129922-31129922 on Build GRCh38
rs56082715 Chr.18: 31129730-31129730 on Build GRCh38
rs989068067 Chr.18: 31129979-31129979 on Build GRCh38
rs917322924 Chr.18: 31129719-31129719 on Build GRCh38
rs7241456 Chr.18: 31129780-31129780 on Build GRCh38
rs570280544 Chr.18: 31129754-31129754 on Build GRCh38
rs536515861 Chr.18: 31129569-31129569 on Build GRCh38
rs964629795 Chr.18: 31129646-31129646 on Build GRCh38
rs534412579 Chr.18: 31129946-31129946 on Build GRCh38
rs747572052 Chr.18: 31129798-31129798 on Build GRCh38
rs1021362457 Chr.18: 31129629-31129629 on Build GRCh38
rs8095354 Chr.18: 31129641-31129641 on Build GRCh38
rs971916343 Chr.18: 31130032-31130032 on Build GRCh38
rs943207231 Chr.18: 31129816-31129816 on Build GRCh38
rs925827016 Chr.18: 31129659-31129659 on Build GRCh38
rs573829157 Chr.18: 31129925-31129925 on Build GRCh38
rs764982246 Chr.18: 31129723-31129723 on Build GRCh38
rs2740995 Chr.18: 31129865-31129865 on Build GRCh38
rs76575208 Chr.18: 31129994-31129994 on Build GRCh38
rs554564577 Chr.18: 31129947-31129947 on Build GRCh38
rs1002382865 Chr.18: 31130052-31130052 on Build GRCh38
rs373168708 Chr.18: 31130011-31130011 on Build GRCh38
rs7240259 Chr.18: 31129596-31129596 on Build GRCh38
rs555506601 Chr.18: 31129872-31129872 on Build GRCh38
rs978628039 Chr.18: 31129653-31129653 on Build GRCh38
rs552957398 Chr.18: 31129956-31129956 on Build GRCh38
rs955916945 Chr.18: 31129974-31129974 on Build GRCh38
rs543824835 Chr.18: 31129588-31129588 on Build GRCh38
rs994748147 Chr.18: 31129844-31129844 on Build GRCh38
rs537603149 Chr.18: 31129840-31129840 on Build GRCh38
rs186368171 Chr.18: 31129660-31129660 on Build GRCh38
rs941633208 Chr.18: 31129741-31129741 on Build GRCh38
rs565920995 Chr.18: 31129702-31129702 on Build GRCh38
rs897326278 Chr.18: 31129580-31129580 on Build GRCh38
rs775229974 Chr.18: 31130036-31130036 on Build GRCh38
rs1044803310 Chr.18: 31129724-31129724 on Build GRCh38
rs897378403 Chr.18: 31129794-31129794 on Build GRCh38
rs923098873 Chr.18: 31129889-31129889 on Build GRCh38
rs746921509 Chr.18: 31129607-31129607 on Build GRCh38
rs988627052 Chr.18: 31129675-31129675 on Build GRCh38
rs7244822 Chr.18: 31130059-31130059 on Build GRCh38
rs181225870 Chr.18: 31129561-31129561 on Build GRCh38
rs950731610 Chr.18: 31129758-31129758 on Build GRCh38
rs577535646 Chr.18: 31129966-31129966 on Build GRCh38
rs1017709833 Chr.18: 31129933-31129933 on Build GRCh38
rs947045216 Chr.18: 31129985-31129985 on Build GRCh38
rs997198174 Chr.18: 31129559-31129559 on Build GRCh38
rs1009000069 Chr.18: 31129614-31129614 on Build GRCh38
rs993281866 Chr.18: 31129585-31129585 on Build GRCh38
rs890218236 Chr.18: 31129912-31129912 on Build GRCh38
rs985291313 Chr.18: 31129773-31129773 on Build GRCh38
rs558140678 Chr.18: 31129745-31129745 on Build GRCh38
rs900171902 Chr.18: 31129949-31129949 on Build GRCh38
rs761241979 Chr.18: 31129975-31129975 on Build GRCh38
rs1032896613 Chr.18: 31129963-31129963 on Build GRCh38

View Full Product Details