Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36138689-36139275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP2
Entrez Gene ID
55250
Gene Name
elongator acetyltransferase complex subunit 2
Gene Aliases
SHINC-2, STATIP1, StIP
UniGene
Hs.606040
Gene Chromosome Location
Chr.18: 36129874-36177931 on Build GRCh38
Transcripts
NM_001242875.2 NM_001242876.2 NM_001242877.2 NM_001242878.2 NM_001242879.2 NM_001324465.1 NM_001324466.1 NM_001324467.1 NM_001324468.1 NM_018255.3 NR_040110.2 NR_136897.1 NR_136898.1 NR_137173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388587 c.510G>A p.L170L Chr.18: 36138859-36138859 on Build GRCh38
6451436 c.522T>C p.D174D Chr.18: 36138871-36138871 on Build GRCh38
1284724 c.479G>A p.G160E Chr.18: 36138828-36138828 on Build GRCh38
226361 c.506T>A p.F169Y Chr.18: 36138855-36138855 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs941419755 Chr.18: 36138954-36138954 on Build GRCh38
rs111298859 Chr.18: 36139017-36139017 on Build GRCh38
rs966096948 Chr.18: 36139080-36139080 on Build GRCh38
rs192906352 Chr.18: 36138989-36138989 on Build GRCh38
rs903587663 Chr.18: 36138739-36138739 on Build GRCh38
rs757764496 Chr.18: 36138783-36138783 on Build GRCh38
rs112931732 Chr.18: 36138731-36138731 on Build GRCh38
rs765162733 Chr.18: 36138849-36138849 on Build GRCh38
rs774055025 Chr.18: 36138828-36138828 on Build GRCh38
rs762249501 Chr.18: 36138906-36138906 on Build GRCh38
rs779649567 Chr.18: 36138801-36138801 on Build GRCh38
rs755533687 Chr.18: 36138798-36138798 on Build GRCh38
rs932440794 Chr.18: 36139102-36139102 on Build GRCh38
rs746428166 Chr.18: 36138790-36138790 on Build GRCh38
rs370583526 Chr.18: 36138761-36138761 on Build GRCh38
rs779050987 Chr.18: 36138925-36138929 on Build GRCh38
rs185109263 Chr.18: 36139165-36139165 on Build GRCh38
rs375440731 Chr.18: 36138758-36138758 on Build GRCh38
rs554004099 Chr.18: 36139045-36139045 on Build GRCh38
rs951593568 Chr.18: 36139208-36139208 on Build GRCh38
rs1033585982 Chr.18: 36139249-36139249 on Build GRCh38
rs189035462 Chr.18: 36138928-36138928 on Build GRCh38
rs922528079 Chr.18: 36139083-36139083 on Build GRCh38
rs151288583 Chr.18: 36139137-36139137 on Build GRCh38
rs200362076 Chr.18: 36138893-36138893 on Build GRCh38
rs752822541 Chr.18: 36138760-36138760 on Build GRCh38
rs578259200 Chr.18: 36138880-36138880 on Build GRCh38
rs957682368 Chr.18: 36138983-36138983 on Build GRCh38
rs3737473 Chr.18: 36139081-36139081 on Build GRCh38
rs773170323 Chr.18: 36138752-36138752 on Build GRCh38
rs748831020 Chr.18: 36138815-36138815 on Build GRCh38
rs751961484 Chr.18: 36138778-36138778 on Build GRCh38
rs767435300 Chr.18: 36138744-36138744 on Build GRCh38
rs147765982 Chr.18: 36138825-36138825 on Build GRCh38
rs945323928 Chr.18: 36139103-36139103 on Build GRCh38
rs983050752 Chr.18: 36139219-36139219 on Build GRCh38
rs1785933 Chr.18: 36139228-36139228 on Build GRCh38
rs756652255 Chr.18: 36138916-36138916 on Build GRCh38
rs547853099 Chr.18: 36138969-36138969 on Build GRCh38
rs1037102593 Chr.18: 36138920-36138920 on Build GRCh38
rs113249789 Chr.18: 36138835-36138835 on Build GRCh38
rs140557040 Chr.18: 36139150-36139150 on Build GRCh38
rs1024508665 Chr.18: 36139196-36139196 on Build GRCh38
rs539643011 Chr.18: 36139131-36139131 on Build GRCh38
rs866711710 Chr.18: 36139214-36139214 on Build GRCh38
rs1043355621 Chr.18: 36139108-36139108 on Build GRCh38
rs368719635 Chr.18: 36138846-36138846 on Build GRCh38
rs146949006 Chr.18: 36139153-36139153 on Build GRCh38
rs755055967 Chr.18: 36138852-36138853 on Build GRCh38
rs921063338 Chr.18: 36138986-36138986 on Build GRCh38
rs1021950135 Chr.18: 36138736-36138736 on Build GRCh38
rs781757063 Chr.18: 36138787-36138787 on Build GRCh38
rs764308459 Chr.18: 36138777-36138777 on Build GRCh38
rs768018753 Chr.18: 36138909-36138909 on Build GRCh38
rs751871798 Chr.18: 36138904-36138904 on Build GRCh38
rs537978540 Chr.18: 36138947-36138947 on Build GRCh38
rs909909342 Chr.18: 36138948-36138948 on Build GRCh38
rs747904116 Chr.18: 36138838-36138838 on Build GRCh38
rs1028038572 Chr.18: 36138770-36138770 on Build GRCh38
rs909757163 Chr.18: 36139110-36139110 on Build GRCh38
rs892091662 Chr.18: 36138711-36138711 on Build GRCh38
rs560443930 Chr.18: 36139238-36139238 on Build GRCh38
rs546890930 Chr.18: 36138842-36138842 on Build GRCh38
rs757324542 Chr.18: 36138980-36138980 on Build GRCh38
rs188032810 Chr.18: 36139186-36139186 on Build GRCh38
rs773082376 Chr.18: 36138845-36138845 on Build GRCh38
rs763001592 Chr.18: 36138889-36138889 on Build GRCh38
rs903534946 Chr.18: 36139128-36139128 on Build GRCh38
rs372307158 Chr.18: 36138911-36138911 on Build GRCh38
rs533557866 Chr.18: 36139016-36139016 on Build GRCh38
rs888202379 Chr.18: 36138852-36138852 on Build GRCh38
rs962750913 Chr.18: 36139232-36139232 on Build GRCh38

View Full Product Details