Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36138112-36138711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP2
Entrez Gene ID
55250
Gene Name
elongator acetyltransferase complex subunit 2
Gene Aliases
SHINC-2, STATIP1, StIP
UniGene
Hs.606040
Gene Chromosome Location
Chr.18: 36129874-36177931 on Build GRCh38
Transcripts
NM_001242875.2 NM_001242876.2 NM_001242877.2 NM_001242878.2 NM_001242879.2 NM_001324465.1 NM_001324466.1 NM_001324467.1 NM_001324468.1 NM_018255.3 NR_040110.2 NR_136897.1 NR_136898.1 NR_137173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000537 c.382A>C p.T128P Chr.18: 36138363-36138363 on Build GRCh38
6653521 c.297A>G p.K99K Chr.18: 36138278-36138278 on Build GRCh38
6653519 c.430delA p.K145fs*73 Chr.18: 36138411-36138411 on Build GRCh38
6221154 c.353A>G p.Q118R Chr.18: 36138334-36138334 on Build GRCh38
4072042 c.438T>C p.G146G Chr.18: 36138419-36138419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563526283 Chr.18: 36138474-36138477 on Build GRCh38
rs370293395 Chr.18: 36138450-36138450 on Build GRCh38
rs189770368 Chr.18: 36138185-36138185 on Build GRCh38
rs766253618 Chr.18: 36138440-36138440 on Build GRCh38
rs761650921 Chr.18: 36138456-36138456 on Build GRCh38
rs377486543 Chr.18: 36138379-36138379 on Build GRCh38
rs961539707 Chr.18: 36138257-36138257 on Build GRCh38
rs1029720746 Chr.18: 36138403-36138403 on Build GRCh38
rs917223925 Chr.18: 36138568-36138568 on Build GRCh38
rs760991896 Chr.18: 36138385-36138385 on Build GRCh38
rs769471732 Chr.18: 36138422-36138422 on Build GRCh38
rs554587235 Chr.18: 36138299-36138299 on Build GRCh38
rs144238695 Chr.18: 36138417-36138417 on Build GRCh38
rs948803626 Chr.18: 36138294-36138294 on Build GRCh38
rs1009238234 Chr.18: 36138420-36138420 on Build GRCh38
rs982821828 Chr.18: 36138599-36138599 on Build GRCh38
rs937641885 Chr.18: 36138181-36138181 on Build GRCh38
rs762865739 Chr.18: 36138231-36138231 on Build GRCh38
rs529659464 Chr.18: 36138270-36138271 on Build GRCh38
rs765057683 Chr.18: 36138220-36138220 on Build GRCh38
rs183528073 Chr.18: 36138208-36138208 on Build GRCh38
rs74956437 Chr.18: 36138238-36138238 on Build GRCh38
rs1024420045 Chr.18: 36138576-36138576 on Build GRCh38
rs75833918 Chr.18: 36138228-36138228 on Build GRCh38
rs909963607 Chr.18: 36138615-36138615 on Build GRCh38
rs776285370 Chr.18: 36138319-36138319 on Build GRCh38
rs753990082 Chr.18: 36138286-36138286 on Build GRCh38
rs769557991 Chr.18: 36138341-36138341 on Build GRCh38
rs761640327 Chr.18: 36138221-36138221 on Build GRCh38
rs1039925091 Chr.18: 36138658-36138658 on Build GRCh38
rs562795859 Chr.18: 36138484-36138484 on Build GRCh38
rs760642020 Chr.18: 36138354-36138354 on Build GRCh38
rs900923236 Chr.18: 36138161-36138161 on Build GRCh38
rs762626883 Chr.18: 36138354-36138354 on Build GRCh38
rs752450625 Chr.18: 36138230-36138230 on Build GRCh38
rs757852630 Chr.18: 36138400-36138400 on Build GRCh38
rs766384110 Chr.18: 36138364-36138364 on Build GRCh38
rs34817357 Chr.18: 36138238-36138238 on Build GRCh38
rs748742569 Chr.18: 36138151-36138151 on Build GRCh38
rs758640091 Chr.18: 36138386-36138386 on Build GRCh38
rs888244494 Chr.18: 36138293-36138293 on Build GRCh38
rs970223484 Chr.18: 36138595-36138595 on Build GRCh38
rs770524620 Chr.18: 36138317-36138317 on Build GRCh38
rs929967082 Chr.18: 36138569-36138569 on Build GRCh38
rs941485062 Chr.18: 36138625-36138625 on Build GRCh38
rs748995997 Chr.18: 36138441-36138441 on Build GRCh38
rs753011010 Chr.18: 36138372-36138372 on Build GRCh38
rs774147661 Chr.18: 36138220-36138221 on Build GRCh38
rs755203115 Chr.18: 36138289-36138289 on Build GRCh38
rs751558018 Chr.18: 36138245-36138245 on Build GRCh38
rs768299769 Chr.18: 36138444-36138444 on Build GRCh38
rs202140481 Chr.18: 36138231-36138232 on Build GRCh38
rs397734289 Chr.18: 36138239-36138240 on Build GRCh38
rs879606331 Chr.18: 36138520-36138520 on Build GRCh38
rs538079080 Chr.18: 36138316-36138316 on Build GRCh38
rs374188452 Chr.18: 36138371-36138371 on Build GRCh38
rs1016279941 Chr.18: 36138255-36138255 on Build GRCh38
rs1049531887 Chr.18: 36138497-36138497 on Build GRCh38
rs781779741 Chr.18: 36138411-36138411 on Build GRCh38
rs373862565 Chr.18: 36138253-36138253 on Build GRCh38
rs74715150 Chr.18: 36138238-36138239 on Build GRCh38
rs1785934 Chr.18: 36138363-36138363 on Build GRCh38
rs927512937 Chr.18: 36138494-36138494 on Build GRCh38
rs868320523 Chr.18: 36138499-36138499 on Build GRCh38
rs925936844 Chr.18: 36138310-36138310 on Build GRCh38
rs759209061 Chr.18: 36138320-36138320 on Build GRCh38
rs1014547941 Chr.18: 36138536-36138536 on Build GRCh38
rs1047188949 Chr.18: 36138271-36138271 on Build GRCh38
rs9950833 Chr.18: 36138142-36138142 on Build GRCh38
rs142403520 Chr.18: 36138503-36138503 on Build GRCh38
rs369683728 Chr.18: 36138309-36138309 on Build GRCh38
rs766715470 Chr.18: 36138210-36138210 on Build GRCh38
rs759195187 Chr.18: 36138513-36138513 on Build GRCh38
rs562147638 Chr.18: 36138671-36138671 on Build GRCh38
rs753898891 Chr.18: 36138368-36138368 on Build GRCh38
rs751407249 Chr.18: 36138357-36138357 on Build GRCh38
rs370651899 Chr.18: 36138398-36138398 on Build GRCh38
rs775282268 Chr.18: 36138346-36138346 on Build GRCh38
rs888090940 Chr.18: 36138500-36138500 on Build GRCh38
rs541850490 Chr.18: 36138552-36138552 on Build GRCh38
rs1015814500 Chr.18: 36138504-36138504 on Build GRCh38
rs774985828 Chr.18: 36138431-36138431 on Build GRCh38
rs377022607 Chr.18: 36138272-36138272 on Build GRCh38
rs537436755 Chr.18: 36138298-36138298 on Build GRCh38
rs558363966 Chr.18: 36138356-36138356 on Build GRCh38
rs200469142 Chr.18: 36138399-36138399 on Build GRCh38
rs956387664 Chr.18: 36138412-36138412 on Build GRCh38
rs879529052 Chr.18: 36138415-36138415 on Build GRCh38
rs767125890 Chr.18: 36138667-36138667 on Build GRCh38

View Full Product Details