Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12730868-12731464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf43
Entrez Gene ID
79002
Gene Name
chromosome 19 open reading frame 43
Gene Aliases
fSAP18
UniGene
Hs.515155
Gene Chromosome Location
Chr.19: 12730640-12734715 on Build GRCh38
Transcripts
NM_024038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6840854 c.514C>G p.R172G Chr.19: 12730978-12730978 on Build GRCh38
5149947 c.381C>T p.L127L Chr.19: 12731386-12731386 on Build GRCh38
1724732 c.389_390insG p.I131fs*11 Chr.19: 12731377-12731378 on Build GRCh38
7075998 c.413C>T p.T138M Chr.19: 12731354-12731354 on Build GRCh38
266123 c.362G>A p.G121D Chr.19: 12731405-12731405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374134076 Chr.19: 12731096-12731096 on Build GRCh38
rs372009797 Chr.19: 12731427-12731427 on Build GRCh38
rs762029407 Chr.19: 12731120-12731120 on Build GRCh38
rs11542951 Chr.19: 12731370-12731370 on Build GRCh38
rs755724441 Chr.19: 12731092-12731092 on Build GRCh38
rs765303818 Chr.19: 12731032-12731032 on Build GRCh38
rs867932101 Chr.19: 12730951-12730951 on Build GRCh38
rs542916907 Chr.19: 12731229-12731229 on Build GRCh38
rs778385132 Chr.19: 12731411-12731411 on Build GRCh38
rs773189461 Chr.19: 12731296-12731296 on Build GRCh38
rs758810543 Chr.19: 12730924-12730925 on Build GRCh38
rs764415193 Chr.19: 12731005-12731005 on Build GRCh38
rs961125706 Chr.19: 12731163-12731163 on Build GRCh38
rs544721720 Chr.19: 12731303-12731303 on Build GRCh38
rs754112487 Chr.19: 12731015-12731015 on Build GRCh38
rs769071271 Chr.19: 12731107-12731107 on Build GRCh38
rs757959761 Chr.19: 12731214-12731214 on Build GRCh38
rs116718949 Chr.19: 12731048-12731048 on Build GRCh38
rs775736306 Chr.19: 12730996-12730996 on Build GRCh38
rs551567529 Chr.19: 12731353-12731353 on Build GRCh38
rs754868268 Chr.19: 12731085-12731085 on Build GRCh38
rs765442897 Chr.19: 12731295-12731295 on Build GRCh38
rs367862325 Chr.19: 12730936-12730936 on Build GRCh38
rs147059853 Chr.19: 12730959-12730959 on Build GRCh38
rs903927818 Chr.19: 12731436-12731436 on Build GRCh38
rs891660843 Chr.19: 12731389-12731389 on Build GRCh38
rs750388405 Chr.19: 12730949-12730949 on Build GRCh38
rs758284079 Chr.19: 12731045-12731045 on Build GRCh38
rs769087859 Chr.19: 12731423-12731423 on Build GRCh38
rs762012753 Chr.19: 12731019-12731019 on Build GRCh38
rs747584062 Chr.19: 12731412-12731412 on Build GRCh38
rs79547465 Chr.19: 12731257-12731257 on Build GRCh38
rs767013231 Chr.19: 12731001-12731001 on Build GRCh38
rs371093491 Chr.19: 12730919-12730919 on Build GRCh38
rs777158461 Chr.19: 12731094-12731094 on Build GRCh38
rs752403130 Chr.19: 12731070-12731070 on Build GRCh38
rs866319514 Chr.19: 12730897-12730897 on Build GRCh38
rs139906319 Chr.19: 12730960-12730960 on Build GRCh38
rs758354275 Chr.19: 12730952-12730952 on Build GRCh38
rs1017924636 Chr.19: 12730945-12730945 on Build GRCh38
rs201118967 Chr.19: 12731321-12731321 on Build GRCh38
rs753626060 Chr.19: 12730935-12730935 on Build GRCh38
rs768363859 Chr.19: 12731326-12731326 on Build GRCh38
rs769485637 Chr.19: 12730973-12730973 on Build GRCh38
rs540128547 Chr.19: 12731238-12731239 on Build GRCh38
rs1000879915 Chr.19: 12731445-12731445 on Build GRCh38
rs753946306 Chr.19: 12730924-12730924 on Build GRCh38
rs990803068 Chr.19: 12731114-12731114 on Build GRCh38
rs370164525 Chr.19: 12730991-12730991 on Build GRCh38
rs935475457 Chr.19: 12731354-12731354 on Build GRCh38
rs748787688 Chr.19: 12731072-12731073 on Build GRCh38
rs567121997 Chr.19: 12730896-12730896 on Build GRCh38
rs760982984 Chr.19: 12730997-12730997 on Build GRCh38
rs778148647 Chr.19: 12731070-12731071 on Build GRCh38
rs999708749 Chr.19: 12730891-12730891 on Build GRCh38
rs370637749 Chr.19: 12731287-12731287 on Build GRCh38
rs747874561 Chr.19: 12731088-12731088 on Build GRCh38
rs368117881 Chr.19: 12731319-12731319 on Build GRCh38
rs760073690 Chr.19: 12730915-12730915 on Build GRCh38
rs764247739 Chr.19: 12730922-12730922 on Build GRCh38
rs752478963 Chr.19: 12731338-12731338 on Build GRCh38
rs553970936 Chr.19: 12731095-12731095 on Build GRCh38
rs746744760 Chr.19: 12730957-12730957 on Build GRCh38
rs767835599 Chr.19: 12730918-12730918 on Build GRCh38
rs938555472 Chr.19: 12731333-12731333 on Build GRCh38
rs1042515072 Chr.19: 12731425-12731425 on Build GRCh38
rs774769035 Chr.19: 12730910-12730910 on Build GRCh38
rs750451295 Chr.19: 12731039-12731039 on Build GRCh38
rs939427879 Chr.19: 12731065-12731065 on Build GRCh38
rs754959353 Chr.19: 12731320-12731320 on Build GRCh38
rs75619212 Chr.19: 12731180-12731180 on Build GRCh38
rs555967740 Chr.19: 12730927-12730927 on Build GRCh38
rs777438434 Chr.19: 12731381-12731381 on Build GRCh38
rs776978076 Chr.19: 12731117-12731117 on Build GRCh38
rs748498938 Chr.19: 12731433-12731433 on Build GRCh38
rs141893958 Chr.19: 12730976-12730976 on Build GRCh38
rs186539758 Chr.19: 12731403-12731403 on Build GRCh38
rs766339455 Chr.19: 12731316-12731316 on Build GRCh38
rs756844642 Chr.19: 12731406-12731406 on Build GRCh38
rs751432254 Chr.19: 12731073-12731073 on Build GRCh38
rs759962400 Chr.19: 12730977-12730977 on Build GRCh38
rs779719752 Chr.19: 12730953-12730953 on Build GRCh38
rs781121211 Chr.19: 12731086-12731086 on Build GRCh38
rs374162606 Chr.19: 12731105-12731105 on Build GRCh38
rs930960720 Chr.19: 12730911-12730911 on Build GRCh38
rs374497183 Chr.19: 12730921-12730921 on Build GRCh38
rs945841727 Chr.19: 12731407-12731407 on Build GRCh38

View Full Product Details