Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35332599-35333076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD22
Entrez Gene ID
933
Gene Name
CD22 molecule
Gene Aliases
SIGLEC-2, SIGLEC2
UniGene
Hs.579691
Gene Chromosome Location
Chr.19: 35329166-35347361 on Build GRCh38
Transcripts
NM_001185099.1 NM_001185100.1 NM_001185101.1 NM_001278417.1 NM_001771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131663 c.198G>A p.K66K Chr.19: 35332710-35332710 on Build GRCh38
368697 c.154G>C p.D52H Chr.19: 35332666-35332666 on Build GRCh38
6211900 c.226_227delCT p.Y77fs*1 Chr.19: 35332738-35332739 on Build GRCh38
3388875 c.117C>T p.C39C Chr.19: 35332629-35332629 on Build GRCh38
6150513 c.168C>T p.F56F Chr.19: 35332680-35332680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950776362 Chr.19: 35333013-35333013 on Build GRCh38
rs200519612 Chr.19: 35332822-35332822 on Build GRCh38
rs767934053 Chr.19: 35332717-35332717 on Build GRCh38
rs774846059 Chr.19: 35332766-35332766 on Build GRCh38
rs747868335 Chr.19: 35332962-35332962 on Build GRCh38
rs146453327 Chr.19: 35332845-35332845 on Build GRCh38
rs139372163 Chr.19: 35332740-35332740 on Build GRCh38
rs756553134 Chr.19: 35332953-35332954 on Build GRCh38
rs61741050 Chr.19: 35332701-35332701 on Build GRCh38
rs756111081 Chr.19: 35332630-35332630 on Build GRCh38
rs766345228 Chr.19: 35332707-35332707 on Build GRCh38
rs758941910 Chr.19: 35332668-35332668 on Build GRCh38
rs750364404 Chr.19: 35332629-35332629 on Build GRCh38
rs1057268405 Chr.19: 35332937-35332937 on Build GRCh38
rs573917789 Chr.19: 35332659-35332659 on Build GRCh38
rs747204167 Chr.19: 35332719-35332719 on Build GRCh38
rs113867292 Chr.19: 35332714-35332714 on Build GRCh38
rs765085221 Chr.19: 35332811-35332811 on Build GRCh38
rs1053162142 Chr.19: 35332794-35332794 on Build GRCh38
rs748552865 Chr.19: 35332912-35332912 on Build GRCh38
rs778714983 Chr.19: 35332718-35332718 on Build GRCh38
rs750745733 Chr.19: 35332939-35332940 on Build GRCh38
rs780112762 Chr.19: 35332881-35332881 on Build GRCh38
rs145824675 Chr.19: 35332836-35332836 on Build GRCh38
rs772876606 Chr.19: 35332923-35332923 on Build GRCh38
rs763921154 Chr.19: 35332962-35332962 on Build GRCh38
rs979872474 Chr.19: 35333051-35333051 on Build GRCh38
rs772387010 Chr.19: 35332922-35332922 on Build GRCh38
rs770355855 Chr.19: 35332939-35332939 on Build GRCh38
rs750677274 Chr.19: 35332970-35332970 on Build GRCh38
rs779955039 Chr.19: 35332636-35332636 on Build GRCh38
rs879748064 Chr.19: 35332961-35332961 on Build GRCh38
rs768906675 Chr.19: 35332764-35332764 on Build GRCh38
rs111284429 Chr.19: 35332643-35332643 on Build GRCh38
rs565080840 Chr.19: 35332882-35332882 on Build GRCh38
rs769951377 Chr.19: 35332749-35332749 on Build GRCh38
rs145580205 Chr.19: 35332796-35332796 on Build GRCh38
rs1029119476 Chr.19: 35333034-35333034 on Build GRCh38
rs373299889 Chr.19: 35332681-35332681 on Build GRCh38
rs201695548 Chr.19: 35332945-35332945 on Build GRCh38
rs948821707 Chr.19: 35333044-35333044 on Build GRCh38
rs928475747 Chr.19: 35333052-35333052 on Build GRCh38
rs762071219 Chr.19: 35332768-35332768 on Build GRCh38
rs1017799238 Chr.19: 35332936-35332936 on Build GRCh38
rs771152146 Chr.19: 35332656-35332656 on Build GRCh38
rs764854283 Chr.19: 35332748-35332748 on Build GRCh38
rs778705464 Chr.19: 35332955-35332956 on Build GRCh38
rs147711132 Chr.19: 35332801-35332801 on Build GRCh38
rs754753405 Chr.19: 35332710-35332710 on Build GRCh38
rs763054158 Chr.19: 35332904-35332904 on Build GRCh38
rs747208002 Chr.19: 35332654-35332654 on Build GRCh38
rs752581963 Chr.19: 35332827-35332827 on Build GRCh38
rs776788774 Chr.19: 35332657-35332657 on Build GRCh38
rs757623667 Chr.19: 35332738-35332738 on Build GRCh38
rs559954767 Chr.19: 35333043-35333043 on Build GRCh38
rs754881065 Chr.19: 35332805-35332805 on Build GRCh38
rs756274692 Chr.19: 35332862-35332862 on Build GRCh38
rs749087732 Chr.19: 35332638-35332638 on Build GRCh38
rs536032771 Chr.19: 35332660-35332660 on Build GRCh38
rs757506378 Chr.19: 35333015-35333015 on Build GRCh38
rs760212048 Chr.19: 35332788-35332788 on Build GRCh38
rs1026727042 Chr.19: 35332995-35332995 on Build GRCh38
rs746036345 Chr.19: 35332847-35332847 on Build GRCh38
rs565011703 Chr.19: 35332708-35332708 on Build GRCh38
rs370423783 Chr.19: 35332649-35332649 on Build GRCh38
rs763916938 Chr.19: 35332943-35332943 on Build GRCh38
rs762860609 Chr.19: 35332959-35332959 on Build GRCh38
rs376715267 Chr.19: 35332808-35332808 on Build GRCh38
rs772821688 Chr.19: 35332785-35332785 on Build GRCh38
rs144011147 Chr.19: 35332625-35332625 on Build GRCh38
rs199814640 Chr.19: 35332942-35332942 on Build GRCh38
rs749993637 Chr.19: 35332696-35332696 on Build GRCh38
rs552700291 Chr.19: 35332662-35332662 on Build GRCh38
rs1008727499 Chr.19: 35333050-35333050 on Build GRCh38
rs773887093 Chr.19: 35332677-35332677 on Build GRCh38
rs777890369 Chr.19: 35332650-35332650 on Build GRCh38
rs749744212 Chr.19: 35332757-35332757 on Build GRCh38
rs142079105 Chr.19: 35332826-35332826 on Build GRCh38
rs767151311 Chr.19: 35332689-35332689 on Build GRCh38
rs377348712 Chr.19: 35332935-35332935 on Build GRCh38
rs767208056 Chr.19: 35332775-35332775 on Build GRCh38
rs749323704 Chr.19: 35332900-35332900 on Build GRCh38
rs572709470 Chr.19: 35332667-35332667 on Build GRCh38
rs1044841386 Chr.19: 35332875-35332875 on Build GRCh38

View Full Product Details