Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129834554-129835148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf78
Entrez Gene ID
51759
Gene Name
chromosome 9 open reading frame 78
Gene Aliases
HCA59, HSPC220, bA409K20.3
UniGene
Hs.278429
Gene Chromosome Location
Chr.9: 129827285-129835293 on Build GRCh38
Transcripts
NM_016520.2 XM_011518780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346097 c.78G>T p.E26D Chr.9: 129835144-129835144 on Build GRCh38
4856926 c.97G>A p.E33K Chr.9: 129834753-129834753 on Build GRCh38
752567 c.123G>T p.R41S Chr.9: 129834727-129834727 on Build GRCh38
5977105 c.123G>A p.R41R Chr.9: 129834727-129834727 on Build GRCh38
1105884 c.82C>A p.R28R Chr.9: 129835140-129835140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773835491 Chr.9: 129834792-129834792 on Build GRCh38
rs1051122480 Chr.9: 129835085-129835085 on Build GRCh38
rs149710224 Chr.9: 129834949-129834949 on Build GRCh38
rs1048217525 Chr.9: 129835122-129835122 on Build GRCh38
rs979228693 Chr.9: 129834885-129834885 on Build GRCh38
rs762351981 Chr.9: 129834698-129834698 on Build GRCh38
rs762962586 Chr.9: 129834694-129834694 on Build GRCh38
rs746878845 Chr.9: 129834672-129834672 on Build GRCh38
rs971467806 Chr.9: 129834890-129834890 on Build GRCh38
rs560383269 Chr.9: 129835055-129835055 on Build GRCh38
rs370957981 Chr.9: 129834727-129834727 on Build GRCh38
rs1020427573 Chr.9: 129834875-129834875 on Build GRCh38
rs755867968 Chr.9: 129835093-129835093 on Build GRCh38
rs775004755 Chr.9: 129834697-129834697 on Build GRCh38
rs993005741 Chr.9: 129834847-129834847 on Build GRCh38
rs375570750 Chr.9: 129834673-129834673 on Build GRCh38
rs544012004 Chr.9: 129834910-129834910 on Build GRCh38
rs926039271 Chr.9: 129834897-129834897 on Build GRCh38
rs1017187361 Chr.9: 129834651-129834651 on Build GRCh38
rs78869308 Chr.9: 129834831-129834831 on Build GRCh38
rs923249491 Chr.9: 129835015-129835015 on Build GRCh38
rs750888060 Chr.9: 129834713-129834713 on Build GRCh38
rs888139402 Chr.9: 129834789-129834789 on Build GRCh38
rs913355062 Chr.9: 129834608-129834608 on Build GRCh38
rs763831212 Chr.9: 129834672-129834674 on Build GRCh38
rs751107953 Chr.9: 129834799-129834799 on Build GRCh38
rs918391001 Chr.9: 129834920-129834920 on Build GRCh38
rs1038566913 Chr.9: 129834844-129834844 on Build GRCh38
rs986096175 Chr.9: 129835045-129835045 on Build GRCh38
rs965278047 Chr.9: 129835065-129835065 on Build GRCh38
rs1048642486 Chr.9: 129834928-129834928 on Build GRCh38
rs960219570 Chr.9: 129834591-129834591 on Build GRCh38
rs745657015 Chr.9: 129834680-129834680 on Build GRCh38
rs778151032 Chr.9: 129834983-129834983 on Build GRCh38
rs760973134 Chr.9: 129834793-129834793 on Build GRCh38
rs888201516 Chr.9: 129834939-129834939 on Build GRCh38
rs761432344 Chr.9: 129835047-129835048 on Build GRCh38
rs571223625 Chr.9: 129835060-129835060 on Build GRCh38
rs751150302 Chr.9: 129834678-129834680 on Build GRCh38
rs566348414 Chr.9: 129834767-129834767 on Build GRCh38
rs368896017 Chr.9: 129834640-129834640 on Build GRCh38
rs766696278 Chr.9: 129834794-129834794 on Build GRCh38
rs762624319 Chr.9: 129834741-129834741 on Build GRCh38
rs375148916 Chr.9: 129834816-129834816 on Build GRCh38
rs953453087 Chr.9: 129835022-129835022 on Build GRCh38
rs3814547 Chr.9: 129834742-129834742 on Build GRCh38
rs777202959 Chr.9: 129834747-129834747 on Build GRCh38
rs768125195 Chr.9: 129834708-129834708 on Build GRCh38
rs183155719 Chr.9: 129834817-129834817 on Build GRCh38
rs761390965 Chr.9: 129835043-129835044 on Build GRCh38
rs780170176 Chr.9: 129834779-129834779 on Build GRCh38
rs749025411 Chr.9: 129835112-129835112 on Build GRCh38
rs769582785 Chr.9: 129834776-129834776 on Build GRCh38
rs937039318 Chr.9: 129835126-129835126 on Build GRCh38
rs772749042 Chr.9: 129834787-129834787 on Build GRCh38
rs3814548 Chr.9: 129834620-129834620 on Build GRCh38
rs911868183 Chr.9: 129835064-129835064 on Build GRCh38
rs41278736 Chr.9: 129834783-129834783 on Build GRCh38
rs575301088 Chr.9: 129834907-129834907 on Build GRCh38
rs929812991 Chr.9: 129834922-129834922 on Build GRCh38
rs896074872 Chr.9: 129834952-129834952 on Build GRCh38
rs779249106 Chr.9: 129835121-129835121 on Build GRCh38
rs953541640 Chr.9: 129834900-129834900 on Build GRCh38
rs769068705 Chr.9: 129835115-129835115 on Build GRCh38
rs529481261 Chr.9: 129834629-129834629 on Build GRCh38
rs1038514233 Chr.9: 129835098-129835098 on Build GRCh38
rs762722188 Chr.9: 129834798-129834798 on Build GRCh38
rs79372164 Chr.9: 129834911-129834911 on Build GRCh38
rs751052761 Chr.9: 129834751-129834751 on Build GRCh38
rs976666926 Chr.9: 129835087-129835087 on Build GRCh38
rs931986198 Chr.9: 129834699-129834699 on Build GRCh38
rs560966247 Chr.9: 129834774-129834776 on Build GRCh38
rs1040874120 Chr.9: 129834918-129834918 on Build GRCh38
rs200525467 Chr.9: 129834682-129834682 on Build GRCh38
rs761073432 Chr.9: 129834716-129834716 on Build GRCh38
rs757906505 Chr.9: 129834668-129834668 on Build GRCh38
rs373823052 Chr.9: 129834769-129834769 on Build GRCh38
rs755939769 Chr.9: 129835089-129835090 on Build GRCh38
rs752301503 Chr.9: 129834796-129834796 on Build GRCh38
rs371644803 Chr.9: 129834802-129834802 on Build GRCh38
rs967508908 Chr.9: 129834880-129834880 on Build GRCh38
rs764849524 Chr.9: 129834722-129834722 on Build GRCh38
rs761981667 Chr.9: 129834786-129834786 on Build GRCh38
rs994835801 Chr.9: 129834862-129834862 on Build GRCh38
rs756736019 Chr.9: 129834815-129834815 on Build GRCh38
rs540092085 Chr.9: 129834780-129834780 on Build GRCh38
rs775089842 Chr.9: 129834674-129834674 on Build GRCh38
rs960271592 Chr.9: 129834839-129834839 on Build GRCh38
rs943917613 Chr.9: 129834917-129834917 on Build GRCh38
rs774632595 Chr.9: 129834785-129834785 on Build GRCh38
rs779841642 Chr.9: 129835096-129835096 on Build GRCh38
rs1000364254 Chr.9: 129835001-129835001 on Build GRCh38
rs77295310 Chr.9: 129834588-129834588 on Build GRCh38
rs1051174958 Chr.9: 129834825-129834825 on Build GRCh38
rs775756516 Chr.9: 129834684-129834684 on Build GRCh38
rs745696468 Chr.9: 129834772-129834772 on Build GRCh38
rs187882057 Chr.9: 129834819-129834819 on Build GRCh38
rs750287903 Chr.9: 129834809-129834810 on Build GRCh38
rs767584648 Chr.9: 129834773-129834774 on Build GRCh38
rs918492887 Chr.9: 129835116-129835116 on Build GRCh38
rs757248973 Chr.9: 129834752-129834752 on Build GRCh38
rs1007047579 Chr.9: 129834833-129834833 on Build GRCh38
rs147629327 Chr.9: 129834656-129834656 on Build GRCh38
rs112144992 Chr.9: 129834743-129834743 on Build GRCh38
rs112226116 Chr.9: 129834776-129834778 on Build GRCh38
rs992994477 Chr.9: 129834594-129834594 on Build GRCh38
rs777715634 Chr.9: 129834670-129834670 on Build GRCh38
rs1014493092 Chr.9: 129834981-129834981 on Build GRCh38
rs764195364 Chr.9: 129834696-129834696 on Build GRCh38
rs199532327 Chr.9: 129835125-129835125 on Build GRCh38
rs552763768 Chr.9: 129835003-129835003 on Build GRCh38
rs1001643209 Chr.9: 129834941-129834941 on Build GRCh38
rs779654293 Chr.9: 129835102-129835102 on Build GRCh38
rs967285701 Chr.9: 129834977-129834977 on Build GRCh38
rs933968446 Chr.9: 129835025-129835025 on Build GRCh38
rs967971722 Chr.9: 129834994-129834994 on Build GRCh38
rs368566789 Chr.9: 129834690-129834690 on Build GRCh38

View Full Product Details