Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127087374-127087973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL2
Entrez Gene ID
23452
Gene Name
angiopoietin like 2
Gene Aliases
ARP2, HARP
UniGene
Hs.653262
Gene Chromosome Location
Chr.9: 127087349-127122803 on Build GRCh38
Transcripts
NM_012098.2 NM_001190728.1 NM_001190729.1 NM_001190730.1 NM_001322320.1 NM_001322321.1 NM_001322322.1 NM_001322323.1 NM_001322324.1 NM_001322325.1 NM_014636.2 XM_006717328.3 XM_011519225.1 XM_011519226.1 XM_011519227.1 XM_011519228.1 XM_011519229.1 XM_011519230.2 XM_011519231.1 XM_011519232.1 XM_011519233.1 XM_011519234.2 XM_011519235.2 XM_011519236.1 XM_011519238.2 XM_017015342.1 XM_017015343.1 XM_017015344.1 XM_017015345.1 XM_017015346.1 XM_017015347.1 XM_017015348.1 XM_017015349.1 XM_017015350.1

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
UniGene
Hs.432842
Gene Chromosome Location
Chr.9: 126914774-127223166 on Build GRCh38
Transcripts
NM_012098.2 NM_001190728.1 NM_001190729.1 NM_001190730.1 NM_001322320.1 NM_001322321.1 NM_001322322.1 NM_001322323.1 NM_001322324.1 NM_001322325.1 NM_014636.2 XM_006717328.3 XM_011519225.1 XM_011519226.1 XM_011519227.1 XM_011519228.1 XM_011519229.1 XM_011519230.2 XM_011519231.1 XM_011519232.1 XM_011519233.1 XM_011519234.2 XM_011519235.2 XM_011519236.1 XM_011519238.2 XM_017015342.1 XM_017015343.1 XM_017015344.1 XM_017015345.1 XM_017015346.1 XM_017015347.1 XM_017015348.1 XM_017015349.1 XM_017015350.1

SNP Detail

SNP ID Chromosome Location
rs559234043 Chr.9: 127087701-127087701 on Build GRCh38
rs999671163 Chr.9: 127087557-127087557 on Build GRCh38
rs900953118 Chr.9: 127087654-127087654 on Build GRCh38
rs927422304 Chr.9: 127087472-127087472 on Build GRCh38
rs1000105970 Chr.9: 127087935-127087935 on Build GRCh38
rs1047434141 Chr.9: 127087441-127087441 on Build GRCh38
rs575684341 Chr.9: 127087556-127087556 on Build GRCh38
rs567058156 Chr.9: 127087519-127087519 on Build GRCh38
rs968702998 Chr.9: 127087437-127087437 on Build GRCh38
rs145539359 Chr.9: 127087577-127087577 on Build GRCh38
rs969237487 Chr.9: 127087866-127087866 on Build GRCh38
rs938758805 Chr.9: 127087479-127087479 on Build GRCh38
rs376537455 Chr.9: 127087714-127087714 on Build GRCh38
rs769789517 Chr.9: 127087759-127087759 on Build GRCh38
rs535550381 Chr.9: 127087540-127087540 on Build GRCh38
rs8838 Chr.9: 127087601-127087601 on Build GRCh38
rs185044047 Chr.9: 127087774-127087774 on Build GRCh38
rs530540820 Chr.9: 127087410-127087410 on Build GRCh38
rs1017881853 Chr.9: 127087603-127087603 on Build GRCh38
rs1046931311 Chr.9: 127087825-127087825 on Build GRCh38
rs909831566 Chr.9: 127087766-127087766 on Build GRCh38
rs11556883 Chr.9: 127087440-127087440 on Build GRCh38
rs188765247 Chr.9: 127087457-127087457 on Build GRCh38
rs749130592 Chr.9: 127087861-127087861 on Build GRCh38
rs7019920 Chr.9: 127087762-127087762 on Build GRCh38
rs906584341 Chr.9: 127087864-127087864 on Build GRCh38
rs545698957 Chr.9: 127087682-127087682 on Build GRCh38
rs748215299 Chr.9: 127087749-127087749 on Build GRCh38
rs980107819 Chr.9: 127087869-127087869 on Build GRCh38
rs374264086 Chr.9: 127087775-127087775 on Build GRCh38
rs565994591 Chr.9: 127087681-127087681 on Build GRCh38
rs866628718 Chr.9: 127087538-127087538 on Build GRCh38
rs1006566498 Chr.9: 127087585-127087585 on Build GRCh38
rs540182631 Chr.9: 127087480-127087480 on Build GRCh38
rs529121958 Chr.9: 127087516-127087516 on Build GRCh38
rs927273915 Chr.9: 127087942-127087942 on Build GRCh38
rs7019153 Chr.9: 127087411-127087411 on Build GRCh38
rs1046584068 Chr.9: 127087541-127087541 on Build GRCh38
rs1012326812 Chr.9: 127087537-127087537 on Build GRCh38
rs989784680 Chr.9: 127087410-127087411 on Build GRCh38
rs549239847 Chr.9: 127087550-127087550 on Build GRCh38
rs951295264 Chr.9: 127087707-127087707 on Build GRCh38
rs10339 Chr.9: 127087451-127087451 on Build GRCh38
rs560959886 Chr.9: 127087455-127087455 on Build GRCh38
rs1031678947 Chr.9: 127087564-127087564 on Build GRCh38
rs948013316 Chr.9: 127087797-127087797 on Build GRCh38
rs530649450 Chr.9: 127087781-127087781 on Build GRCh38
rs192894514 Chr.9: 127087491-127087491 on Build GRCh38
rs12554810 Chr.9: 127087796-127087796 on Build GRCh38
rs376862055 Chr.9: 127087593-127087593 on Build GRCh38
rs544180766 Chr.9: 127087567-127087567 on Build GRCh38

View Full Product Details