Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101401490-101402081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLA
Entrez Gene ID
2717
Gene Name
galactosidase alpha
Gene Aliases
GALA
UniGene
Hs.69089
Gene Chromosome Location
Chr.X: 101397791-101408013 on Build GRCh38
Transcripts
NM_000169.2 NM_001199973.1 NM_001199974.1

Gene Symbol
RPL36A-HNRNPH2
Entrez Gene ID
100529097
Gene Name
RPL36A-HNRNPH2 readthrough
Gene Aliases
-
UniGene
Hs.432485
Gene Chromosome Location
Chr.X: 101390890-101414140 on Build GRCh38
Transcripts
NM_000169.2 NM_001199973.1 NM_001199974.1

SNP Detail

SNP ID Chromosome Location
rs781860029 Chr.X: 101401641-101401641 on Build GRCh38
rs730880445 Chr.X: 101401674-101401674 on Build GRCh38
rs727503951 Chr.X: 101401779-101401779 on Build GRCh38
rs782336447 Chr.X: 101401802-101401802 on Build GRCh38
rs869312320 Chr.X: 101401631-101401631 on Build GRCh38
rs113419388 Chr.X: 101401716-101401716 on Build GRCh38
rs869312143 Chr.X: 101401718-101401718 on Build GRCh38
rs782652843 Chr.X: 101401627-101401627 on Build GRCh38
rs782418101 Chr.X: 101401533-101401533 on Build GRCh38
rs869312299 Chr.X: 101401780-101401780 on Build GRCh38
rs869312317 Chr.X: 101401668-101401668 on Build GRCh38
rs869312305 Chr.X: 101401726-101401726 on Build GRCh38
rs77934640 Chr.X: 101401750-101401750 on Build GRCh38
rs782126435 Chr.X: 101401560-101401560 on Build GRCh38
rs961297357 Chr.X: 101401849-101401849 on Build GRCh38
rs782519616 Chr.X: 101401987-101401987 on Build GRCh38
rs730880444 Chr.X: 101401811-101401811 on Build GRCh38
rs398123210 Chr.X: 101401670-101401670 on Build GRCh38
rs782209213 Chr.X: 101402047-101402047 on Build GRCh38
rs181562693 Chr.X: 101401659-101401659 on Build GRCh38
rs869312315 Chr.X: 101401671-101401671 on Build GRCh38
rs869312287 Chr.X: 101401810-101401810 on Build GRCh38
rs869312300 Chr.X: 101401777-101401777 on Build GRCh38
rs531010133 Chr.X: 101401570-101401570 on Build GRCh38
rs916701411 Chr.X: 101402056-101402056 on Build GRCh38
rs782703603 Chr.X: 101401591-101401591 on Build GRCh38
rs869312323 Chr.X: 101401623-101401623 on Build GRCh38
rs869312306 Chr.X: 101401720-101401722 on Build GRCh38
rs886044845 Chr.X: 101401755-101401755 on Build GRCh38
rs869312290 Chr.X: 101401879-101401879 on Build GRCh38
rs782777183 Chr.X: 101401566-101401566 on Build GRCh38
rs28935196 Chr.X: 101401695-101401695 on Build GRCh38
rs138886989 Chr.X: 101401763-101401763 on Build GRCh38
rs781873631 Chr.X: 101401967-101401967 on Build GRCh38
rs1019520442 Chr.X: 101401840-101401840 on Build GRCh38
rs869312311 Chr.X: 101401693-101401693 on Build GRCh38
rs869312295 Chr.X: 101401898-101401898 on Build GRCh38
rs869312321 Chr.X: 101401626-101401631 on Build GRCh38
rs727503072 Chr.X: 101401793-101401793 on Build GRCh38
rs869312285 Chr.X: 101401823-101401823 on Build GRCh38
rs869312291 Chr.X: 101401885-101401889 on Build GRCh38
rs727504350 Chr.X: 101401694-101401694 on Build GRCh38
rs869312318 Chr.X: 101401664-101401664 on Build GRCh38
rs201275759 Chr.X: 101401605-101401605 on Build GRCh38
rs869312145 Chr.X: 101401639-101401639 on Build GRCh38
rs782343976 Chr.X: 101401525-101401525 on Build GRCh38
rs35549513 Chr.X: 101401901-101401901 on Build GRCh38
rs782810583 Chr.X: 101401818-101401818 on Build GRCh38
rs782722844 Chr.X: 101401654-101401654 on Build GRCh38
rs781833461 Chr.X: 101401855-101401855 on Build GRCh38
rs869312293 Chr.X: 101401889-101401893 on Build GRCh38
rs782079025 Chr.X: 101401584-101401584 on Build GRCh38
rs782195985 Chr.X: 101401885-101401886 on Build GRCh38
rs782486830 Chr.X: 101401678-101401678 on Build GRCh38
rs370299397 Chr.X: 101401722-101401722 on Build GRCh38
rs782164542 Chr.X: 101401815-101401815 on Build GRCh38
rs1011744471 Chr.X: 101401557-101401557 on Build GRCh38
rs782100696 Chr.X: 101401786-101401786 on Build GRCh38
rs869312312 Chr.X: 101401686-101401686 on Build GRCh38
rs869312286 Chr.X: 101401827-101401828 on Build GRCh38
rs104894845 Chr.X: 101401752-101401752 on Build GRCh38
rs782003690 Chr.X: 101401837-101401837 on Build GRCh38
rs782755334 Chr.X: 101401853-101401853 on Build GRCh38
rs782538815 Chr.X: 101401612-101401612 on Build GRCh38
rs28935195 Chr.X: 101401713-101401713 on Build GRCh38
rs869312298 Chr.X: 101401787-101401787 on Build GRCh38
rs782047833 Chr.X: 101401531-101401531 on Build GRCh38
rs149391489 Chr.X: 101401803-101401803 on Build GRCh38
rs782013736 Chr.X: 101401538-101401538 on Build GRCh38
rs869312296 Chr.X: 101401806-101401806 on Build GRCh38
rs869312288 Chr.X: 101401871-101401871 on Build GRCh38
rs869312301 Chr.X: 101401766-101401766 on Build GRCh38
rs869312324 Chr.X: 101401632-101401632 on Build GRCh38
rs150228150 Chr.X: 101401760-101401760 on Build GRCh38
rs782682748 Chr.X: 101401808-101401809 on Build GRCh38
rs869312319 Chr.X: 101401660-101401660 on Build GRCh38
rs782623177 Chr.X: 101401988-101401988 on Build GRCh38
rs869312307 Chr.X: 101401712-101401712 on Build GRCh38
rs104894837 Chr.X: 101401743-101401743 on Build GRCh38
rs782547736 Chr.X: 101401902-101401902 on Build GRCh38
rs782811871 Chr.X: 101401912-101401912 on Build GRCh38
rs869312314 Chr.X: 101401675-101401675 on Build GRCh38
rs869312294 Chr.X: 101401892-101401896 on Build GRCh38
rs991305661 Chr.X: 101401949-101401949 on Build GRCh38
rs869312322 Chr.X: 101401594-101401594 on Build GRCh38
rs3027588 Chr.X: 101401530-101401530 on Build GRCh38
rs782118672 Chr.X: 101401847-101401847 on Build GRCh38
rs1023432 Chr.X: 101401889-101401889 on Build GRCh38
rs869312144 Chr.X: 101401689-101401689 on Build GRCh38
rs797044702 Chr.X: 101401710-101401710 on Build GRCh38
rs869312289 Chr.X: 101401876-101401876 on Build GRCh38
rs886044843 Chr.X: 101401757-101401757 on Build GRCh38
rs782615630 Chr.X: 101401705-101401705 on Build GRCh38
rs186273861 Chr.X: 101401581-101401581 on Build GRCh38
rs869312302 Chr.X: 101401745-101401745 on Build GRCh38
rs869312292 Chr.X: 101401880-101401890 on Build GRCh38
rs869312284 Chr.X: 101401920-101401920 on Build GRCh38
rs869312308 Chr.X: 101401655-101401656 on Build GRCh38
rs782085638 Chr.X: 101401748-101401748 on Build GRCh38
rs782197072 Chr.X: 101401714-101401714 on Build GRCh38

View Full Product Details