Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 126551900-126552443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF12L1
Entrez Gene ID
139170
Gene Name
DDB1 and CUL4 associated factor 12 like 1
Gene Aliases
KIAA1892L, WDR40B
UniGene
Hs.120403
Gene Chromosome Location
Chr.X: 126549383-126552859 on Build GRCh38
Transcripts
NM_178470.4

SNP Detail

SNP ID Chromosome Location
rs377390890 Chr.X: 126552200-126552200 on Build GRCh38
rs778190369 Chr.X: 126552331-126552331 on Build GRCh38
rs375986905 Chr.X: 126551970-126551970 on Build GRCh38
rs770105970 Chr.X: 126551937-126551937 on Build GRCh38
rs199989323 Chr.X: 126552217-126552217 on Build GRCh38
rs765863389 Chr.X: 126552390-126552390 on Build GRCh38
rs777544582 Chr.X: 126552045-126552045 on Build GRCh38
rs763026132 Chr.X: 126551963-126551963 on Build GRCh38
rs764645023 Chr.X: 126551985-126551985 on Build GRCh38
rs775856177 Chr.X: 126551955-126551955 on Build GRCh38
rs902593684 Chr.X: 126552360-126552360 on Build GRCh38
rs769832852 Chr.X: 126552236-126552236 on Build GRCh38
rs748351807 Chr.X: 126552214-126552214 on Build GRCh38
rs775537701 Chr.X: 126552246-126552246 on Build GRCh38
rs780327912 Chr.X: 126552102-126552102 on Build GRCh38
rs752424251 Chr.X: 126552155-126552155 on Build GRCh38
rs370562344 Chr.X: 126552340-126552340 on Build GRCh38
rs747055344 Chr.X: 126552332-126552332 on Build GRCh38
rs776600036 Chr.X: 126552336-126552336 on Build GRCh38
rs866998454 Chr.X: 126551973-126551973 on Build GRCh38
rs780136639 Chr.X: 126552288-126552288 on Build GRCh38
rs757985768 Chr.X: 126552041-126552041 on Build GRCh38
rs373543121 Chr.X: 126552048-126552048 on Build GRCh38
rs746326807 Chr.X: 126552090-126552090 on Build GRCh38
rs144796770 Chr.X: 126552185-126552185 on Build GRCh38
rs779076294 Chr.X: 126552111-126552111 on Build GRCh38
rs763904128 Chr.X: 126552276-126552276 on Build GRCh38
rs201996325 Chr.X: 126552188-126552188 on Build GRCh38
rs929945930 Chr.X: 126552184-126552184 on Build GRCh38
rs774678665 Chr.X: 126552110-126552110 on Build GRCh38
rs373280004 Chr.X: 126552226-126552226 on Build GRCh38
rs755444137 Chr.X: 126552395-126552395 on Build GRCh38
rs772235462 Chr.X: 126552113-126552113 on Build GRCh38
rs374606828 Chr.X: 126552180-126552180 on Build GRCh38
rs753863510 Chr.X: 126552008-126552008 on Build GRCh38
rs761307271 Chr.X: 126551933-126551933 on Build GRCh38
rs887131101 Chr.X: 126552417-126552417 on Build GRCh38
rs766393165 Chr.X: 126551991-126551991 on Build GRCh38
rs41310460 Chr.X: 126552264-126552264 on Build GRCh38
rs911471419 Chr.X: 126552058-126552058 on Build GRCh38
rs752543292 Chr.X: 126552028-126552028 on Build GRCh38
rs145069543 Chr.X: 126552369-126552369 on Build GRCh38
rs755461394 Chr.X: 126552321-126552321 on Build GRCh38
rs768485642 Chr.X: 126552359-126552359 on Build GRCh38
rs375613009 Chr.X: 126552098-126552098 on Build GRCh38
rs867455944 Chr.X: 126552109-126552109 on Build GRCh38
rs149373470 Chr.X: 126552132-126552132 on Build GRCh38
rs369328900 Chr.X: 126552012-126552012 on Build GRCh38
rs754329629 Chr.X: 126552320-126552320 on Build GRCh38
rs780712210 Chr.X: 126552068-126552068 on Build GRCh38
rs758875700 Chr.X: 126552412-126552412 on Build GRCh38
rs759482810 Chr.X: 126552133-126552133 on Build GRCh38
rs749727856 Chr.X: 126552107-126552107 on Build GRCh38
rs1004432170 Chr.X: 126552422-126552422 on Build GRCh38
rs148550538 Chr.X: 126552206-126552206 on Build GRCh38
rs756928184 Chr.X: 126552199-126552199 on Build GRCh38
rs779175095 Chr.X: 126552213-126552213 on Build GRCh38
rs369850510 Chr.X: 126552141-126552141 on Build GRCh38
rs747145337 Chr.X: 126552231-126552231 on Build GRCh38
rs747060032 Chr.X: 126552303-126552303 on Build GRCh38
rs1021876212 Chr.X: 126551956-126551956 on Build GRCh38
rs749590330 Chr.X: 126552210-126552210 on Build GRCh38
rs758681741 Chr.X: 126552329-126552329 on Build GRCh38
rs377115767 Chr.X: 126552323-126552323 on Build GRCh38
rs762859614 Chr.X: 126552172-126552172 on Build GRCh38
rs967681645 Chr.X: 126551967-126551967 on Build GRCh38
rs150927042 Chr.X: 126552372-126552372 on Build GRCh38
rs780664644 Chr.X: 126552207-126552207 on Build GRCh38
rs758565746 Chr.X: 126552287-126552287 on Build GRCh38
rs1056266108 Chr.X: 126552219-126552219 on Build GRCh38
rs145669288 Chr.X: 126552318-126552318 on Build GRCh38
rs777172062 Chr.X: 126551928-126551928 on Build GRCh38
rs1022819725 Chr.X: 126552078-126552078 on Build GRCh38
rs767130770 Chr.X: 126552375-126552375 on Build GRCh38
rs907259332 Chr.X: 126552215-126552215 on Build GRCh38
rs772887112 Chr.X: 126552376-126552376 on Build GRCh38
rs769104621 Chr.X: 126552211-126552211 on Build GRCh38
rs771099108 Chr.X: 126552333-126552333 on Build GRCh38
rs140792759 Chr.X: 126552400-126552400 on Build GRCh38
rs139822084 Chr.X: 126552060-126552060 on Build GRCh38
rs778313079 Chr.X: 126552418-126552418 on Build GRCh38
rs986958118 Chr.X: 126552055-126552055 on Build GRCh38
rs753090078 Chr.X: 126552403-126552403 on Build GRCh38
rs768747321 Chr.X: 126552108-126552108 on Build GRCh38
rs760351168 Chr.X: 126552384-126552384 on Build GRCh38

View Full Product Details