Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 150814734-150815328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD99L2
Entrez Gene ID
83692
Gene Name
CD99 molecule like 2
Gene Aliases
CD99B, MIC2L1
UniGene
Hs.522805
Gene Chromosome Location
Chr.X: 150766336-150898816 on Build GRCh38
Transcripts
NM_001184808.1 NM_001242614.1 NM_031462.3 NM_134445.3 NM_134446.3 XM_017029890.1 XM_017029891.1

SNP Detail

SNP ID Chromosome Location
rs191303266 Chr.X: 150815287-150815287 on Build GRCh38
rs782431561 Chr.X: 150814822-150814822 on Build GRCh38
rs141111348 Chr.X: 150814926-150814926 on Build GRCh38
rs151085209 Chr.X: 150814907-150814907 on Build GRCh38
rs371316554 Chr.X: 150814990-150814990 on Build GRCh38
rs781813833 Chr.X: 150814961-150814961 on Build GRCh38
rs367713103 Chr.X: 150814947-150814947 on Build GRCh38
rs782673799 Chr.X: 150814996-150814996 on Build GRCh38
rs942796684 Chr.X: 150815156-150815156 on Build GRCh38
rs781823345 Chr.X: 150815185-150815185 on Build GRCh38
rs368278860 Chr.X: 150814838-150814838 on Build GRCh38
rs782012430 Chr.X: 150814906-150814906 on Build GRCh38
rs782762279 Chr.X: 150814932-150814932 on Build GRCh38
rs782518983 Chr.X: 150815119-150815119 on Build GRCh38
rs782728279 Chr.X: 150815057-150815057 on Build GRCh38
rs782550171 Chr.X: 150814942-150814942 on Build GRCh38
rs782201225 Chr.X: 150814871-150814871 on Build GRCh38
rs368595725 Chr.X: 150814804-150814804 on Build GRCh38
rs912640008 Chr.X: 150815147-150815147 on Build GRCh38
rs782521465 Chr.X: 150814872-150814872 on Build GRCh38
rs374445547 Chr.X: 150814890-150814890 on Build GRCh38
rs577351043 Chr.X: 150814878-150814878 on Build GRCh38
rs782443250 Chr.X: 150814974-150814974 on Build GRCh38
rs902449988 Chr.X: 150815295-150815295 on Build GRCh38
rs782818628 Chr.X: 150814896-150814896 on Build GRCh38
rs1039725934 Chr.X: 150815165-150815165 on Build GRCh38
rs201325686 Chr.X: 150814981-150814981 on Build GRCh38
rs782412229 Chr.X: 150814880-150814880 on Build GRCh38
rs782639257 Chr.X: 150815139-150815139 on Build GRCh38
rs782393976 Chr.X: 150814995-150814995 on Build GRCh38
rs781862677 Chr.X: 150814938-150814938 on Build GRCh38
rs782695723 Chr.X: 150814912-150814912 on Build GRCh38
rs782272226 Chr.X: 150814998-150814998 on Build GRCh38
rs782619126 Chr.X: 150814977-150814977 on Build GRCh38
rs781803873 Chr.X: 150814821-150814821 on Build GRCh38
rs782485088 Chr.X: 150815195-150815195 on Build GRCh38
rs781907198 Chr.X: 150815009-150815009 on Build GRCh38
rs781873877 Chr.X: 150814982-150814982 on Build GRCh38
rs781889438 Chr.X: 150814879-150814879 on Build GRCh38
rs781837500 Chr.X: 150814913-150814913 on Build GRCh38
rs987398812 Chr.X: 150815086-150815086 on Build GRCh38
rs782224638 Chr.X: 150815202-150815202 on Build GRCh38
rs146438753 Chr.X: 150815211-150815211 on Build GRCh38

View Full Product Details