Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83461320-83461873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175A
Entrez Gene ID
84142
Gene Name
family with sequence similarity 175 member A
Gene Aliases
ABRA1, CCDC98
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83459515-83485181 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1 NM_001297767.1 NM_001297769.1 NM_001297770.1 NM_016067.3

Gene Symbol
MRPS18C
Entrez Gene ID
51023
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
CGI-134, MRP-S18-1, MRP-S18-c, MRPS18-1, S18mt-c, mrps18-c
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83455932-83461723 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1 NM_001297767.1 NM_001297769.1 NM_001297770.1 NM_016067.3

SNP Detail

SNP ID Chromosome Location
rs1003456050 Chr.4: 83461517-83461517 on Build GRCh38
rs775659975 Chr.4: 83461428-83461428 on Build GRCh38
rs748643690 Chr.4: 83461773-83461773 on Build GRCh38
rs137940592 Chr.4: 83461544-83461544 on Build GRCh38
rs906388655 Chr.4: 83461510-83461510 on Build GRCh38
rs189572362 Chr.4: 83461792-83461792 on Build GRCh38
rs184785302 Chr.4: 83461513-83461513 on Build GRCh38
rs910795900 Chr.4: 83461712-83461712 on Build GRCh38
rs749646384 Chr.4: 83461530-83461530 on Build GRCh38
rs1026468152 Chr.4: 83461672-83461672 on Build GRCh38
rs879067054 Chr.4: 83461375-83461375 on Build GRCh38
rs796294567 Chr.4: 83461842-83461845 on Build GRCh38
rs893908611 Chr.4: 83461414-83461414 on Build GRCh38
rs770052837 Chr.4: 83461425-83461425 on Build GRCh38
rs150152879 Chr.4: 83461665-83461665 on Build GRCh38
rs374979187 Chr.4: 83461845-83461848 on Build GRCh38
rs1005229888 Chr.4: 83461568-83461568 on Build GRCh38
rs769016871 Chr.4: 83461536-83461536 on Build GRCh38
rs915269046 Chr.4: 83461789-83461789 on Build GRCh38
rs140997571 Chr.4: 83461661-83461661 on Build GRCh38
rs375716331 Chr.4: 83461714-83461714 on Build GRCh38
rs191585231 Chr.4: 83461683-83461683 on Build GRCh38
rs11099601 Chr.4: 83461610-83461610 on Build GRCh38
rs910543125 Chr.4: 83461395-83461395 on Build GRCh38
rs945452909 Chr.4: 83461801-83461801 on Build GRCh38
rs1126971 Chr.4: 83461399-83461399 on Build GRCh38
rs1034052383 Chr.4: 83461554-83461554 on Build GRCh38
rs768123031 Chr.4: 83461635-83461635 on Build GRCh38
rs989459128 Chr.4: 83461762-83461762 on Build GRCh38
rs187712373 Chr.4: 83461602-83461602 on Build GRCh38
rs184898857 Chr.4: 83461697-83461697 on Build GRCh38
rs1052473526 Chr.4: 83461370-83461370 on Build GRCh38
rs1042207215 Chr.4: 83461469-83461469 on Build GRCh38
rs774724187 Chr.4: 83461609-83461609 on Build GRCh38
rs894774303 Chr.4: 83461565-83461565 on Build GRCh38
rs77519137 Chr.4: 83461397-83461397 on Build GRCh38
rs945189705 Chr.4: 83461444-83461444 on Build GRCh38
rs192857866 Chr.4: 83461493-83461493 on Build GRCh38

View Full Product Details