Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 98880921-98881475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E
Entrez Gene ID
1977
Gene Name
eukaryotic translation initiation factor 4E
Gene Aliases
AUTS19, CBP, EIF4E1, EIF4EL1, EIF4F, eIF-4E
UniGene
Hs.249718
Gene Chromosome Location
Chr.4: 98878456-98930635 on Build GRCh38
Transcripts
NM_001130678.1 NM_001130679.1 NM_001968.3 XM_006714126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195655 c.572C>T p.P191L Chr.4: 98881110-98881110 on Build GRCh38
6405086 c.557G>A p.R186K Chr.4: 98881125-98881125 on Build GRCh38
1619267 c.540-8delT p.? Chr.4: 98881150-98881150 on Build GRCh38
1059185 c.624C>T p.G208G Chr.4: 98881058-98881058 on Build GRCh38
6972897 c.587G>C p.G196A Chr.4: 98881095-98881095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013823891 Chr.4: 98881377-98881377 on Build GRCh38
rs754660302 Chr.4: 98881150-98881151 on Build GRCh38
rs768240575 Chr.4: 98881127-98881127 on Build GRCh38
rs935434248 Chr.4: 98881396-98881396 on Build GRCh38
rs967400435 Chr.4: 98881099-98881099 on Build GRCh38
rs768419229 Chr.4: 98880982-98880982 on Build GRCh38
rs898424280 Chr.4: 98881269-98881269 on Build GRCh38
rs763285621 Chr.4: 98881168-98881168 on Build GRCh38
rs113849091 Chr.4: 98881161-98881161 on Build GRCh38
rs572665562 Chr.4: 98881330-98881330 on Build GRCh38
rs184309947 Chr.4: 98881246-98881246 on Build GRCh38
rs759973110 Chr.4: 98881149-98881149 on Build GRCh38
rs553664019 Chr.4: 98881160-98881161 on Build GRCh38
rs763230565 Chr.4: 98881028-98881028 on Build GRCh38
rs903935241 Chr.4: 98881358-98881358 on Build GRCh38
rs751767073 Chr.4: 98881175-98881175 on Build GRCh38
rs144779880 Chr.4: 98881124-98881124 on Build GRCh38
rs112318982 Chr.4: 98881224-98881224 on Build GRCh38
rs911035815 Chr.4: 98881335-98881335 on Build GRCh38
rs905325032 Chr.4: 98881429-98881429 on Build GRCh38
rs200056873 Chr.4: 98881158-98881161 on Build GRCh38
rs775892925 Chr.4: 98881160-98881160 on Build GRCh38
rs778594185 Chr.4: 98881129-98881129 on Build GRCh38
rs780061762 Chr.4: 98881119-98881119 on Build GRCh38
rs201876303 Chr.4: 98881426-98881431 on Build GRCh38
rs1043777028 Chr.4: 98881356-98881356 on Build GRCh38
rs770285965 Chr.4: 98881158-98881158 on Build GRCh38
rs748882618 Chr.4: 98880981-98880981 on Build GRCh38
rs779529404 Chr.4: 98881150-98881150 on Build GRCh38
rs771379922 Chr.4: 98881012-98881012 on Build GRCh38
rs554415891 Chr.4: 98881321-98881321 on Build GRCh38
rs1024319694 Chr.4: 98881428-98881428 on Build GRCh38
rs751640787 Chr.4: 98881069-98881069 on Build GRCh38
rs1051479041 Chr.4: 98881279-98881279 on Build GRCh38
rs576392381 Chr.4: 98881423-98881423 on Build GRCh38
rs201272862 Chr.4: 98881159-98881161 on Build GRCh38
rs200243582 Chr.4: 98881081-98881081 on Build GRCh38
rs1037990923 Chr.4: 98881250-98881250 on Build GRCh38
rs562194458 Chr.4: 98880979-98880979 on Build GRCh38
rs745946178 Chr.4: 98881425-98881426 on Build GRCh38
rs796969921 Chr.4: 98881160-98881161 on Build GRCh38
rs555853444 Chr.4: 98881265-98881265 on Build GRCh38
rs777151481 Chr.4: 98881021-98881021 on Build GRCh38
rs374891583 Chr.4: 98881149-98881150 on Build GRCh38
rs762398533 Chr.4: 98881357-98881357 on Build GRCh38
rs759913560 Chr.4: 98881022-98881022 on Build GRCh38
rs755977303 Chr.4: 98881105-98881105 on Build GRCh38
rs1024417404 Chr.4: 98881388-98881388 on Build GRCh38
rs535497182 Chr.4: 98881307-98881307 on Build GRCh38
rs747639784 Chr.4: 98881131-98881131 on Build GRCh38
rs757955938 Chr.4: 98881024-98881025 on Build GRCh38
rs79710293 Chr.4: 98881213-98881213 on Build GRCh38
rs761363263 Chr.4: 98881003-98881003 on Build GRCh38
rs558465706 Chr.4: 98881387-98881387 on Build GRCh38
rs764367114 Chr.4: 98881060-98881060 on Build GRCh38
rs1023785703 Chr.4: 98881153-98881153 on Build GRCh38
rs781023498 Chr.4: 98881082-98881082 on Build GRCh38
rs546566408 Chr.4: 98881386-98881386 on Build GRCh38
rs777133297 Chr.4: 98881140-98881140 on Build GRCh38
rs767577200 Chr.4: 98881177-98881177 on Build GRCh38
rs1002036762 Chr.4: 98881283-98881283 on Build GRCh38
rs181201776 Chr.4: 98881194-98881194 on Build GRCh38
rs886837703 Chr.4: 98881236-98881236 on Build GRCh38
rs527622522 Chr.4: 98881214-98881214 on Build GRCh38
rs568169675 Chr.4: 98881281-98881281 on Build GRCh38
rs750338066 Chr.4: 98881085-98881085 on Build GRCh38
rs149506945 Chr.4: 98881427-98881427 on Build GRCh38
rs1052419721 Chr.4: 98881406-98881406 on Build GRCh38
rs933913145 Chr.4: 98881271-98881271 on Build GRCh38
rs138645913 Chr.4: 98881138-98881138 on Build GRCh38
rs749910948 Chr.4: 98881448-98881448 on Build GRCh38
rs11729285 Chr.4: 98881244-98881244 on Build GRCh38
rs752925618 Chr.4: 98881027-98881027 on Build GRCh38
rs967452929 Chr.4: 98881405-98881407 on Build GRCh38
rs773789119 Chr.4: 98880993-98880993 on Build GRCh38
rs942542636 Chr.4: 98881349-98881349 on Build GRCh38
rs765530897 Chr.4: 98881023-98881023 on Build GRCh38
rs764265247 Chr.4: 98881170-98881170 on Build GRCh38
rs962315517 Chr.4: 98881221-98881221 on Build GRCh38
rs960540761 Chr.4: 98881361-98881361 on Build GRCh38

View Full Product Details