Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83075495-83076037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS4
Entrez Gene ID
51138
Gene Name
COP9 signalosome subunit 4
Gene Aliases
CSN4
UniGene
Hs.190384
Gene Chromosome Location
Chr.4: 83035086-83075818 on Build GRCh38
Transcripts
NM_001258006.1 NM_016129.2 XM_011532019.1

SNP Detail

SNP ID Chromosome Location
rs577634815 Chr.4: 83075550-83075550 on Build GRCh38
rs1024809751 Chr.4: 83075898-83075898 on Build GRCh38
rs942192658 Chr.4: 83075625-83075625 on Build GRCh38
rs931345725 Chr.4: 83075598-83075598 on Build GRCh38
rs188159951 Chr.4: 83075579-83075579 on Build GRCh38
rs963693131 Chr.4: 83075944-83075944 on Build GRCh38
rs937340694 Chr.4: 83075572-83075572 on Build GRCh38
rs11944557 Chr.4: 83075876-83075876 on Build GRCh38
rs909469492 Chr.4: 83075602-83075602 on Build GRCh38
rs1027052343 Chr.4: 83075964-83075964 on Build GRCh38
rs944260615 Chr.4: 83075659-83075659 on Build GRCh38
rs536336406 Chr.4: 83075790-83075790 on Build GRCh38
rs1020812982 Chr.4: 83075833-83075833 on Build GRCh38
rs895075053 Chr.4: 83075875-83075875 on Build GRCh38
rs763556381 Chr.4: 83075906-83075907 on Build GRCh38
rs903593397 Chr.4: 83075824-83075824 on Build GRCh38
rs961541984 Chr.4: 83075903-83075903 on Build GRCh38
rs1002933456 Chr.4: 83075870-83075870 on Build GRCh38
rs933498345 Chr.4: 83076013-83076013 on Build GRCh38
rs143607253 Chr.4: 83075642-83075642 on Build GRCh38
rs774763797 Chr.4: 83075821-83075821 on Build GRCh38
rs559170493 Chr.4: 83075936-83075936 on Build GRCh38
rs1041227199 Chr.4: 83075661-83075661 on Build GRCh38
rs745995717 Chr.4: 83075536-83075536 on Build GRCh38
rs981835045 Chr.4: 83075534-83075535 on Build GRCh38
rs565782529 Chr.4: 83075653-83075653 on Build GRCh38
rs763248076 Chr.4: 83075699-83075699 on Build GRCh38
rs145220085 Chr.4: 83075907-83075907 on Build GRCh38
rs1000556786 Chr.4: 83075847-83075847 on Build GRCh38
rs897558715 Chr.4: 83075647-83075647 on Build GRCh38
rs961491424 Chr.4: 83075576-83075576 on Build GRCh38
rs972750696 Chr.4: 83075587-83075587 on Build GRCh38
rs768835491 Chr.4: 83075766-83075766 on Build GRCh38
rs11947785 Chr.4: 83076005-83076005 on Build GRCh38
rs1007188727 Chr.4: 83075804-83075804 on Build GRCh38
rs888753503 Chr.4: 83075798-83075798 on Build GRCh38
rs991528991 Chr.4: 83075916-83075916 on Build GRCh38
rs1032180313 Chr.4: 83075820-83075820 on Build GRCh38
rs952777425 Chr.4: 83075976-83075976 on Build GRCh38
rs920006209 Chr.4: 83075592-83075592 on Build GRCh38
rs577479421 Chr.4: 83075997-83075997 on Build GRCh38
rs946233976 Chr.4: 83075750-83075750 on Build GRCh38
rs544944554 Chr.4: 83076003-83076003 on Build GRCh38
rs992022322 Chr.4: 83075528-83075528 on Build GRCh38
rs747858374 Chr.4: 83075921-83075928 on Build GRCh38
rs571464490 Chr.4: 83075533-83075533 on Build GRCh38
rs138982593 Chr.4: 83075848-83075848 on Build GRCh38
rs911180647 Chr.4: 83076011-83076011 on Build GRCh38
rs1025198406 Chr.4: 83075555-83075555 on Build GRCh38
rs537269640 Chr.4: 83075918-83075918 on Build GRCh38

View Full Product Details