Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112638106-112638694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC80
Entrez Gene ID
151887
Gene Name
coiled-coil domain containing 80
Gene Aliases
DRO1, SSG1, URB, okuribin
UniGene
Hs.477128
Gene Chromosome Location
Chr.3: 112604386-112641143 on Build GRCh38
Transcripts
NM_199511.2 NM_199512.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327974 c.1666A>T p.N556Y Chr.3: 112638240-112638240 on Build GRCh38
445309 c.1630G>C p.E544Q Chr.3: 112638276-112638276 on Build GRCh38
6219094 c.1561G>T p.E521* Chr.3: 112638345-112638345 on Build GRCh38
727221 c.1390C>T p.R464W Chr.3: 112638516-112638516 on Build GRCh38
4910157 c.1444C>T p.P482S Chr.3: 112638462-112638462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769656953 Chr.3: 112638495-112638495 on Build GRCh38
rs779073205 Chr.3: 112638193-112638195 on Build GRCh38
rs1036673507 Chr.3: 112638162-112638162 on Build GRCh38
rs534306920 Chr.3: 112638667-112638667 on Build GRCh38
rs144252779 Chr.3: 112638309-112638309 on Build GRCh38
rs760102914 Chr.3: 112638455-112638455 on Build GRCh38
rs770453194 Chr.3: 112638631-112638631 on Build GRCh38
rs958169301 Chr.3: 112638298-112638298 on Build GRCh38
rs774414772 Chr.3: 112638397-112638397 on Build GRCh38
rs754415158 Chr.3: 112638449-112638449 on Build GRCh38
rs777714728 Chr.3: 112638365-112638365 on Build GRCh38
rs747318222 Chr.3: 112638592-112638592 on Build GRCh38
rs201190794 Chr.3: 112638574-112638574 on Build GRCh38
rs763087683 Chr.3: 112638151-112638151 on Build GRCh38
rs1048515481 Chr.3: 112638139-112638139 on Build GRCh38
rs760782964 Chr.3: 112638505-112638505 on Build GRCh38
rs776851723 Chr.3: 112638515-112638515 on Build GRCh38
rs376265934 Chr.3: 112638496-112638496 on Build GRCh38
rs756595001 Chr.3: 112638321-112638321 on Build GRCh38
rs773123702 Chr.3: 112638235-112638235 on Build GRCh38
rs746184884 Chr.3: 112638629-112638629 on Build GRCh38
rs376493692 Chr.3: 112638645-112638645 on Build GRCh38
rs745884821 Chr.3: 112638494-112638494 on Build GRCh38
rs771380755 Chr.3: 112638506-112638506 on Build GRCh38
rs200296261 Chr.3: 112638347-112638347 on Build GRCh38
rs752186550 Chr.3: 112638473-112638473 on Build GRCh38
rs757283225 Chr.3: 112638144-112638145 on Build GRCh38
rs745961319 Chr.3: 112638591-112638593 on Build GRCh38
rs768783026 Chr.3: 112638385-112638385 on Build GRCh38
rs772041379 Chr.3: 112638232-112638232 on Build GRCh38
rs773474180 Chr.3: 112638414-112638414 on Build GRCh38
rs199850583 Chr.3: 112638536-112638536 on Build GRCh38
rs116307644 Chr.3: 112638489-112638489 on Build GRCh38
rs777697705 Chr.3: 112638583-112638583 on Build GRCh38
rs35149092 Chr.3: 112638130-112638130 on Build GRCh38
rs1055570649 Chr.3: 112638212-112638212 on Build GRCh38
rs142225247 Chr.3: 112638286-112638286 on Build GRCh38
rs770832651 Chr.3: 112638265-112638265 on Build GRCh38
rs750253669 Chr.3: 112638660-112638660 on Build GRCh38
rs752276251 Chr.3: 112638522-112638522 on Build GRCh38
rs966948299 Chr.3: 112638573-112638573 on Build GRCh38
rs753317240 Chr.3: 112638329-112638329 on Build GRCh38
rs145200206 Chr.3: 112638231-112638231 on Build GRCh38
rs138501198 Chr.3: 112638521-112638521 on Build GRCh38
rs770807716 Chr.3: 112638366-112638366 on Build GRCh38
rs758118294 Chr.3: 112638357-112638357 on Build GRCh38
rs376955776 Chr.3: 112638433-112638433 on Build GRCh38
rs754143659 Chr.3: 112638133-112638137 on Build GRCh38
rs895568130 Chr.3: 112638523-112638523 on Build GRCh38
rs58031351 Chr.3: 112638314-112638314 on Build GRCh38
rs569499375 Chr.3: 112638542-112638542 on Build GRCh38
rs148461149 Chr.3: 112638197-112638197 on Build GRCh38
rs145579533 Chr.3: 112638179-112638179 on Build GRCh38
rs150318536 Chr.3: 112638486-112638486 on Build GRCh38
rs762009043 Chr.3: 112638289-112638289 on Build GRCh38
rs370640914 Chr.3: 112638538-112638538 on Build GRCh38
rs140232948 Chr.3: 112638447-112638447 on Build GRCh38
rs765287493 Chr.3: 112638141-112638141 on Build GRCh38
rs1045851177 Chr.3: 112638432-112638432 on Build GRCh38
rs995218613 Chr.3: 112638213-112638213 on Build GRCh38
rs146203552 Chr.3: 112638196-112638196 on Build GRCh38
rs771072496 Chr.3: 112638158-112638158 on Build GRCh38
rs767940513 Chr.3: 112638301-112638301 on Build GRCh38
rs772313881 Chr.3: 112638502-112638502 on Build GRCh38
rs754571897 Chr.3: 112638339-112638339 on Build GRCh38
rs529604031 Chr.3: 112638534-112638534 on Build GRCh38
rs774394821 Chr.3: 112638516-112638516 on Build GRCh38
rs749189595 Chr.3: 112638560-112638560 on Build GRCh38
rs748776813 Chr.3: 112638205-112638205 on Build GRCh38
rs777067306 Chr.3: 112638617-112638617 on Build GRCh38
rs371496097 Chr.3: 112638650-112638650 on Build GRCh38
rs145745813 Chr.3: 112638497-112638497 on Build GRCh38
rs773521549 Chr.3: 112638503-112638503 on Build GRCh38
rs937115514 Chr.3: 112638519-112638519 on Build GRCh38
rs372294401 Chr.3: 112638651-112638651 on Build GRCh38
rs763490845 Chr.3: 112638532-112638532 on Build GRCh38
rs762594374 Chr.3: 112638530-112638530 on Build GRCh38
rs763727517 Chr.3: 112638648-112638648 on Build GRCh38
rs371056820 Chr.3: 112638380-112638380 on Build GRCh38
rs775631710 Chr.3: 112638145-112638145 on Build GRCh38
rs527428637 Chr.3: 112638379-112638379 on Build GRCh38
rs377187744 Chr.3: 112638193-112638193 on Build GRCh38
rs928811113 Chr.3: 112638471-112638471 on Build GRCh38
rs767411894 Chr.3: 112638611-112638611 on Build GRCh38
rs373216819 Chr.3: 112638480-112638480 on Build GRCh38
rs903114682 Chr.3: 112638565-112638565 on Build GRCh38
rs766863633 Chr.3: 112638322-112638322 on Build GRCh38
rs750164014 Chr.3: 112638539-112638539 on Build GRCh38
rs187703841 Chr.3: 112638259-112638259 on Build GRCh38
rs764253810 Chr.3: 112638154-112638154 on Build GRCh38
rs58622942 Chr.3: 112638312-112638312 on Build GRCh38
rs151141158 Chr.3: 112638398-112638398 on Build GRCh38
rs770991717 Chr.3: 112638599-112638599 on Build GRCh38
rs775443915 Chr.3: 112638270-112638270 on Build GRCh38
rs137969304 Chr.3: 112638488-112638488 on Build GRCh38
rs763049995 Chr.3: 112638280-112638280 on Build GRCh38
rs116334737 Chr.3: 112638668-112638668 on Build GRCh38
rs544919809 Chr.3: 112638340-112638340 on Build GRCh38
rs767850556 Chr.3: 112638189-112638189 on Build GRCh38
rs772227726 Chr.3: 112638413-112638413 on Build GRCh38
rs749986115 Chr.3: 112638637-112638637 on Build GRCh38
rs143049160 Chr.3: 112638652-112638652 on Build GRCh38
rs758136333 Chr.3: 112638479-112638479 on Build GRCh38
rs754607927 Chr.3: 112638227-112638227 on Build GRCh38
rs1014035278 Chr.3: 112638229-112638229 on Build GRCh38

View Full Product Details