Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175882465-175883063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX2
Entrez Gene ID
10814
Gene Name
complexin 2
Gene Aliases
921-L, CPX-2, CPX2, Hfb1
UniGene
Hs.193235
Gene Chromosome Location
Chr.5: 175796539-175884021 on Build GRCh38
Transcripts
NM_001008220.1 NM_006650.3 XM_005265798.1 XM_005265799.1 XM_011534419.1 XM_017008964.1

SNP Detail

SNP ID Chromosome Location
rs915826142 Chr.5: 175882533-175882533 on Build GRCh38
rs898249047 Chr.5: 175883038-175883038 on Build GRCh38
rs996965426 Chr.5: 175882921-175882921 on Build GRCh38
rs991103070 Chr.5: 175882834-175882834 on Build GRCh38
rs780259528 Chr.5: 175882850-175882850 on Build GRCh38
rs1019756078 Chr.5: 175882742-175882742 on Build GRCh38
rs925726146 Chr.5: 175882763-175882763 on Build GRCh38
rs966786165 Chr.5: 175882815-175882815 on Build GRCh38
rs958062514 Chr.5: 175882894-175882894 on Build GRCh38
rs557239635 Chr.5: 175882592-175882592 on Build GRCh38
rs978105390 Chr.5: 175882508-175882508 on Build GRCh38
rs1008447935 Chr.5: 175882951-175882951 on Build GRCh38
rs886981627 Chr.5: 175882521-175882521 on Build GRCh38
rs753755456 Chr.5: 175882503-175882503 on Build GRCh38
rs1045562953 Chr.5: 175882520-175882520 on Build GRCh38
rs898475676 Chr.5: 175882542-175882542 on Build GRCh38
rs187275728 Chr.5: 175883009-175883009 on Build GRCh38
rs543471301 Chr.5: 175882865-175882865 on Build GRCh38
rs935778538 Chr.5: 175882786-175882786 on Build GRCh38
rs1008825109 Chr.5: 175882741-175882741 on Build GRCh38
rs990815607 Chr.5: 175882895-175882895 on Build GRCh38
rs376280483 Chr.5: 175882772-175882772 on Build GRCh38
rs143025190 Chr.5: 175882947-175882947 on Build GRCh38
rs939866193 Chr.5: 175882876-175882876 on Build GRCh38
rs894379225 Chr.5: 175882497-175882497 on Build GRCh38
rs761104751 Chr.5: 175882814-175882814 on Build GRCh38
rs56873824 Chr.5: 175882598-175882598 on Build GRCh38
rs570040705 Chr.5: 175883029-175883029 on Build GRCh38
rs967290087 Chr.5: 175882658-175882658 on Build GRCh38
rs1021639579 Chr.5: 175882961-175882961 on Build GRCh38
rs949996497 Chr.5: 175882851-175882851 on Build GRCh38
rs1004265623 Chr.5: 175882522-175882522 on Build GRCh38
rs996831542 Chr.5: 175882556-175882556 on Build GRCh38
rs1021081322 Chr.5: 175882650-175882650 on Build GRCh38
rs987497374 Chr.5: 175882640-175882640 on Build GRCh38
rs758699368 Chr.5: 175882797-175882797 on Build GRCh38
rs939783012 Chr.5: 175882964-175882964 on Build GRCh38
rs1000972303 Chr.5: 175883035-175883035 on Build GRCh38
rs761017833 Chr.5: 175882888-175882888 on Build GRCh38
rs572157919 Chr.5: 175882568-175882568 on Build GRCh38
rs1045558373 Chr.5: 175882546-175882546 on Build GRCh38
rs978136487 Chr.5: 175882839-175882840 on Build GRCh38
rs966882232 Chr.5: 175883026-175883026 on Build GRCh38
rs528997079 Chr.5: 175882898-175882898 on Build GRCh38
rs1046064843 Chr.5: 175882852-175882852 on Build GRCh38
rs898134402 Chr.5: 175882621-175882621 on Build GRCh38
rs183499703 Chr.5: 175882892-175882892 on Build GRCh38
rs1036660782 Chr.5: 175882611-175882611 on Build GRCh38
rs1036776689 Chr.5: 175883011-175883011 on Build GRCh38
rs747648896 Chr.5: 175882614-175882614 on Build GRCh38
rs559703128 Chr.5: 175882933-175882933 on Build GRCh38
rs3749801 Chr.5: 175882537-175882537 on Build GRCh38
rs536677950 Chr.5: 175882743-175882743 on Build GRCh38
rs778586832 Chr.5: 175882525-175882525 on Build GRCh38
rs977113747 Chr.5: 175882675-175882675 on Build GRCh38
rs3199427 Chr.5: 175882918-175882918 on Build GRCh38
rs564413988 Chr.5: 175882616-175882616 on Build GRCh38

View Full Product Details