Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30594514-30595012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GARS
Entrez Gene ID
2617
Gene Name
glycyl-tRNA synthetase
Gene Aliases
CMT2D, DSMAV, GlyRS, HMN5, SMAD1
UniGene
Hs.404321
Gene Chromosome Location
Chr.7: 30594735-30634033 on Build GRCh38
Transcripts
NM_001316772.1 NM_002047.3 XM_006715686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422433 c.83C>T p.P28L Chr.7: 30595004-30595004 on Build GRCh38
4881718 c.13C>T p.R5C Chr.7: 30594934-30594934 on Build GRCh38
1724763 c.46_48delCTG p.L20delL Chr.7: 30594967-30594969 on Build GRCh38
3028249 c.59T>C p.L20P Chr.7: 30594980-30594980 on Build GRCh38
4971540 c.59_61delTGC p.L20delL Chr.7: 30594980-30594982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562788409 Chr.7: 30594807-30594807 on Build GRCh38
rs529166485 Chr.7: 30594918-30594918 on Build GRCh38
rs527307703 Chr.7: 30594853-30594853 on Build GRCh38
rs948489135 Chr.7: 30594856-30594856 on Build GRCh38
rs759734139 Chr.7: 30594859-30594859 on Build GRCh38
rs753413723 Chr.7: 30594960-30594960 on Build GRCh38
rs17159262 Chr.7: 30594717-30594717 on Build GRCh38
rs540431164 Chr.7: 30594685-30594685 on Build GRCh38
rs1040518108 Chr.7: 30594764-30594764 on Build GRCh38
rs985496696 Chr.7: 30594966-30594966 on Build GRCh38
rs534002285 Chr.7: 30594735-30594735 on Build GRCh38
rs1040174267 Chr.7: 30594874-30594874 on Build GRCh38
rs1054637945 Chr.7: 30594884-30594884 on Build GRCh38
rs1022351707 Chr.7: 30594785-30594785 on Build GRCh38
rs150213018 Chr.7: 30594967-30594969 on Build GRCh38
rs545878519 Chr.7: 30594870-30594870 on Build GRCh38
rs987352468 Chr.7: 30594701-30594701 on Build GRCh38
rs991714987 Chr.7: 30594687-30594687 on Build GRCh38
rs902970712 Chr.7: 30594769-30594769 on Build GRCh38
rs766172101 Chr.7: 30594959-30594959 on Build GRCh38
rs886062270 Chr.7: 30594842-30594842 on Build GRCh38
rs999019691 Chr.7: 30594779-30594779 on Build GRCh38
rs560069959 Chr.7: 30594831-30594831 on Build GRCh38
rs2970504 Chr.7: 30594705-30594705 on Build GRCh38
rs892811963 Chr.7: 30594808-30594808 on Build GRCh38
rs932663016 Chr.7: 30594858-30594858 on Build GRCh38
rs371124400 Chr.7: 30594713-30594713 on Build GRCh38
rs910505084 Chr.7: 30594866-30594866 on Build GRCh38
rs968133921 Chr.7: 30594791-30594791 on Build GRCh38
rs575067868 Chr.7: 30594812-30594812 on Build GRCh38
rs1050218200 Chr.7: 30594584-30594584 on Build GRCh38
rs983688104 Chr.7: 30594611-30594623 on Build GRCh38
rs886062271 Chr.7: 30594863-30594863 on Build GRCh38
rs995164000 Chr.7: 30594568-30594568 on Build GRCh38
rs780383773 Chr.7: 30594988-30594988 on Build GRCh38
rs777045330 Chr.7: 30594837-30594837 on Build GRCh38
rs374341679 Chr.7: 30594746-30594746 on Build GRCh38
rs899120209 Chr.7: 30594563-30594563 on Build GRCh38
rs956087619 Chr.7: 30594621-30594621 on Build GRCh38
rs1044146793 Chr.7: 30594861-30594861 on Build GRCh38
rs762106233 Chr.7: 30594945-30594945 on Build GRCh38
rs773138162 Chr.7: 30594947-30594947 on Build GRCh38
rs774753271 Chr.7: 30594943-30594943 on Build GRCh38
rs367989555 Chr.7: 30594640-30594640 on Build GRCh38
rs1008764831 Chr.7: 30594756-30594756 on Build GRCh38
rs760759910 Chr.7: 30594966-30594967 on Build GRCh38
rs78980639 Chr.7: 30594745-30594745 on Build GRCh38
rs988228020 Chr.7: 30594631-30594631 on Build GRCh38
rs762624758 Chr.7: 30594990-30594990 on Build GRCh38
rs750436094 Chr.7: 30594900-30594900 on Build GRCh38
rs765145813 Chr.7: 30594885-30594885 on Build GRCh38
rs1002774216 Chr.7: 30594765-30594765 on Build GRCh38
rs760668265 Chr.7: 30594952-30594952 on Build GRCh38
rs940068066 Chr.7: 30594543-30594543 on Build GRCh38
rs888962455 Chr.7: 30594597-30594597 on Build GRCh38
rs73294431 Chr.7: 30594642-30594642 on Build GRCh38
rs907427143 Chr.7: 30594912-30594912 on Build GRCh38
rs1011456833 Chr.7: 30594781-30594781 on Build GRCh38
rs576984233 Chr.7: 30594753-30594753 on Build GRCh38
rs971563727 Chr.7: 30594703-30594703 on Build GRCh38
rs948354997 Chr.7: 30594747-30594747 on Build GRCh38
rs368574634 Chr.7: 30594973-30594973 on Build GRCh38
rs768665866 Chr.7: 30594993-30594993 on Build GRCh38
rs775425723 Chr.7: 30594882-30594882 on Build GRCh38
rs919928419 Chr.7: 30594742-30594742 on Build GRCh38
rs919647902 Chr.7: 30594854-30594854 on Build GRCh38
rs886062269 Chr.7: 30594725-30594725 on Build GRCh38
rs538051301 Chr.7: 30594638-30594638 on Build GRCh38
rs944520337 Chr.7: 30594872-30594872 on Build GRCh38
rs62636572 Chr.7: 30594932-30594932 on Build GRCh38
rs568199609 Chr.7: 30594913-30594913 on Build GRCh38
rs756664556 Chr.7: 30594980-30594980 on Build GRCh38
rs201132307 Chr.7: 30594940-30594940 on Build GRCh38
rs569310951 Chr.7: 30594903-30594903 on Build GRCh38
rs986033444 Chr.7: 30594865-30594865 on Build GRCh38
rs564177184 Chr.7: 30594879-30594879 on Build GRCh38
rs891712368 Chr.7: 30594881-30594881 on Build GRCh38
rs746437033 Chr.7: 30594927-30594927 on Build GRCh38
rs1008627610 Chr.7: 30594602-30594602 on Build GRCh38
rs753221402 Chr.7: 30594910-30594910 on Build GRCh38
rs531483802 Chr.7: 30594886-30594886 on Build GRCh38
rs781520666 Chr.7: 30594924-30594924 on Build GRCh38
rs955394532 Chr.7: 30594949-30594949 on Build GRCh38
rs761679763 Chr.7: 30594849-30594849 on Build GRCh38
rs961449034 Chr.7: 30594842-30594842 on Build GRCh38
rs202117737 Chr.7: 30594972-30594972 on Build GRCh38
rs116125961 Chr.7: 30594605-30594605 on Build GRCh38
rs775650014 Chr.7: 30594933-30594933 on Build GRCh38
rs940014942 Chr.7: 30594727-30594727 on Build GRCh38
rs778383373 Chr.7: 30594914-30594914 on Build GRCh38
rs967434049 Chr.7: 30594617-30594617 on Build GRCh38
rs907088979 Chr.7: 30594754-30594754 on Build GRCh38
rs1038082371 Chr.7: 30594732-30594732 on Build GRCh38
rs747409671 Chr.7: 30594916-30594916 on Build GRCh38
rs765982079 Chr.7: 30594904-30594904 on Build GRCh38
rs761660649 Chr.7: 30594895-30594895 on Build GRCh38
rs764698848 Chr.7: 30594964-30594964 on Build GRCh38
rs1032651070 Chr.7: 30594790-30594790 on Build GRCh38
rs1036682839 Chr.7: 30594557-30594557 on Build GRCh38
rs544405016 Chr.7: 30594778-30594778 on Build GRCh38
rs955582963 Chr.7: 30594825-30594825 on Build GRCh38
rs758933506 Chr.7: 30594963-30594963 on Build GRCh38
rs1049399 Chr.7: 30594917-30594917 on Build GRCh38
rs758037738 Chr.7: 30594965-30594965 on Build GRCh38
rs530359858 Chr.7: 30594680-30594680 on Build GRCh38
rs542192277 Chr.7: 30594819-30594819 on Build GRCh38
rs148593022 Chr.7: 30594697-30594697 on Build GRCh38
rs549993976 Chr.7: 30594891-30594891 on Build GRCh38
rs774307716 Chr.7: 30594892-30594892 on Build GRCh38
rs1038333196 Chr.7: 30594741-30594741 on Build GRCh38
rs903029869 Chr.7: 30594877-30594877 on Build GRCh38
rs981616071 Chr.7: 30594832-30594832 on Build GRCh38

View Full Product Details