Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 14146114-14146630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
UniGene
Hs.567255
Gene Chromosome Location
Chr.7: 14145049-14974777 on Build GRCh38
Transcripts
NM_004080.2

SNP Detail

SNP ID Chromosome Location
rs909590655 Chr.7: 14146253-14146253 on Build GRCh38
rs750237712 Chr.7: 14146393-14146393 on Build GRCh38
rs984611174 Chr.7: 14146476-14146476 on Build GRCh38
rs965588666 Chr.7: 14146446-14146446 on Build GRCh38
rs547047245 Chr.7: 14146333-14146333 on Build GRCh38
rs115322449 Chr.7: 14146145-14146145 on Build GRCh38
rs967945537 Chr.7: 14146429-14146429 on Build GRCh38
rs999129075 Chr.7: 14146439-14146439 on Build GRCh38
rs891892955 Chr.7: 14146425-14146425 on Build GRCh38
rs111989759 Chr.7: 14146243-14146243 on Build GRCh38
rs939992921 Chr.7: 14146327-14146327 on Build GRCh38
rs139586687 Chr.7: 14146579-14146579 on Build GRCh38
rs766620939 Chr.7: 14146396-14146396 on Build GRCh38
rs918323309 Chr.7: 14146285-14146285 on Build GRCh38
rs992695009 Chr.7: 14146231-14146231 on Build GRCh38
rs906238505 Chr.7: 14146336-14146336 on Build GRCh38
rs1001839079 Chr.7: 14146339-14146339 on Build GRCh38
rs570464407 Chr.7: 14146458-14146458 on Build GRCh38
rs908365862 Chr.7: 14146323-14146323 on Build GRCh38
rs944000211 Chr.7: 14146277-14146277 on Build GRCh38
rs893557930 Chr.7: 14146392-14146392 on Build GRCh38
rs918226569 Chr.7: 14146519-14146519 on Build GRCh38
rs927902833 Chr.7: 14146575-14146575 on Build GRCh38
rs979868 Chr.7: 14146251-14146251 on Build GRCh38
rs34120194 Chr.7: 14146183-14146184 on Build GRCh38
rs957998155 Chr.7: 14146217-14146217 on Build GRCh38
rs527607700 Chr.7: 14146316-14146316 on Build GRCh38
rs921231722 Chr.7: 14146460-14146460 on Build GRCh38
rs549510121 Chr.7: 14146503-14146503 on Build GRCh38
rs577232212 Chr.7: 14146171-14146171 on Build GRCh38
rs1054694177 Chr.7: 14146357-14146357 on Build GRCh38
rs933762130 Chr.7: 14146533-14146533 on Build GRCh38
rs1048234112 Chr.7: 14146313-14146313 on Build GRCh38
rs1019863713 Chr.7: 14146445-14146445 on Build GRCh38
rs1024530866 Chr.7: 14146234-14146234 on Build GRCh38
rs990351017 Chr.7: 14146220-14146220 on Build GRCh38
rs187127069 Chr.7: 14146224-14146224 on Build GRCh38
rs1021827769 Chr.7: 14146421-14146421 on Build GRCh38
rs569098004 Chr.7: 14146521-14146521 on Build GRCh38
rs952863352 Chr.7: 14146473-14146473 on Build GRCh38
rs1011038227 Chr.7: 14146407-14146407 on Build GRCh38
rs955900643 Chr.7: 14146230-14146230 on Build GRCh38
rs770152706 Chr.7: 14146256-14146256 on Build GRCh38
rs538006809 Chr.7: 14146556-14146556 on Build GRCh38
rs887035405 Chr.7: 14146142-14146142 on Build GRCh38
rs977659499 Chr.7: 14146262-14146262 on Build GRCh38
rs939845197 Chr.7: 14146529-14146529 on Build GRCh38
rs774269122 Chr.7: 14146449-14146449 on Build GRCh38
rs192081390 Chr.7: 14146308-14146308 on Build GRCh38
rs904566203 Chr.7: 14146391-14146391 on Build GRCh38
rs554743875 Chr.7: 14146561-14146561 on Build GRCh38
rs570448009 Chr.7: 14146288-14146288 on Build GRCh38
rs931351846 Chr.7: 14146311-14146311 on Build GRCh38
rs972409019 Chr.7: 14146516-14146516 on Build GRCh38
rs1000275501 Chr.7: 14146207-14146207 on Build GRCh38
rs1016849490 Chr.7: 14146466-14146466 on Build GRCh38
rs777085744 Chr.7: 14146249-14146249 on Build GRCh38
rs986913491 Chr.7: 14146540-14146540 on Build GRCh38
rs769013155 Chr.7: 14146221-14146221 on Build GRCh38
rs375074445 Chr.7: 14146181-14146181 on Build GRCh38
rs150559448 Chr.7: 14146146-14146146 on Build GRCh38
rs376698044 Chr.7: 14146200-14146200 on Build GRCh38
rs943634598 Chr.7: 14146326-14146326 on Build GRCh38
rs759553106 Chr.7: 14146530-14146530 on Build GRCh38
rs747601624 Chr.7: 14146206-14146206 on Build GRCh38
rs539140555 Chr.7: 14146465-14146465 on Build GRCh38
rs904494012 Chr.7: 14146205-14146205 on Build GRCh38
rs866561254 Chr.7: 14146273-14146273 on Build GRCh38
rs1024632697 Chr.7: 14146147-14146147 on Build GRCh38
rs1001933956 Chr.7: 14146173-14146173 on Build GRCh38
rs763259928 Chr.7: 14146303-14146303 on Build GRCh38
rs1046187599 Chr.7: 14146572-14146572 on Build GRCh38
rs908395617 Chr.7: 14146477-14146477 on Build GRCh38

View Full Product Details