Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91267461-91267957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD1
Entrez Gene ID
8321
Gene Name
frizzled class receptor 1
Gene Aliases
-
UniGene
Hs.94234
Gene Chromosome Location
Chr.7: 91264468-91268817 on Build GRCh38
Transcripts
NM_003505.1

SNP Detail

SNP ID Chromosome Location
rs1011076632 Chr.7: 91267563-91267563 on Build GRCh38
rs894056876 Chr.7: 91267542-91267542 on Build GRCh38
rs188223046 Chr.7: 91267486-91267486 on Build GRCh38
rs1039014784 Chr.7: 91267861-91267861 on Build GRCh38
rs550392104 Chr.7: 91267829-91267829 on Build GRCh38
rs1030721151 Chr.7: 91267741-91267741 on Build GRCh38
rs918440988 Chr.7: 91267742-91267743 on Build GRCh38
rs990477392 Chr.7: 91267657-91267657 on Build GRCh38
rs918405279 Chr.7: 91267498-91267498 on Build GRCh38
rs3750145 Chr.7: 91267516-91267516 on Build GRCh38
rs879887652 Chr.7: 91267625-91267625 on Build GRCh38
rs967252522 Chr.7: 91267586-91267586 on Build GRCh38
rs192284995 Chr.7: 91267543-91267543 on Build GRCh38
rs903653463 Chr.7: 91267667-91267667 on Build GRCh38
rs79504063 Chr.7: 91267647-91267647 on Build GRCh38
rs942377923 Chr.7: 91267620-91267620 on Build GRCh38
rs891977090 Chr.7: 91267771-91267771 on Build GRCh38
rs941265687 Chr.7: 91267694-91267694 on Build GRCh38
rs1010364315 Chr.7: 91267779-91267779 on Build GRCh38
rs537195277 Chr.7: 91267645-91267645 on Build GRCh38
rs1017843267 Chr.7: 91267530-91267530 on Build GRCh38
rs567498853 Chr.7: 91267858-91267858 on Build GRCh38
rs571106547 Chr.7: 91267551-91267551 on Build GRCh38
rs1035164854 Chr.7: 91267617-91267617 on Build GRCh38
rs1039337689 Chr.7: 91267631-91267631 on Build GRCh38
rs1051473679 Chr.7: 91267522-91267522 on Build GRCh38
rs890527585 Chr.7: 91267856-91267856 on Build GRCh38
rs971630115 Chr.7: 91267793-91267793 on Build GRCh38
rs1051884116 Chr.7: 91267744-91267744 on Build GRCh38
rs751867283 Chr.7: 91267696-91267696 on Build GRCh38
rs765543797 Chr.7: 91267857-91267857 on Build GRCh38
rs933799510 Chr.7: 91267742-91267742 on Build GRCh38
rs891003302 Chr.7: 91267532-91267532 on Build GRCh38
rs1024983794 Chr.7: 91267781-91267781 on Build GRCh38
rs536474451 Chr.7: 91267904-91267904 on Build GRCh38
rs981904989 Chr.7: 91267613-91267613 on Build GRCh38
rs777135522 Chr.7: 91267529-91267529 on Build GRCh38
rs1021089661 Chr.7: 91267564-91267564 on Build GRCh38
rs972617395 Chr.7: 91267721-91267721 on Build GRCh38
rs980136916 Chr.7: 91267811-91267811 on Build GRCh38

View Full Product Details