Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120988924-120989523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPED1
Entrez Gene ID
79974
Gene Name
cadherin like and PC-esterase domain containing 1
Gene Aliases
C7orf58
UniGene
Hs.189652
Gene Chromosome Location
Chr.7: 120988697-121297444 on Build GRCh38
Transcripts
NM_001105533.1 NM_024913.4 XM_017012649.1

SNP Detail

SNP ID Chromosome Location
rs904397943 Chr.7: 120989271-120989271 on Build GRCh38
rs921530473 Chr.7: 120989467-120989467 on Build GRCh38
rs184538585 Chr.7: 120989060-120989060 on Build GRCh38
rs990596912 Chr.7: 120989491-120989491 on Build GRCh38
rs2525740 Chr.7: 120989427-120989427 on Build GRCh38
rs1021530385 Chr.7: 120989105-120989105 on Build GRCh38
rs534353465 Chr.7: 120989194-120989194 on Build GRCh38
rs2525738 Chr.7: 120989172-120989172 on Build GRCh38
rs902770295 Chr.7: 120989429-120989429 on Build GRCh38
rs949270163 Chr.7: 120989229-120989229 on Build GRCh38
rs1039993231 Chr.7: 120989120-120989120 on Build GRCh38
rs150871188 Chr.7: 120989061-120989061 on Build GRCh38
rs540400068 Chr.7: 120989152-120989153 on Build GRCh38
rs533565642 Chr.7: 120989009-120989009 on Build GRCh38
rs1034300026 Chr.7: 120989349-120989349 on Build GRCh38
rs962931614 Chr.7: 120989453-120989453 on Build GRCh38
rs1049105587 Chr.7: 120988959-120988959 on Build GRCh38
rs189461125 Chr.7: 120989382-120989382 on Build GRCh38
rs763708606 Chr.7: 120989463-120989463 on Build GRCh38
rs577537662 Chr.7: 120989286-120989286 on Build GRCh38
rs2525739 Chr.7: 120989216-120989216 on Build GRCh38
rs997718765 Chr.7: 120989423-120989423 on Build GRCh38
rs546623540 Chr.7: 120989182-120989182 on Build GRCh38
rs144547146 Chr.7: 120989442-120989442 on Build GRCh38
rs55666906 Chr.7: 120989162-120989162 on Build GRCh38
rs990461020 Chr.7: 120989214-120989214 on Build GRCh38
rs941599859 Chr.7: 120989111-120989111 on Build GRCh38
rs959123671 Chr.7: 120989205-120989205 on Build GRCh38
rs368713267 Chr.7: 120989498-120989498 on Build GRCh38
rs762637722 Chr.7: 120989082-120989083 on Build GRCh38
rs560130076 Chr.7: 120989474-120989474 on Build GRCh38
rs972714912 Chr.7: 120989231-120989231 on Build GRCh38
rs759356034 Chr.7: 120989245-120989245 on Build GRCh38
rs748376955 Chr.7: 120988968-120988968 on Build GRCh38
rs980010162 Chr.7: 120989163-120989163 on Build GRCh38
rs1048718862 Chr.7: 120989275-120989275 on Build GRCh38
rs931127874 Chr.7: 120989272-120989272 on Build GRCh38
rs768567827 Chr.7: 120989341-120989341 on Build GRCh38
rs1017517223 Chr.7: 120989449-120989449 on Build GRCh38
rs139672757 Chr.7: 120989094-120989094 on Build GRCh38
rs146633346 Chr.7: 120989456-120989456 on Build GRCh38
rs753364642 Chr.7: 120989473-120989473 on Build GRCh38
rs570134352 Chr.7: 120989375-120989375 on Build GRCh38
rs901889040 Chr.7: 120989417-120989417 on Build GRCh38
rs377528126 Chr.7: 120989370-120989370 on Build GRCh38
rs1017579367 Chr.7: 120989023-120989023 on Build GRCh38
rs369606353 Chr.7: 120989066-120989066 on Build GRCh38
rs142413727 Chr.7: 120989043-120989043 on Build GRCh38
rs943482082 Chr.7: 120989338-120989338 on Build GRCh38
rs1024355031 Chr.7: 120989413-120989413 on Build GRCh38
rs770071525 Chr.7: 120988969-120988969 on Build GRCh38
rs910100027 Chr.7: 120989106-120989106 on Build GRCh38
rs537533250 Chr.7: 120989016-120989016 on Build GRCh38

View Full Product Details