Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11424275-11424824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167A
Entrez Gene ID
83648
Gene Name
family with sequence similarity 167 member A
Gene Aliases
C8orf13, D8S265
UniGene
Hs.124299
Gene Chromosome Location
Chr.8: 11421464-11475904 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Gene Symbol
FAM167A-AS1
Entrez Gene ID
83656
Gene Name
FAM167A antisense RNA 1
Gene Aliases
C8orf12
UniGene
Hs.583896
Gene Chromosome Location
Chr.8: 11368402-11438657 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643471 c.563C>A p.S188Y Chr.8: 11424455-11424455 on Build GRCh38
4749961 c.520C>T p.R174W Chr.8: 11424498-11424498 on Build GRCh38
244371 c.601G>A p.V201M Chr.8: 11424417-11424417 on Build GRCh38
7367129 c.460G>C p.E154Q Chr.8: 11424558-11424558 on Build GRCh38
266501 c.437G>T p.G146V Chr.8: 11424581-11424581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747371658 Chr.8: 11424369-11424369 on Build GRCh38
rs922544676 Chr.8: 11424777-11424777 on Build GRCh38
rs768510757 Chr.8: 11424454-11424454 on Build GRCh38
rs1030721261 Chr.8: 11424343-11424343 on Build GRCh38
rs760183930 Chr.8: 11424626-11424626 on Build GRCh38
rs780501946 Chr.8: 11424558-11424558 on Build GRCh38
rs111729419 Chr.8: 11424684-11424684 on Build GRCh38
rs755737011 Chr.8: 11424399-11424399 on Build GRCh38
rs752012096 Chr.8: 11424488-11424488 on Build GRCh38
rs773785540 Chr.8: 11424323-11424341 on Build GRCh38
rs567362067 Chr.8: 11424642-11424642 on Build GRCh38
rs963199025 Chr.8: 11424320-11424320 on Build GRCh38
rs761572595 Chr.8: 11424394-11424394 on Build GRCh38
rs547076790 Chr.8: 11424336-11424336 on Build GRCh38
rs538682492 Chr.8: 11424719-11424719 on Build GRCh38
rs140423769 Chr.8: 11424545-11424545 on Build GRCh38
rs538000985 Chr.8: 11424698-11424698 on Build GRCh38
rs368459570 Chr.8: 11424417-11424417 on Build GRCh38
rs750839979 Chr.8: 11424494-11424494 on Build GRCh38
rs984126198 Chr.8: 11424448-11424448 on Build GRCh38
rs148850114 Chr.8: 11424712-11424712 on Build GRCh38
rs756046453 Chr.8: 11424654-11424655 on Build GRCh38
rs1011016398 Chr.8: 11424672-11424672 on Build GRCh38
rs202235860 Chr.8: 11424340-11424340 on Build GRCh38
rs764004616 Chr.8: 11424337-11424337 on Build GRCh38
rs940693820 Chr.8: 11424607-11424607 on Build GRCh38
rs778894458 Chr.8: 11424568-11424568 on Build GRCh38
rs759963618 Chr.8: 11424450-11424452 on Build GRCh38
rs980256350 Chr.8: 11424746-11424746 on Build GRCh38
rs761836545 Chr.8: 11424793-11424796 on Build GRCh38
rs762583224 Chr.8: 11424593-11424593 on Build GRCh38
rs764168254 Chr.8: 11424393-11424393 on Build GRCh38
rs564594491 Chr.8: 11424646-11424647 on Build GRCh38
rs1022062195 Chr.8: 11424559-11424559 on Build GRCh38
rs376807405 Chr.8: 11424325-11424325 on Build GRCh38
rs1017007026 Chr.8: 11424763-11424763 on Build GRCh38
rs373564873 Chr.8: 11424339-11424339 on Build GRCh38
rs996008684 Chr.8: 11424321-11424321 on Build GRCh38
rs577482093 Chr.8: 11424713-11424713 on Build GRCh38
rs369090343 Chr.8: 11424497-11424497 on Build GRCh38
rs761747530 Chr.8: 11424464-11424464 on Build GRCh38
rs533627555 Chr.8: 11424634-11424634 on Build GRCh38
rs143743025 Chr.8: 11424421-11424421 on Build GRCh38
rs566681543 Chr.8: 11424395-11424395 on Build GRCh38
rs139424835 Chr.8: 11424455-11424455 on Build GRCh38
rs762338989 Chr.8: 11424533-11424533 on Build GRCh38
rs1041736999 Chr.8: 11424680-11424680 on Build GRCh38
rs755367917 Chr.8: 11424606-11424606 on Build GRCh38
rs774348671 Chr.8: 11424571-11424571 on Build GRCh38
rs370927037 Chr.8: 11424596-11424596 on Build GRCh38
rs561322004 Chr.8: 11424483-11424483 on Build GRCh38
rs537742836 Chr.8: 11424330-11424330 on Build GRCh38
rs757296081 Chr.8: 11424668-11424668 on Build GRCh38
rs565530389 Chr.8: 11424548-11424548 on Build GRCh38
rs146816358 Chr.8: 11424490-11424490 on Build GRCh38
rs763106621 Chr.8: 11424552-11424552 on Build GRCh38
rs150406646 Chr.8: 11424550-11424550 on Build GRCh38
rs779476502 Chr.8: 11424350-11424350 on Build GRCh38
rs17153216 Chr.8: 11424757-11424757 on Build GRCh38
rs200511231 Chr.8: 11424328-11424328 on Build GRCh38
rs752201866 Chr.8: 11424532-11424532 on Build GRCh38
rs748294847 Chr.8: 11424575-11424575 on Build GRCh38
rs1055995813 Chr.8: 11424705-11424705 on Build GRCh38
rs754451449 Chr.8: 11424405-11424405 on Build GRCh38
rs765900981 Chr.8: 11424400-11424400 on Build GRCh38
rs892100302 Chr.8: 11424670-11424670 on Build GRCh38
rs755206316 Chr.8: 11424567-11424567 on Build GRCh38
rs745542182 Chr.8: 11424590-11424590 on Build GRCh38
rs1016067086 Chr.8: 11424318-11424318 on Build GRCh38
rs141292162 Chr.8: 11424418-11424418 on Build GRCh38
rs780996003 Chr.8: 11424628-11424628 on Build GRCh38
rs765541335 Chr.8: 11424609-11424609 on Build GRCh38
rs771780754 Chr.8: 11424465-11424465 on Build GRCh38
rs550216545 Chr.8: 11424531-11424531 on Build GRCh38
rs901019403 Chr.8: 11424514-11424514 on Build GRCh38
rs960504292 Chr.8: 11424768-11424768 on Build GRCh38
rs372968845 Chr.8: 11424505-11424505 on Build GRCh38
rs1025810371 Chr.8: 11424739-11424739 on Build GRCh38
rs1038192816 Chr.8: 11424440-11424440 on Build GRCh38
rs566603440 Chr.8: 11424625-11424625 on Build GRCh38
rs372350472 Chr.8: 11424473-11424473 on Build GRCh38
rs758382682 Chr.8: 11424579-11424579 on Build GRCh38
rs755649418 Chr.8: 11424347-11424347 on Build GRCh38
rs553728707 Chr.8: 11424724-11424724 on Build GRCh38
rs754305260 Chr.8: 11424734-11424734 on Build GRCh38
rs778137618 Chr.8: 11424367-11424367 on Build GRCh38
rs765620953 Chr.8: 11424515-11424522 on Build GRCh38
rs771433749 Chr.8: 11424378-11424381 on Build GRCh38
rs1043834323 Chr.8: 11424676-11424676 on Build GRCh38
rs772010209 Chr.8: 11424508-11424508 on Build GRCh38
rs746092041 Chr.8: 11424429-11424429 on Build GRCh38
rs370672901 Chr.8: 11424314-11424314 on Build GRCh38
rs913743855 Chr.8: 11424802-11424802 on Build GRCh38
rs766283803 Chr.8: 11424668-11424669 on Build GRCh38
rs779654158 Chr.8: 11424687-11424687 on Build GRCh38
rs989648123 Chr.8: 11424381-11424381 on Build GRCh38
rs750355812 Chr.8: 11424686-11424686 on Build GRCh38
rs749519377 Chr.8: 11424498-11424498 on Build GRCh38
rs373127114 Chr.8: 11424660-11424660 on Build GRCh38
rs903470906 Chr.8: 11424683-11424683 on Build GRCh38
rs558608053 Chr.8: 11424766-11424766 on Build GRCh38
rs777251081 Chr.8: 11424652-11424652 on Build GRCh38
rs776331059 Chr.8: 11424523-11424523 on Build GRCh38
rs753301707 Chr.8: 11424403-11424403 on Build GRCh38
rs146979421 Chr.8: 11424591-11424591 on Build GRCh38
rs757255814 Chr.8: 11424507-11424507 on Build GRCh38
rs199968652 Chr.8: 11424392-11424392 on Build GRCh38
rs561393537 Chr.8: 11424779-11424779 on Build GRCh38
rs1001750743 Chr.8: 11424558-11424559 on Build GRCh38
rs770696934 Chr.8: 11424522-11424522 on Build GRCh38
rs761299096 Chr.8: 11424649-11424649 on Build GRCh38
rs201058431 Chr.8: 11424528-11424528 on Build GRCh38
rs370416023 Chr.8: 11424354-11424354 on Build GRCh38
rs754766828 Chr.8: 11424671-11424679 on Build GRCh38
rs1008791752 Chr.8: 11424650-11424650 on Build GRCh38
rs149662282 Chr.8: 11424511-11424511 on Build GRCh38
rs942595478 Chr.8: 11424496-11424496 on Build GRCh38
rs777181659 Chr.8: 11424423-11424424 on Build GRCh38
rs368537961 Chr.8: 11424342-11424342 on Build GRCh38
rs775453862 Chr.8: 11424398-11424398 on Build GRCh38
rs145481836 Chr.8: 11424584-11424584 on Build GRCh38
rs138151717 Chr.8: 11424600-11424600 on Build GRCh38
rs572193573 Chr.8: 11424435-11424435 on Build GRCh38
rs770101595 Chr.8: 11424431-11424431 on Build GRCh38
rs768845937 Chr.8: 11424444-11424444 on Build GRCh38
rs771301144 Chr.8: 11424375-11424375 on Build GRCh38
rs746960113 Chr.8: 11424627-11424627 on Build GRCh38
rs1022321826 Chr.8: 11424351-11424351 on Build GRCh38
rs760272883 Chr.8: 11424479-11424479 on Build GRCh38
rs931797359 Chr.8: 11424633-11424633 on Build GRCh38
rs373612696 Chr.8: 11424341-11424341 on Build GRCh38
rs754218407 Chr.8: 11424602-11424602 on Build GRCh38
rs143521673 Chr.8: 11424635-11424635 on Build GRCh38
rs775785936 Chr.8: 11424516-11424517 on Build GRCh38
rs138628802 Chr.8: 11424565-11424565 on Build GRCh38
rs752970462 Chr.8: 11424657-11424657 on Build GRCh38
rs200337863 Chr.8: 11424595-11424595 on Build GRCh38
rs752846903 Chr.8: 11424612-11424612 on Build GRCh38
rs374639232 Chr.8: 11424409-11424409 on Build GRCh38
rs757724438 Chr.8: 11424489-11424489 on Build GRCh38
rs758850719 Chr.8: 11424355-11424355 on Build GRCh38
rs542744918 Chr.8: 11424776-11424776 on Build GRCh38
rs200487180 Chr.8: 11424546-11424546 on Build GRCh38
rs753694360 Chr.8: 11424670-11424670 on Build GRCh38
rs759102263 Chr.8: 11424484-11424484 on Build GRCh38
rs781164142 Chr.8: 11424673-11424673 on Build GRCh38
rs767037170 Chr.8: 11424651-11424651 on Build GRCh38
rs769766563 Chr.8: 11424388-11424388 on Build GRCh38
rs760000055 Chr.8: 11424654-11424654 on Build GRCh38
rs368405922 Chr.8: 11424492-11424492 on Build GRCh38
rs773953753 Chr.8: 11424648-11424648 on Build GRCh38
rs775280270 Chr.8: 11424647-11424647 on Build GRCh38
rs749935355 Chr.8: 11424585-11424585 on Build GRCh38
rs760508594 Chr.8: 11424516-11424516 on Build GRCh38
rs892200041 Chr.8: 11424542-11424542 on Build GRCh38
rs746097006 Chr.8: 11424387-11424387 on Build GRCh38
rs766056567 Chr.8: 11424481-11424481 on Build GRCh38
rs763754784 Chr.8: 11424343-11424344 on Build GRCh38
rs938768205 Chr.8: 11424301-11424301 on Build GRCh38
rs751599012 Chr.8: 11424661-11424661 on Build GRCh38
rs951596015 Chr.8: 11424749-11424749 on Build GRCh38
rs940760420 Chr.8: 11424434-11424434 on Build GRCh38
rs778299082 Chr.8: 11424408-11424408 on Build GRCh38
rs988423024 Chr.8: 11424791-11424794 on Build GRCh38
rs1022034019 Chr.8: 11424690-11424690 on Build GRCh38
rs761556428 Chr.8: 11424580-11424580 on Build GRCh38
rs761098784 Chr.8: 11424378-11424378 on Build GRCh38
rs896059633 Chr.8: 11424711-11424711 on Build GRCh38

View Full Product Details