Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140739652-140740123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6332508 c.2029G>A p.D677N Chr.7: 140739910-140739910 on Build GRCh38
4385977 c.2040G>T p.K680N Chr.7: 140739899-140739899 on Build GRCh38
6957667 c.2021T>A p.L674Q Chr.7: 140739918-140739918 on Build GRCh38
5872002 c.2056C>T p.P686S Chr.7: 140739883-140739883 on Build GRCh38
6475278 c.2034C>T p.L678L Chr.7: 140739905-140739905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757016514 Chr.7: 140739933-140739933 on Build GRCh38
rs748847875 Chr.7: 140739796-140739796 on Build GRCh38
rs755852332 Chr.7: 140739777-140739777 on Build GRCh38
rs960546121 Chr.7: 140740022-140740022 on Build GRCh38
rs371976102 Chr.7: 140739809-140739809 on Build GRCh38
rs376554544 Chr.7: 140739766-140739766 on Build GRCh38
rs55715359 Chr.7: 140739864-140739864 on Build GRCh38
rs748680512 Chr.7: 140739987-140739987 on Build GRCh38
rs971451154 Chr.7: 140739955-140739955 on Build GRCh38
rs1034603754 Chr.7: 140739956-140739956 on Build GRCh38
rs558990907 Chr.7: 140739966-140739966 on Build GRCh38
rs201793951 Chr.7: 140739929-140739929 on Build GRCh38
rs765734118 Chr.7: 140739956-140739957 on Build GRCh38
rs367812694 Chr.7: 140739772-140739772 on Build GRCh38
rs575667092 Chr.7: 140739677-140739677 on Build GRCh38
rs757029878 Chr.7: 140739765-140739765 on Build GRCh38
rs751336763 Chr.7: 140739764-140739764 on Build GRCh38
rs886766660 Chr.7: 140739750-140739750 on Build GRCh38
rs750297886 Chr.7: 140739957-140739957 on Build GRCh38
rs916046242 Chr.7: 140740050-140740050 on Build GRCh38
rs916100507 Chr.7: 140739704-140739704 on Build GRCh38
rs938849898 Chr.7: 140739673-140739673 on Build GRCh38
rs754567621 Chr.7: 140739980-140739980 on Build GRCh38
rs984889160 Chr.7: 140739958-140739958 on Build GRCh38
rs541015003 Chr.7: 140740011-140740011 on Build GRCh38
rs761066454 Chr.7: 140740069-140740069 on Build GRCh38
rs759664527 Chr.7: 140739920-140739920 on Build GRCh38
rs775021122 Chr.7: 140739804-140739804 on Build GRCh38
rs999291320 Chr.7: 140740002-140740002 on Build GRCh38
rs772330392 Chr.7: 140739808-140739808 on Build GRCh38
rs776058053 Chr.7: 140739955-140739956 on Build GRCh38
rs397507485 Chr.7: 140739927-140739927 on Build GRCh38
rs779422978 Chr.7: 140739791-140739791 on Build GRCh38
rs991759529 Chr.7: 140740049-140740049 on Build GRCh38
rs886041257 Chr.7: 140739838-140739838 on Build GRCh38
rs576801042 Chr.7: 140739713-140739713 on Build GRCh38
rs886041260 Chr.7: 140739910-140739910 on Build GRCh38
rs1028549781 Chr.7: 140739874-140739874 on Build GRCh38
rs776664982 Chr.7: 140739895-140739895 on Build GRCh38
rs1035708024 Chr.7: 140740015-140740015 on Build GRCh38
rs745567884 Chr.7: 140739697-140739697 on Build GRCh38
rs972521995 Chr.7: 140740034-140740034 on Build GRCh38
rs397507486 Chr.7: 140739813-140739813 on Build GRCh38
rs767199788 Chr.7: 140739954-140739954 on Build GRCh38
rs375174370 Chr.7: 140739857-140739857 on Build GRCh38
rs1040947183 Chr.7: 140739759-140739759 on Build GRCh38
rs748758036 Chr.7: 140739967-140739967 on Build GRCh38
rs1054299757 Chr.7: 140739782-140739782 on Build GRCh38
rs779495487 Chr.7: 140739984-140739984 on Build GRCh38
rs775808846 Chr.7: 140739959-140739959 on Build GRCh38
rs939706621 Chr.7: 140739751-140739751 on Build GRCh38
rs371857758 Chr.7: 140739805-140739805 on Build GRCh38
rs893650301 Chr.7: 140739898-140739898 on Build GRCh38
rs901111900 Chr.7: 140739788-140739788 on Build GRCh38
rs770572009 Chr.7: 140739950-140739951 on Build GRCh38
rs769263015 Chr.7: 140739801-140739801 on Build GRCh38
rs1048178832 Chr.7: 140739733-140739733 on Build GRCh38
rs745516557 Chr.7: 140739776-140739776 on Build GRCh38
rs967334726 Chr.7: 140740001-140740001 on Build GRCh38
rs201618982 Chr.7: 140739992-140739992 on Build GRCh38
rs766692331 Chr.7: 140739861-140739861 on Build GRCh38
rs759069424 Chr.7: 140739957-140739957 on Build GRCh38

View Full Product Details