Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 43085359-43085922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNTA
Entrez Gene ID
2339
Gene Name
farnesyltransferase, CAAX box, alpha
Gene Aliases
FPTA, PGGT1A, PTAR2
UniGene
Hs.370312
Gene Chromosome Location
Chr.8: 43056299-43085788 on Build GRCh38
Transcripts
NM_002027.2 NR_033698.1

SNP Detail

SNP ID Chromosome Location
rs11554712 Chr.8: 43085670-43085670 on Build GRCh38
rs1042618846 Chr.8: 43085794-43085794 on Build GRCh38
rs1021185961 Chr.8: 43085598-43085598 on Build GRCh38
rs923234474 Chr.8: 43085447-43085447 on Build GRCh38
rs755916862 Chr.8: 43085477-43085477 on Build GRCh38
rs923398171 Chr.8: 43085740-43085740 on Build GRCh38
rs190378282 Chr.8: 43085760-43085760 on Build GRCh38
rs868106502 Chr.8: 43085833-43085833 on Build GRCh38
rs1034793840 Chr.8: 43085461-43085461 on Build GRCh38
rs1028751532 Chr.8: 43085538-43085538 on Build GRCh38
rs979420068 Chr.8: 43085520-43085520 on Build GRCh38
rs528788946 Chr.8: 43085423-43085423 on Build GRCh38
rs762888952 Chr.8: 43085703-43085703 on Build GRCh38
rs573144758 Chr.8: 43085781-43085781 on Build GRCh38
rs983886524 Chr.8: 43085770-43085770 on Build GRCh38
rs550647765 Chr.8: 43085590-43085590 on Build GRCh38
rs941189820 Chr.8: 43085491-43085491 on Build GRCh38
rs954129355 Chr.8: 43085542-43085542 on Build GRCh38
rs926411773 Chr.8: 43085844-43085844 on Build GRCh38
rs183382931 Chr.8: 43085839-43085839 on Build GRCh38
rs968411035 Chr.8: 43085516-43085516 on Build GRCh38
rs1020863459 Chr.8: 43085514-43085514 on Build GRCh38
rs976102787 Chr.8: 43085728-43085728 on Build GRCh38
rs896247279 Chr.8: 43085851-43085855 on Build GRCh38
rs570601316 Chr.8: 43085640-43085640 on Build GRCh38
rs557155893 Chr.8: 43085853-43085853 on Build GRCh38
rs937703364 Chr.8: 43085709-43085709 on Build GRCh38
rs968239409 Chr.8: 43085386-43085386 on Build GRCh38
rs142364879 Chr.8: 43085397-43085397 on Build GRCh38
rs976155112 Chr.8: 43085434-43085434 on Build GRCh38
rs377746719 Chr.8: 43085607-43085607 on Build GRCh38
rs896143982 Chr.8: 43085545-43085545 on Build GRCh38
rs766597324 Chr.8: 43085586-43085586 on Build GRCh38
rs1001922251 Chr.8: 43085424-43085424 on Build GRCh38
rs904906550 Chr.8: 43085419-43085419 on Build GRCh38
rs3196850 Chr.8: 43085725-43085725 on Build GRCh38
rs887048403 Chr.8: 43085486-43085486 on Build GRCh38
rs1009355848 Chr.8: 43085751-43085751 on Build GRCh38
rs777558857 Chr.8: 43085722-43085722 on Build GRCh38
rs536859657 Chr.8: 43085644-43085644 on Build GRCh38
rs912677829 Chr.8: 43085567-43085567 on Build GRCh38
rs1048695089 Chr.8: 43085681-43085681 on Build GRCh38
rs1056222149 Chr.8: 43085848-43085848 on Build GRCh38
rs140553256 Chr.8: 43085865-43085865 on Build GRCh38
rs1046973358 Chr.8: 43085463-43085463 on Build GRCh38
rs1001205913 Chr.8: 43085840-43085840 on Build GRCh38
rs1049206870 Chr.8: 43085400-43085400 on Build GRCh38
rs898774561 Chr.8: 43085388-43085388 on Build GRCh38
rs548466550 Chr.8: 43085425-43085425 on Build GRCh38
rs954012766 Chr.8: 43085888-43085888 on Build GRCh38
rs536699177 Chr.8: 43085582-43085582 on Build GRCh38
rs931564420 Chr.8: 43085650-43085650 on Build GRCh38
rs905047862 Chr.8: 43085515-43085515 on Build GRCh38
rs753776897 Chr.8: 43085727-43085727 on Build GRCh38
rs146781943 Chr.8: 43085506-43085506 on Build GRCh38
rs187422693 Chr.8: 43085704-43085704 on Build GRCh38
rs986863678 Chr.8: 43085543-43085543 on Build GRCh38
rs937862585 Chr.8: 43085845-43085845 on Build GRCh38
rs909228301 Chr.8: 43085802-43085802 on Build GRCh38

View Full Product Details