Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102834543-102835107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
UniGene
Hs.459106
Gene Chromosome Location
Chr.8: 102826302-102864200 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Gene Symbol
LOC107986873
Entrez Gene ID
107986873
Gene Name
forkhead box protein L2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 102810302-102863067 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4834335 c.589C>T p.H197Y Chr.8: 102834743-102834743 on Build GRCh38
6472214 c.614C>T p.S205F Chr.8: 102834718-102834718 on Build GRCh38
213870 c.610G>C p.E204Q Chr.8: 102834722-102834722 on Build GRCh38
3732137 c.632C>T p.A211V Chr.8: 102834700-102834700 on Build GRCh38
6055777 c.605G>A p.C202Y Chr.8: 102834727-102834727 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs549486654 Chr.8: 102834650-102834650 on Build GRCh38
rs771366422 Chr.8: 102834933-102834933 on Build GRCh38
rs753914794 Chr.8: 102834871-102834871 on Build GRCh38
rs779121314 Chr.8: 102834968-102834968 on Build GRCh38
rs900184312 Chr.8: 102835023-102835023 on Build GRCh38
rs779638813 Chr.8: 102834713-102834713 on Build GRCh38
rs187777894 Chr.8: 102834818-102834818 on Build GRCh38
rs531309471 Chr.8: 102834650-102834654 on Build GRCh38
rs545432558 Chr.8: 102834602-102834603 on Build GRCh38
rs766897529 Chr.8: 102834741-102834741 on Build GRCh38
rs930108208 Chr.8: 102834587-102834587 on Build GRCh38
rs771858692 Chr.8: 102834688-102834688 on Build GRCh38
rs750222537 Chr.8: 102834672-102834672 on Build GRCh38
rs373718982 Chr.8: 102834653-102834657 on Build GRCh38
rs1055020977 Chr.8: 102834983-102834983 on Build GRCh38
rs188435821 Chr.8: 102834595-102834595 on Build GRCh38
rs145786248 Chr.8: 102834718-102834718 on Build GRCh38
rs890296410 Chr.8: 102834717-102834717 on Build GRCh38
rs777986584 Chr.8: 102834636-102834636 on Build GRCh38
rs117057089 Chr.8: 102834903-102834903 on Build GRCh38
rs763664481 Chr.8: 102834707-102834707 on Build GRCh38
rs760259243 Chr.8: 102834700-102834700 on Build GRCh38
rs79031142 Chr.8: 102835019-102835019 on Build GRCh38
rs756694151 Chr.8: 102834727-102834727 on Build GRCh38
rs1047479069 Chr.8: 102834602-102834603 on Build GRCh38
rs749355793 Chr.8: 102834797-102834797 on Build GRCh38
rs550963830 Chr.8: 102834954-102834954 on Build GRCh38
rs757965469 Chr.8: 102834696-102834696 on Build GRCh38
rs368044057 Chr.8: 102834776-102834776 on Build GRCh38
rs563695442 Chr.8: 102834626-102834630 on Build GRCh38
rs941783685 Chr.8: 102834605-102834605 on Build GRCh38
rs1020114313 Chr.8: 102834735-102834735 on Build GRCh38
rs774704018 Chr.8: 102834629-102834631 on Build GRCh38
rs957539285 Chr.8: 102834732-102834732 on Build GRCh38
rs993045796 Chr.8: 102834840-102834840 on Build GRCh38
rs913303591 Chr.8: 102834847-102834847 on Build GRCh38
rs759269331 Chr.8: 102834685-102834685 on Build GRCh38
rs781563194 Chr.8: 102834760-102834760 on Build GRCh38
rs888294131 Chr.8: 102834995-102834995 on Build GRCh38
rs369549318 Chr.8: 102834758-102834758 on Build GRCh38
rs780158511 Chr.8: 102834870-102834870 on Build GRCh38
rs773576663 Chr.8: 102834651-102834654 on Build GRCh38
rs574206393 Chr.8: 102835082-102835082 on Build GRCh38
rs978857559 Chr.8: 102834801-102834801 on Build GRCh38
rs67964073 Chr.8: 102834596-102834597 on Build GRCh38
rs749806611 Chr.8: 102834928-102834928 on Build GRCh38
rs375020522 Chr.8: 102835026-102835026 on Build GRCh38
rs539842781 Chr.8: 102835061-102835061 on Build GRCh38
rs1037795316 Chr.8: 102834635-102834635 on Build GRCh38
rs760976847 Chr.8: 102834657-102834658 on Build GRCh38
rs996107007 Chr.8: 102834650-102834653 on Build GRCh38
rs755116551 Chr.8: 102834872-102834872 on Build GRCh38
rs569270256 Chr.8: 102835009-102835009 on Build GRCh38
rs749304461 Chr.8: 102834638-102834638 on Build GRCh38
rs145454106 Chr.8: 102834699-102834699 on Build GRCh38
rs917435329 Chr.8: 102834569-102834569 on Build GRCh38
rs931596718 Chr.8: 102835080-102835081 on Build GRCh38
rs539689983 Chr.8: 102834922-102834922 on Build GRCh38
rs3214633 Chr.8: 102834597-102834598 on Build GRCh38
rs1034778136 Chr.8: 102834804-102834804 on Build GRCh38
rs71303443 Chr.8: 102834595-102834596 on Build GRCh38
rs778039122 Chr.8: 102834780-102834780 on Build GRCh38
rs762163920 Chr.8: 102834644-102834648 on Build GRCh38
rs766837545 Chr.8: 102834663-102834664 on Build GRCh38
rs755304216 Chr.8: 102834759-102834759 on Build GRCh38
rs770008496 Chr.8: 102834620-102834620 on Build GRCh38
rs772142600 Chr.8: 102835062-102835062 on Build GRCh38
rs149083461 Chr.8: 102834779-102834779 on Build GRCh38
rs927238785 Chr.8: 102834937-102834937 on Build GRCh38

View Full Product Details