Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182386878-182387464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUL
Entrez Gene ID
2752
Gene Name
glutamate-ammonia ligase
Gene Aliases
GLNS, GS, PIG43, PIG59
UniGene
Hs.132016
Gene Chromosome Location
Chr.1: 182378093-182392206 on Build GRCh38
Transcripts
NM_001033044.3 NM_001033056.3 NM_002065.6 XM_006711278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336917 c.186C>T p.F62F Chr.1: 182387273-182387273 on Build GRCh38
3479905 c.274G>A p.D92N Chr.1: 182387185-182387185 on Build GRCh38
6447914 c.237C>T p.L79L Chr.1: 182387222-182387222 on Build GRCh38
4576660 c.168G>A p.E56E Chr.1: 182387291-182387291 on Build GRCh38
123382 c.319A>G p.R107G Chr.1: 182387140-182387140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906773155 Chr.1: 182386964-182386964 on Build GRCh38
rs539793139 Chr.1: 182387082-182387082 on Build GRCh38
rs955248197 Chr.1: 182387439-182387439 on Build GRCh38
rs141126924 Chr.1: 182387273-182387273 on Build GRCh38
rs969146542 Chr.1: 182387267-182387267 on Build GRCh38
rs17462824 Chr.1: 182387112-182387112 on Build GRCh38
rs781327746 Chr.1: 182387120-182387121 on Build GRCh38
rs771767939 Chr.1: 182387064-182387064 on Build GRCh38
rs370606415 Chr.1: 182387237-182387237 on Build GRCh38
rs748116431 Chr.1: 182387306-182387307 on Build GRCh38
rs897582610 Chr.1: 182387436-182387436 on Build GRCh38
rs149058083 Chr.1: 182387152-182387152 on Build GRCh38
rs770631160 Chr.1: 182387123-182387123 on Build GRCh38
rs776266756 Chr.1: 182387124-182387124 on Build GRCh38
rs774042474 Chr.1: 182387341-182387341 on Build GRCh38
rs543259650 Chr.1: 182386992-182386992 on Build GRCh38
rs768583558 Chr.1: 182387336-182387336 on Build GRCh38
rs771775154 Chr.1: 182387114-182387114 on Build GRCh38
rs199769578 Chr.1: 182387255-182387264 on Build GRCh38
rs1058111 Chr.1: 182387264-182387264 on Build GRCh38
rs910198515 Chr.1: 182387359-182387359 on Build GRCh38
rs367550174 Chr.1: 182387105-182387105 on Build GRCh38
rs765202450 Chr.1: 182387142-182387142 on Build GRCh38
rs745333737 Chr.1: 182387227-182387227 on Build GRCh38
rs932917304 Chr.1: 182387327-182387328 on Build GRCh38
rs199746338 Chr.1: 182387191-182387191 on Build GRCh38
rs376306093 Chr.1: 182387147-182387147 on Build GRCh38
rs775388317 Chr.1: 182387145-182387145 on Build GRCh38
rs1035569086 Chr.1: 182387121-182387122 on Build GRCh38
rs888834174 Chr.1: 182387081-182387081 on Build GRCh38
rs1055178827 Chr.1: 182387009-182387009 on Build GRCh38
rs910516060 Chr.1: 182387141-182387141 on Build GRCh38
rs371858972 Chr.1: 182387108-182387108 on Build GRCh38
rs924724847 Chr.1: 182387282-182387282 on Build GRCh38
rs748476678 Chr.1: 182387322-182387322 on Build GRCh38
rs774185084 Chr.1: 182387250-182387250 on Build GRCh38
rs896899743 Chr.1: 182387390-182387390 on Build GRCh38
rs756642285 Chr.1: 182387190-182387190 on Build GRCh38
rs756231421 Chr.1: 182387291-182387291 on Build GRCh38
rs548878766 Chr.1: 182387005-182387005 on Build GRCh38
rs993955517 Chr.1: 182387393-182387393 on Build GRCh38
rs751683068 Chr.1: 182387084-182387084 on Build GRCh38
rs748431581 Chr.1: 182387078-182387078 on Build GRCh38
rs116513736 Chr.1: 182386948-182386948 on Build GRCh38
rs367852420 Chr.1: 182387304-182387304 on Build GRCh38
rs111524794 Chr.1: 182387135-182387135 on Build GRCh38
rs371238132 Chr.1: 182387115-182387115 on Build GRCh38
rs201312968 Chr.1: 182387330-182387330 on Build GRCh38
rs766410836 Chr.1: 182387298-182387298 on Build GRCh38
rs1038081970 Chr.1: 182387389-182387389 on Build GRCh38
rs1003825523 Chr.1: 182386974-182386974 on Build GRCh38
rs554405325 Chr.1: 182387050-182387050 on Build GRCh38
rs377614330 Chr.1: 182387154-182387154 on Build GRCh38
rs201791327 Chr.1: 182387202-182387202 on Build GRCh38
rs543481170 Chr.1: 182387125-182387125 on Build GRCh38
rs779331105 Chr.1: 182387209-182387209 on Build GRCh38
rs750433606 Chr.1: 182387185-182387185 on Build GRCh38
rs1029530925 Chr.1: 182387109-182387109 on Build GRCh38
rs78681284 Chr.1: 182387331-182387331 on Build GRCh38
rs113454052 Chr.1: 182387295-182387295 on Build GRCh38
rs750521238 Chr.1: 182387286-182387286 on Build GRCh38
rs777819987 Chr.1: 182387083-182387083 on Build GRCh38
rs943434572 Chr.1: 182386901-182386901 on Build GRCh38
rs564037834 Chr.1: 182387354-182387354 on Build GRCh38
rs755338601 Chr.1: 182387204-182387204 on Build GRCh38
rs924785234 Chr.1: 182386938-182386938 on Build GRCh38
rs757499402 Chr.1: 182387155-182387155 on Build GRCh38
rs574532372 Chr.1: 182387219-182387219 on Build GRCh38
rs749244267 Chr.1: 182387335-182387335 on Build GRCh38
rs763866364 Chr.1: 182387246-182387246 on Build GRCh38
rs1042603010 Chr.1: 182386959-182386959 on Build GRCh38
rs796496885 Chr.1: 182387103-182387103 on Build GRCh38
rs1005223612 Chr.1: 182387383-182387383 on Build GRCh38
rs12735664 Chr.1: 182386944-182386944 on Build GRCh38
rs780945383 Chr.1: 182387326-182387326 on Build GRCh38
rs757335121 Chr.1: 182387085-182387085 on Build GRCh38
rs776287282 Chr.1: 182387222-182387222 on Build GRCh38
rs746641298 Chr.1: 182387116-182387116 on Build GRCh38
rs779156551 Chr.1: 182387318-182387318 on Build GRCh38
rs762920855 Chr.1: 182387244-182387244 on Build GRCh38
rs113485734 Chr.1: 182387130-182387130 on Build GRCh38
rs757002617 Chr.1: 182387323-182387323 on Build GRCh38
rs979316823 Chr.1: 182386919-182386919 on Build GRCh38
rs151138433 Chr.1: 182387221-182387221 on Build GRCh38
rs566236605 Chr.1: 182387074-182387074 on Build GRCh38
rs6666230 Chr.1: 182387189-182387189 on Build GRCh38
rs560340787 Chr.1: 182387248-182387248 on Build GRCh38
rs188126943 Chr.1: 182386916-182386916 on Build GRCh38
rs1029887677 Chr.1: 182387437-182387437 on Build GRCh38
rs373022393 Chr.1: 182387110-182387110 on Build GRCh38
rs755497075 Chr.1: 182387309-182387309 on Build GRCh38
rs779616294 Chr.1: 182387332-182387332 on Build GRCh38
rs767643850 Chr.1: 182387159-182387159 on Build GRCh38
rs779945271 Chr.1: 182387057-182387057 on Build GRCh38

View Full Product Details