Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196675693-196676273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFH
Entrez Gene ID
3075
Gene Name
complement factor H
Gene Aliases
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS
UniGene
Hs.363396
Gene Chromosome Location
Chr.1: 196651878-196747504 on Build GRCh38
Transcripts
NM_000186.3 NM_001014975.2 XM_017001108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6060124 c.381T>C p.R127R Chr.1: 196676019-196676019 on Build GRCh38
209290 c.397G>A p.G133R Chr.1: 196676035-196676035 on Build GRCh38
3771573 c.427G>A p.V143I Chr.1: 196676065-196676065 on Build GRCh38
4489989 c.355C>T p.Q119* Chr.1: 196675993-196675993 on Build GRCh38
1730011 c.418A>G p.I140V Chr.1: 196676056-196676056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903230936 Chr.1: 196675969-196675969 on Build GRCh38
rs746517444 Chr.1: 196676139-196676139 on Build GRCh38
rs773693442 Chr.1: 196675986-196675986 on Build GRCh38
rs996989995 Chr.1: 196676231-196676231 on Build GRCh38
rs1045691296 Chr.1: 196676197-196676197 on Build GRCh38
rs373339058 Chr.1: 196676035-196676035 on Build GRCh38
rs1054666899 Chr.1: 196676010-196676010 on Build GRCh38
rs373961921 Chr.1: 196676103-196676103 on Build GRCh38
rs528144204 Chr.1: 196675810-196675810 on Build GRCh38
rs34842495 Chr.1: 196676123-196676123 on Build GRCh38
rs747852489 Chr.1: 196675964-196675964 on Build GRCh38
rs997127130 Chr.1: 196676040-196676040 on Build GRCh38
rs377211562 Chr.1: 196676122-196676123 on Build GRCh38
rs56237321 Chr.1: 196675815-196675816 on Build GRCh38
rs949277414 Chr.1: 196675748-196675748 on Build GRCh38
rs768247864 Chr.1: 196675954-196675954 on Build GRCh38
rs993203818 Chr.1: 196676215-196676215 on Build GRCh38
rs920457743 Chr.1: 196676173-196676173 on Build GRCh38
rs749261592 Chr.1: 196676106-196676106 on Build GRCh38
rs1055624130 Chr.1: 196675929-196675929 on Build GRCh38
rs1008442813 Chr.1: 196675861-196675861 on Build GRCh38
rs750289844 Chr.1: 196676042-196676042 on Build GRCh38
rs147002633 Chr.1: 196676026-196676026 on Build GRCh38
rs112026050 Chr.1: 196675883-196675883 on Build GRCh38
rs573149699 Chr.1: 196676182-196676182 on Build GRCh38
rs534021367 Chr.1: 196676185-196676185 on Build GRCh38
rs35108970 Chr.1: 196676172-196676173 on Build GRCh38
rs369221218 Chr.1: 196675943-196675943 on Build GRCh38
rs759999515 Chr.1: 196676008-196676008 on Build GRCh38
rs546762242 Chr.1: 196675831-196675831 on Build GRCh38
rs1050013120 Chr.1: 196676244-196676244 on Build GRCh38
rs182386164 Chr.1: 196675729-196675729 on Build GRCh38
rs550967449 Chr.1: 196675928-196675928 on Build GRCh38
rs748855994 Chr.1: 196675952-196675952 on Build GRCh38
rs568051993 Chr.1: 196675879-196675879 on Build GRCh38
rs776146154 Chr.1: 196676091-196676093 on Build GRCh38
rs972216992 Chr.1: 196676134-196676134 on Build GRCh38
rs1018459574 Chr.1: 196675876-196675876 on Build GRCh38
rs557886216 Chr.1: 196676181-196676181 on Build GRCh38
rs1032896735 Chr.1: 196676125-196676125 on Build GRCh38
rs750038396 Chr.1: 196675904-196675904 on Build GRCh38
rs1028694346 Chr.1: 196675777-196675777 on Build GRCh38
rs376321145 Chr.1: 196675992-196675992 on Build GRCh38
rs768234992 Chr.1: 196676087-196676088 on Build GRCh38
rs573913360 Chr.1: 196676116-196676116 on Build GRCh38
rs962194001 Chr.1: 196675864-196675864 on Build GRCh38
rs773951949 Chr.1: 196675955-196675955 on Build GRCh38
rs771558168 Chr.1: 196675975-196675975 on Build GRCh38
rs375271367 Chr.1: 196676104-196676107 on Build GRCh38
rs961860133 Chr.1: 196676129-196676129 on Build GRCh38
rs758647278 Chr.1: 196676029-196676029 on Build GRCh38
rs573930117 Chr.1: 196676227-196676227 on Build GRCh38
rs1033685753 Chr.1: 196675717-196675717 on Build GRCh38
rs189220686 Chr.1: 196676192-196676192 on Build GRCh38
rs780125819 Chr.1: 196675951-196675951 on Build GRCh38
rs777094657 Chr.1: 196676000-196676000 on Build GRCh38
rs915935339 Chr.1: 196675937-196675937 on Build GRCh38
rs987645941 Chr.1: 196675796-196675796 on Build GRCh38
rs55965675 Chr.1: 196676181-196676181 on Build GRCh38
rs140618003 Chr.1: 196675921-196675921 on Build GRCh38
rs564280628 Chr.1: 196675795-196675795 on Build GRCh38
rs368897930 Chr.1: 196676112-196676112 on Build GRCh38
rs754907102 Chr.1: 196676107-196676107 on Build GRCh38
rs941896496 Chr.1: 196676005-196676005 on Build GRCh38
rs371462079 Chr.1: 196675995-196675995 on Build GRCh38
rs930595650 Chr.1: 196675927-196675927 on Build GRCh38
rs956501696 Chr.1: 196676172-196676172 on Build GRCh38
rs1054529081 Chr.1: 196676224-196676224 on Build GRCh38
rs994208309 Chr.1: 196675815-196675815 on Build GRCh38
rs755841584 Chr.1: 196676079-196676079 on Build GRCh38
rs953008464 Chr.1: 196675791-196675791 on Build GRCh38
rs765265569 Chr.1: 196676115-196676116 on Build GRCh38
rs113472243 Chr.1: 196676190-196676190 on Build GRCh38
rs1044895126 Chr.1: 196675784-196675784 on Build GRCh38
rs919043884 Chr.1: 196676235-196676235 on Build GRCh38
rs980622798 Chr.1: 196675970-196675970 on Build GRCh38
rs934793293 Chr.1: 196676223-196676223 on Build GRCh38
rs758497308 Chr.1: 196675942-196675942 on Build GRCh38
rs747608778 Chr.1: 196676116-196676116 on Build GRCh38
rs192715337 Chr.1: 196675718-196675718 on Build GRCh38
rs980385812 Chr.1: 196676210-196676210 on Build GRCh38
rs121913058 Chr.1: 196676018-196676018 on Build GRCh38
rs1028557096 Chr.1: 196676086-196676086 on Build GRCh38
rs780179183 Chr.1: 196676096-196676096 on Build GRCh38
rs953176788 Chr.1: 196676090-196676090 on Build GRCh38
rs754116374 Chr.1: 196675812-196675813 on Build GRCh38
rs369487905 Chr.1: 196675811-196675812 on Build GRCh38
rs773280333 Chr.1: 196675847-196675853 on Build GRCh38
rs372157046 Chr.1: 196676173-196676173 on Build GRCh38
rs957567917 Chr.1: 196675720-196675720 on Build GRCh38
rs113004102 Chr.1: 196675987-196675987 on Build GRCh38
rs373036907 Chr.1: 196675965-196675965 on Build GRCh38
rs971883192 Chr.1: 196675882-196675882 on Build GRCh38
rs771972035 Chr.1: 196675813-196675814 on Build GRCh38

View Full Product Details