Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 30892614-30893196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC5
Entrez Gene ID
100506627
Gene Name
doublecortin domain containing 5
Gene Aliases
-
UniGene
Hs.530438
Gene Chromosome Location
Chr.11: 30863603-30992686 on Build GRCh38
Transcripts
NM_020869.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998050 c.1072C>T p.R358* Chr.11: 30892889-30892889 on Build GRCh38
1353427 c.1123T>C p.W375R Chr.11: 30892838-30892838 on Build GRCh38
6537291 c.990G>C p.M330I Chr.11: 30892971-30892971 on Build GRCh38
7099311 c.985G>C p.E329Q Chr.11: 30892976-30892976 on Build GRCh38
7415613 c.982C>A p.Q328K Chr.11: 30892979-30892979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769773585 Chr.11: 30892976-30892976 on Build GRCh38
rs550850566 Chr.11: 30893021-30893021 on Build GRCh38
rs183741564 Chr.11: 30892775-30892775 on Build GRCh38
rs973371259 Chr.11: 30892669-30892669 on Build GRCh38
rs767877253 Chr.11: 30892801-30892801 on Build GRCh38
rs767136587 Chr.11: 30892961-30892961 on Build GRCh38
rs749122881 Chr.11: 30892939-30892939 on Build GRCh38
rs767519935 Chr.11: 30893047-30893047 on Build GRCh38
rs1039979964 Chr.11: 30892834-30892834 on Build GRCh38
rs116099904 Chr.11: 30892920-30892920 on Build GRCh38
rs761166563 Chr.11: 30893011-30893011 on Build GRCh38
rs193258092 Chr.11: 30892676-30892676 on Build GRCh38
rs754093190 Chr.11: 30893028-30893028 on Build GRCh38
rs188215210 Chr.11: 30892649-30892649 on Build GRCh38
rs781321680 Chr.11: 30892952-30892952 on Build GRCh38
rs776033574 Chr.11: 30893000-30893000 on Build GRCh38
rs770437666 Chr.11: 30892981-30892981 on Build GRCh38
rs764427413 Chr.11: 30892926-30892926 on Build GRCh38
rs372974839 Chr.11: 30893143-30893143 on Build GRCh38
rs969242485 Chr.11: 30893135-30893135 on Build GRCh38
rs887520809 Chr.11: 30892811-30892811 on Build GRCh38
rs112299430 Chr.11: 30892761-30892761 on Build GRCh38
rs770544810 Chr.11: 30892896-30892896 on Build GRCh38
rs762504427 Chr.11: 30892895-30892895 on Build GRCh38
rs368503716 Chr.11: 30892949-30892949 on Build GRCh38
rs992113172 Chr.11: 30893051-30893051 on Build GRCh38
rs759817535 Chr.11: 30892656-30892656 on Build GRCh38
rs928059820 Chr.11: 30892772-30892772 on Build GRCh38
rs1036016331 Chr.11: 30892863-30892863 on Build GRCh38
rs560230357 Chr.11: 30892765-30892765 on Build GRCh38
rs995054733 Chr.11: 30893067-30893067 on Build GRCh38
rs772499561 Chr.11: 30892831-30892831 on Build GRCh38
rs776912393 Chr.11: 30892644-30892644 on Build GRCh38
rs773050294 Chr.11: 30892889-30892889 on Build GRCh38
rs771547180 Chr.11: 30892998-30892998 on Build GRCh38
rs372269095 Chr.11: 30892980-30892980 on Build GRCh38
rs761692309 Chr.11: 30892797-30892797 on Build GRCh38
rs748079326 Chr.11: 30892963-30892963 on Build GRCh38
rs139655298 Chr.11: 30893132-30893132 on Build GRCh38
rs760106610 Chr.11: 30892914-30892914 on Build GRCh38
rs916042557 Chr.11: 30892789-30892789 on Build GRCh38
rs569583697 Chr.11: 30893100-30893100 on Build GRCh38
rs761573034 Chr.11: 30892784-30892784 on Build GRCh38
rs368137781 Chr.11: 30892835-30892835 on Build GRCh38
rs778853457 Chr.11: 30892850-30892850 on Build GRCh38
rs759005244 Chr.11: 30892996-30892996 on Build GRCh38
rs762566885 Chr.11: 30892790-30892790 on Build GRCh38
rs779064286 Chr.11: 30892945-30892945 on Build GRCh38
rs753007745 Chr.11: 30892808-30892808 on Build GRCh38
rs777709051 Chr.11: 30892977-30892977 on Build GRCh38
rs761075007 Chr.11: 30892921-30892921 on Build GRCh38
rs569432827 Chr.11: 30893058-30893058 on Build GRCh38
rs908988951 Chr.11: 30893144-30893144 on Build GRCh38
rs757353786 Chr.11: 30892937-30892937 on Build GRCh38
rs545497859 Chr.11: 30892888-30892888 on Build GRCh38
rs940392726 Chr.11: 30893168-30893168 on Build GRCh38
rs963534332 Chr.11: 30892655-30892655 on Build GRCh38
rs754975870 Chr.11: 30892951-30892951 on Build GRCh38
rs764935024 Chr.11: 30892786-30892786 on Build GRCh38
rs746972463 Chr.11: 30892858-30892858 on Build GRCh38
rs752170900 Chr.11: 30892795-30892795 on Build GRCh38
rs373949850 Chr.11: 30892809-30892809 on Build GRCh38
rs746907666 Chr.11: 30892773-30892773 on Build GRCh38
rs1021704138 Chr.11: 30892909-30892909 on Build GRCh38
rs916155226 Chr.11: 30893090-30893090 on Build GRCh38
rs374142291 Chr.11: 30892936-30892936 on Build GRCh38
rs1023953504 Chr.11: 30892726-30892726 on Build GRCh38
rs747913513 Chr.11: 30892887-30892887 on Build GRCh38
rs773940703 Chr.11: 30892992-30892992 on Build GRCh38
rs777571557 Chr.11: 30892886-30892886 on Build GRCh38
rs768420393 Chr.11: 30892884-30892884 on Build GRCh38
rs755369617 Chr.11: 30892800-30892800 on Build GRCh38
rs376149695 Chr.11: 30892783-30892783 on Build GRCh38
rs931650592 Chr.11: 30892891-30892891 on Build GRCh38
rs946862435 Chr.11: 30892817-30892817 on Build GRCh38
rs187923172 Chr.11: 30892791-30892791 on Build GRCh38
rs940360400 Chr.11: 30892847-30892847 on Build GRCh38
rs762119017 Chr.11: 30893042-30893042 on Build GRCh38
rs773674256 Chr.11: 30892907-30892907 on Build GRCh38
rs768424839 Chr.11: 30892729-30892729 on Build GRCh38
rs757415492 Chr.11: 30892844-30892844 on Build GRCh38

View Full Product Details