Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31432518-31432982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
UniGene
Hs.187269
Gene Chromosome Location
Chr.11: 31369829-31432835 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1 NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

Gene Symbol
IMMP1L
Entrez Gene ID
196294
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
IMMP1, IMP1, IMP1-LIKE
UniGene
Hs.502223
Gene Chromosome Location
Chr.11: 31432399-31509644 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1 NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604478 c.461delT p.L154fs*>13 Chr.11: 31432540-31432540 on Build GRCh38
5630250 c.476A>T p.N159I Chr.11: 31432525-31432525 on Build GRCh38
4494938 c.442C>T p.L148L Chr.11: 31432559-31432559 on Build GRCh38
6685585 c.433-4A>G p.? Chr.11: 31432572-31432572 on Build GRCh38
4032344 c.437G>T p.W146L Chr.11: 31432564-31432564 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759966128 Chr.11: 31432576-31432576 on Build GRCh38
rs998715611 Chr.11: 31432774-31432774 on Build GRCh38
rs560637187 Chr.11: 31432887-31432887 on Build GRCh38
rs752027802 Chr.11: 31432569-31432569 on Build GRCh38
rs750903334 Chr.11: 31432780-31432780 on Build GRCh38
rs756584770 Chr.11: 31432927-31432927 on Build GRCh38
rs905979453 Chr.11: 31432957-31432957 on Build GRCh38
rs767900311 Chr.11: 31432616-31432616 on Build GRCh38
rs766645264 Chr.11: 31432899-31432899 on Build GRCh38
rs768439612 Chr.11: 31432606-31432616 on Build GRCh38
rs899625654 Chr.11: 31432710-31432710 on Build GRCh38
rs926157268 Chr.11: 31432693-31432693 on Build GRCh38
rs987126611 Chr.11: 31432658-31432658 on Build GRCh38
rs566046408 Chr.11: 31432879-31432879 on Build GRCh38
rs772546006 Chr.11: 31432547-31432547 on Build GRCh38
rs911683675 Chr.11: 31432675-31432675 on Build GRCh38
rs766728264 Chr.11: 31432554-31432554 on Build GRCh38
rs951387184 Chr.11: 31432778-31432779 on Build GRCh38
rs372412278 Chr.11: 31432537-31432537 on Build GRCh38
rs893707456 Chr.11: 31432661-31432661 on Build GRCh38
rs768018814 Chr.11: 31432680-31432680 on Build GRCh38
rs950086247 Chr.11: 31432855-31432855 on Build GRCh38
rs936146564 Chr.11: 31432741-31432741 on Build GRCh38
rs533363287 Chr.11: 31432689-31432689 on Build GRCh38
rs1029741509 Chr.11: 31432775-31432775 on Build GRCh38
rs551432389 Chr.11: 31432816-31432816 on Build GRCh38
rs562727683 Chr.11: 31432617-31432617 on Build GRCh38
rs112431824 Chr.11: 31432538-31432538 on Build GRCh38
rs375632237 Chr.11: 31432653-31432653 on Build GRCh38
rs1016920711 Chr.11: 31432870-31432870 on Build GRCh38
rs892006507 Chr.11: 31432821-31432821 on Build GRCh38
rs372250112 Chr.11: 31432626-31432626 on Build GRCh38
rs375832595 Chr.11: 31432549-31432549 on Build GRCh38
rs752677500 Chr.11: 31432847-31432849 on Build GRCh38

View Full Product Details