Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61301984-61302440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
UniGene
Hs.290758
Gene Chromosome Location
Chr.11: 61299447-61333212 on Build GRCh38
Transcripts
NM_001923.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439245 c.3138C>T p.S1046S Chr.11: 61302334-61302334 on Build GRCh38
6461927 c.3182T>C p.V1061A Chr.11: 61302290-61302290 on Build GRCh38
122530 c.3201G>C p.K1067N Chr.11: 61302271-61302271 on Build GRCh38
4776290 c.3201G>T p.K1067N Chr.11: 61302271-61302271 on Build GRCh38
6641226 c.3142T>C p.Y1048H Chr.11: 61302330-61302330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369420599 Chr.11: 61302409-61302409 on Build GRCh38
rs750382896 Chr.11: 61302241-61302241 on Build GRCh38
rs28720351 Chr.11: 61302262-61302262 on Build GRCh38
rs760883222 Chr.11: 61302350-61302350 on Build GRCh38
rs780649504 Chr.11: 61302284-61302284 on Build GRCh38
rs949499509 Chr.11: 61302042-61302042 on Build GRCh38
rs766161677 Chr.11: 61302385-61302385 on Build GRCh38
rs552948022 Chr.11: 61302078-61302078 on Build GRCh38
rs150781675 Chr.11: 61302215-61302215 on Build GRCh38
rs142156411 Chr.11: 61302157-61302157 on Build GRCh38
rs186672621 Chr.11: 61302405-61302405 on Build GRCh38
rs919801104 Chr.11: 61302030-61302030 on Build GRCh38
rs752312432 Chr.11: 61302392-61302392 on Build GRCh38
rs750481253 Chr.11: 61302376-61302376 on Build GRCh38
rs747712060 Chr.11: 61302300-61302300 on Build GRCh38
rs970073422 Chr.11: 61302086-61302086 on Build GRCh38
rs972675860 Chr.11: 61302005-61302005 on Build GRCh38
rs112738410 Chr.11: 61302256-61302256 on Build GRCh38
rs982170442 Chr.11: 61302410-61302410 on Build GRCh38
rs529609793 Chr.11: 61302387-61302387 on Build GRCh38
rs777290980 Chr.11: 61302394-61302394 on Build GRCh38
rs756089166 Chr.11: 61302376-61302381 on Build GRCh38
rs768938953 Chr.11: 61302366-61302366 on Build GRCh38
rs552252536 Chr.11: 61302043-61302043 on Build GRCh38
rs1023186704 Chr.11: 61302084-61302084 on Build GRCh38
rs749684692 Chr.11: 61302100-61302100 on Build GRCh38
rs184167072 Chr.11: 61302039-61302039 on Build GRCh38
rs1016183899 Chr.11: 61302048-61302048 on Build GRCh38
rs779439822 Chr.11: 61302319-61302319 on Build GRCh38
rs560588802 Chr.11: 61302349-61302349 on Build GRCh38
rs949349290 Chr.11: 61302400-61302400 on Build GRCh38
rs962871030 Chr.11: 61302053-61302053 on Build GRCh38
rs762051498 Chr.11: 61302374-61302374 on Build GRCh38
rs995675675 Chr.11: 61302404-61302404 on Build GRCh38
rs138003606 Chr.11: 61302340-61302340 on Build GRCh38
rs988879065 Chr.11: 61302162-61302162 on Build GRCh38
rs1046463609 Chr.11: 61302061-61302061 on Build GRCh38
rs573104064 Chr.11: 61302143-61302143 on Build GRCh38
rs750454569 Chr.11: 61302167-61302167 on Build GRCh38
rs774488814 Chr.11: 61302183-61302183 on Build GRCh38
rs754714183 Chr.11: 61302388-61302388 on Build GRCh38
rs764880050 Chr.11: 61302370-61302372 on Build GRCh38
rs923999325 Chr.11: 61302416-61302416 on Build GRCh38
rs935531970 Chr.11: 61302421-61302421 on Build GRCh38
rs889449696 Chr.11: 61302198-61302198 on Build GRCh38
rs772412053 Chr.11: 61302346-61302346 on Build GRCh38
rs769285778 Chr.11: 61302312-61302312 on Build GRCh38
rs779921300 Chr.11: 61302255-61302255 on Build GRCh38
rs372159527 Chr.11: 61302235-61302235 on Build GRCh38
rs902693679 Chr.11: 61302062-61302062 on Build GRCh38
rs758316619 Chr.11: 61302248-61302248 on Build GRCh38
rs200814352 Chr.11: 61302214-61302214 on Build GRCh38
rs981420460 Chr.11: 61302110-61302110 on Build GRCh38
rs575538984 Chr.11: 61302184-61302184 on Build GRCh38
rs776704384 Chr.11: 61302370-61302370 on Build GRCh38
rs763013579 Chr.11: 61302384-61302384 on Build GRCh38
rs916561839 Chr.11: 61302041-61302041 on Build GRCh38
rs182128993 Chr.11: 61302326-61302326 on Build GRCh38
rs780960836 Chr.11: 61302390-61302390 on Build GRCh38
rs28720352 Chr.11: 61302040-61302040 on Build GRCh38
rs928746148 Chr.11: 61302116-61302116 on Build GRCh38
rs115359696 Chr.11: 61302199-61302199 on Build GRCh38
rs146926940 Chr.11: 61302268-61302268 on Build GRCh38
rs561941161 Chr.11: 61302181-61302181 on Build GRCh38

View Full Product Details