Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108693246-108693775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX10
Entrez Gene ID
1662
Gene Name
DEAD-box helicase 10
Gene Aliases
HRH-J8
UniGene
Hs.591931
Gene Chromosome Location
Chr.11: 108665025-108940930 on Build GRCh38
Transcripts
NM_004398.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242179 c.1160T>C p.V387A Chr.11: 108693537-108693537 on Build GRCh38
6641515 c.1145C>T p.P382L Chr.11: 108693522-108693522 on Build GRCh38
6988743 c.1177C>T p.P393S Chr.11: 108693554-108693554 on Build GRCh38
6497796 c.1182G>T p.E394D Chr.11: 108693559-108693559 on Build GRCh38
1351107 c.1152G>A p.V384V Chr.11: 108693529-108693529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896665269 Chr.11: 108693708-108693708 on Build GRCh38
rs560501559 Chr.11: 108693433-108693433 on Build GRCh38
rs576259947 Chr.11: 108693327-108693327 on Build GRCh38
rs1050314771 Chr.11: 108693270-108693270 on Build GRCh38
rs776311119 Chr.11: 108693603-108693603 on Build GRCh38
rs910367750 Chr.11: 108693271-108693271 on Build GRCh38
rs540999302 Chr.11: 108693409-108693409 on Build GRCh38
rs763224346 Chr.11: 108693637-108693637 on Build GRCh38
rs186583559 Chr.11: 108693351-108693351 on Build GRCh38
rs966752451 Chr.11: 108693473-108693473 on Build GRCh38
rs113758301 Chr.11: 108693426-108693426 on Build GRCh38
rs748924341 Chr.11: 108693550-108693550 on Build GRCh38
rs745782561 Chr.11: 108693488-108693488 on Build GRCh38
rs760521052 Chr.11: 108693501-108693501 on Build GRCh38
rs182860416 Chr.11: 108693333-108693333 on Build GRCh38
rs774168176 Chr.11: 108693696-108693696 on Build GRCh38
rs141530263 Chr.11: 108693527-108693527 on Build GRCh38
rs373284598 Chr.11: 108693508-108693508 on Build GRCh38
rs919034862 Chr.11: 108693537-108693537 on Build GRCh38
rs1016671424 Chr.11: 108693318-108693318 on Build GRCh38
rs766986030 Chr.11: 108693626-108693626 on Build GRCh38
rs896134811 Chr.11: 108693692-108693692 on Build GRCh38
rs1014199663 Chr.11: 108693739-108693739 on Build GRCh38
rs115882119 Chr.11: 108693482-108693482 on Build GRCh38
rs372562723 Chr.11: 108693612-108693612 on Build GRCh38
rs370402551 Chr.11: 108693634-108693634 on Build GRCh38
rs761774918 Chr.11: 108693505-108693505 on Build GRCh38
rs111518330 Chr.11: 108693504-108693504 on Build GRCh38
rs539519228 Chr.11: 108693321-108693324 on Build GRCh38
rs777533142 Chr.11: 108693547-108693547 on Build GRCh38
rs1020965451 Chr.11: 108693444-108693444 on Build GRCh38
rs369219748 Chr.11: 108693490-108693490 on Build GRCh38
rs768488109 Chr.11: 108693598-108693598 on Build GRCh38
rs755567260 Chr.11: 108693631-108693631 on Build GRCh38
rs756892882 Chr.11: 108693552-108693552 on Build GRCh38
rs376427484 Chr.11: 108693512-108693512 on Build GRCh38
rs549958175 Chr.11: 108693663-108693663 on Build GRCh38
rs755457943 Chr.11: 108693536-108693536 on Build GRCh38
rs748915747 Chr.11: 108693336-108693336 on Build GRCh38
rs373881594 Chr.11: 108693458-108693458 on Build GRCh38
rs773281180 Chr.11: 108693621-108693621 on Build GRCh38
rs758907230 Chr.11: 108693522-108693522 on Build GRCh38
rs1040505599 Chr.11: 108693284-108693284 on Build GRCh38
rs554392120 Chr.11: 108693311-108693311 on Build GRCh38
rs536310508 Chr.11: 108693276-108693276 on Build GRCh38
rs767002502 Chr.11: 108693526-108693526 on Build GRCh38
rs376254764 Chr.11: 108693477-108693477 on Build GRCh38
rs887406901 Chr.11: 108693665-108693665 on Build GRCh38
rs775475549 Chr.11: 108693570-108693570 on Build GRCh38
rs746776546 Chr.11: 108693595-108693595 on Build GRCh38
rs561769821 Chr.11: 108693384-108693384 on Build GRCh38
rs199952549 Chr.11: 108693517-108693517 on Build GRCh38
rs752149447 Chr.11: 108693627-108693627 on Build GRCh38
rs567621638 Chr.11: 108693364-108693365 on Build GRCh38
rs1055760960 Chr.11: 108693677-108693677 on Build GRCh38
rs78029525 Chr.11: 108693580-108693580 on Build GRCh38
rs769620305 Chr.11: 108693475-108693475 on Build GRCh38
rs979950326 Chr.11: 108693434-108693434 on Build GRCh38
rs896609018 Chr.11: 108693417-108693417 on Build GRCh38
rs897510757 Chr.11: 108693302-108693302 on Build GRCh38
rs571536952 Chr.11: 108693676-108693676 on Build GRCh38
rs765381428 Chr.11: 108693617-108693617 on Build GRCh38
rs76765504 Chr.11: 108693352-108693352 on Build GRCh38
rs1032300038 Chr.11: 108693478-108693478 on Build GRCh38
rs778008421 Chr.11: 108693622-108693623 on Build GRCh38
rs1004393728 Chr.11: 108693277-108693277 on Build GRCh38
rs933907886 Chr.11: 108693507-108693507 on Build GRCh38
rs370270676 Chr.11: 108693562-108693562 on Build GRCh38
rs762054512 Chr.11: 108693611-108693611 on Build GRCh38
rs775248138 Chr.11: 108693499-108693499 on Build GRCh38
rs778424578 Chr.11: 108693559-108693559 on Build GRCh38
rs377212478 Chr.11: 108693624-108693624 on Build GRCh38

View Full Product Details