Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10015704-10016295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC12B
Entrez Gene ID
387837
Gene Name
C-type lectin domain family 12 member B
Gene Aliases
UNQ5782
UniGene
Hs.127937
Gene Chromosome Location
Chr.12: 10006138-10018800 on Build GRCh38
Transcripts
NM_001129998.2 NM_001319241.1 NM_001319242.1 NM_205852.3 NR_135049.1 XM_011520658.2 XM_011520663.2 XM_017019295.1 NR_120484.1

Gene Symbol
LOC102724020
Entrez Gene ID
102724020
Gene Name
uncharacterized LOC102724020
Gene Aliases
-
UniGene
Hs.737731
Gene Chromosome Location
Chr.12: 10015352-10030606 on Build GRCh38
Transcripts
NM_001129998.2 NM_001319241.1 NM_001319242.1 NM_205852.3 NR_135049.1 XM_011520658.2 XM_011520663.2 XM_017019295.1 NR_120484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630091 c.660C>T p.G220G Chr.12: 10015707-10015707 on Build GRCh38
4439406 c.678C>T p.S226S Chr.12: 10015725-10015725 on Build GRCh38
4439404 c.677C>T p.S226F Chr.12: 10015724-10015724 on Build GRCh38
6630092 c.660C>T p.G220G Chr.12: 10015707-10015707 on Build GRCh38
1322056 c.663_679del17 p.P223fs*3 Chr.12: 10015710-10015726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147703765 Chr.12: 10015731-10015731 on Build GRCh38
rs918549636 Chr.12: 10016217-10016217 on Build GRCh38
rs563573983 Chr.12: 10015788-10015788 on Build GRCh38
rs142428871 Chr.12: 10015736-10015736 on Build GRCh38
rs1045655568 Chr.12: 10015986-10015986 on Build GRCh38
rs1030299279 Chr.12: 10016048-10016048 on Build GRCh38
rs566843273 Chr.12: 10016011-10016011 on Build GRCh38
rs968458521 Chr.12: 10015846-10015846 on Build GRCh38
rs117936789 Chr.12: 10015960-10015960 on Build GRCh38
rs1037033610 Chr.12: 10015868-10015868 on Build GRCh38
rs190493547 Chr.12: 10016140-10016140 on Build GRCh38
rs924362718 Chr.12: 10016118-10016118 on Build GRCh38
rs535449119 Chr.12: 10016038-10016038 on Build GRCh38
rs953273731 Chr.12: 10015728-10015728 on Build GRCh38
rs977113921 Chr.12: 10015869-10015869 on Build GRCh38
rs1007361934 Chr.12: 10015958-10015958 on Build GRCh38
rs949695155 Chr.12: 10015857-10015859 on Build GRCh38
rs494312 Chr.12: 10016112-10016112 on Build GRCh38
rs992904882 Chr.12: 10016151-10016151 on Build GRCh38
rs186030326 Chr.12: 10016004-10016004 on Build GRCh38
rs1020583867 Chr.12: 10015991-10015991 on Build GRCh38
rs772832018 Chr.12: 10015727-10015727 on Build GRCh38
rs779602955 Chr.12: 10016085-10016085 on Build GRCh38
rs959136577 Chr.12: 10016068-10016068 on Build GRCh38
rs370229418 Chr.12: 10015732-10015732 on Build GRCh38
rs773227544 Chr.12: 10016255-10016255 on Build GRCh38
rs764274926 Chr.12: 10015754-10015754 on Build GRCh38
rs183383572 Chr.12: 10016193-10016193 on Build GRCh38
rs531439346 Chr.12: 10015743-10015743 on Build GRCh38
rs1048467394 Chr.12: 10015948-10015948 on Build GRCh38
rs10845019 Chr.12: 10015964-10015964 on Build GRCh38
rs938333507 Chr.12: 10015917-10015917 on Build GRCh38
rs1007453976 Chr.12: 10016058-10016058 on Build GRCh38
rs1036428432 Chr.12: 10016026-10016026 on Build GRCh38
rs757506410 Chr.12: 10015787-10015787 on Build GRCh38
rs924311814 Chr.12: 10015913-10015913 on Build GRCh38
rs887588910 Chr.12: 10015952-10015953 on Build GRCh38
rs118186444 Chr.12: 10015994-10015994 on Build GRCh38
rs990987371 Chr.12: 10016074-10016074 on Build GRCh38
rs977166053 Chr.12: 10016113-10016113 on Build GRCh38
rs561520318 Chr.12: 10015981-10015981 on Build GRCh38
rs918164327 Chr.12: 10015848-10015848 on Build GRCh38
rs760365286 Chr.12: 10015730-10015730 on Build GRCh38
rs759327362 Chr.12: 10015733-10015733 on Build GRCh38
rs537959085 Chr.12: 10016142-10016142 on Build GRCh38
rs533664439 Chr.12: 10015983-10015983 on Build GRCh38
rs762917656 Chr.12: 10015747-10015747 on Build GRCh38
rs1006338643 Chr.12: 10016002-10016002 on Build GRCh38
rs900144832 Chr.12: 10015910-10015910 on Build GRCh38
rs997510561 Chr.12: 10016041-10016041 on Build GRCh38
rs547241038 Chr.12: 10016149-10016149 on Build GRCh38
rs953421413 Chr.12: 10016056-10016056 on Build GRCh38
rs751720828 Chr.12: 10015769-10015769 on Build GRCh38
rs767867368 Chr.12: 10015790-10015790 on Build GRCh38
rs897198406 Chr.12: 10015909-10015909 on Build GRCh38
rs377180681 Chr.12: 10016034-10016034 on Build GRCh38

View Full Product Details