Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64194332-64194892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf66
Entrez Gene ID
144577
Gene Name
chromosome 12 open reading frame 66
Gene Aliases
-
UniGene
Hs.444671
Gene Chromosome Location
Chr.12: 64186312-64222296 on Build GRCh38
Transcripts
NM_001300940.1 NM_001300941.1 NM_152440.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5822966 c.677G>C p.W226S Chr.12: 64194503-64194503 on Build GRCh38
7080405 c.718C>T p.L240L Chr.12: 64194462-64194462 on Build GRCh38
5909212 c.522-8C>T p.? Chr.12: 64194666-64194666 on Build GRCh38
7120098 c.691G>C p.E231Q Chr.12: 64194489-64194489 on Build GRCh38
5841703 c.800C>T p.A267V Chr.12: 64194380-64194380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7959774 Chr.12: 64194848-64194848 on Build GRCh38
rs578180187 Chr.12: 64194669-64194669 on Build GRCh38
rs768565313 Chr.12: 64194671-64194671 on Build GRCh38
rs774708461 Chr.12: 64194371-64194371 on Build GRCh38
rs789721 Chr.12: 64194785-64194785 on Build GRCh38
rs202008538 Chr.12: 64194430-64194430 on Build GRCh38
rs915121779 Chr.12: 64194803-64194803 on Build GRCh38
rs531382321 Chr.12: 64194819-64194819 on Build GRCh38
rs1006163203 Chr.12: 64194868-64194868 on Build GRCh38
rs760097293 Chr.12: 64194615-64194615 on Build GRCh38
rs115571670 Chr.12: 64194764-64194764 on Build GRCh38
rs779034288 Chr.12: 64194631-64194631 on Build GRCh38
rs759204574 Chr.12: 64194435-64194435 on Build GRCh38
rs565220715 Chr.12: 64194866-64194866 on Build GRCh38
rs780374090 Chr.12: 64194526-64194526 on Build GRCh38
rs201307069 Chr.12: 64194354-64194354 on Build GRCh38
rs747048695 Chr.12: 64194536-64194536 on Build GRCh38
rs762325973 Chr.12: 64194570-64194570 on Build GRCh38
rs949023324 Chr.12: 64194774-64194774 on Build GRCh38
rs780137539 Chr.12: 64194665-64194665 on Build GRCh38
rs773754996 Chr.12: 64194692-64194692 on Build GRCh38
rs763412536 Chr.12: 64194603-64194603 on Build GRCh38
rs746091775 Chr.12: 64194383-64194383 on Build GRCh38
rs1023609901 Chr.12: 64194548-64194548 on Build GRCh38
rs778990508 Chr.12: 64194513-64194513 on Build GRCh38
rs777059739 Chr.12: 64194554-64194554 on Build GRCh38
rs757817610 Chr.12: 64194680-64194680 on Build GRCh38
rs764488113 Chr.12: 64194401-64194401 on Build GRCh38
rs775464266 Chr.12: 64194390-64194390 on Build GRCh38
rs772132769 Chr.12: 64194389-64194389 on Build GRCh38
rs946641186 Chr.12: 64194812-64194812 on Build GRCh38
rs750221764 Chr.12: 64194641-64194641 on Build GRCh38
rs755276688 Chr.12: 64194447-64194447 on Build GRCh38
rs140615462 Chr.12: 64194623-64194623 on Build GRCh38
rs558474340 Chr.12: 64194644-64194644 on Build GRCh38
rs752113585 Chr.12: 64194475-64194475 on Build GRCh38
rs1013091403 Chr.12: 64194541-64194541 on Build GRCh38
rs923639649 Chr.12: 64194855-64194855 on Build GRCh38
rs747281376 Chr.12: 64194548-64194548 on Build GRCh38
rs939115523 Chr.12: 64194400-64194400 on Build GRCh38
rs535524580 Chr.12: 64194512-64194512 on Build GRCh38
rs762164607 Chr.12: 64194422-64194422 on Build GRCh38
rs977877520 Chr.12: 64194733-64194733 on Build GRCh38
rs1015576784 Chr.12: 64194683-64194683 on Build GRCh38
rs770905074 Chr.12: 64194359-64194359 on Build GRCh38
rs770251237 Chr.12: 64194687-64194687 on Build GRCh38
rs752449322 Chr.12: 64194606-64194606 on Build GRCh38
rs568868863 Chr.12: 64194539-64194539 on Build GRCh38
rs751158955 Chr.12: 64194433-64194433 on Build GRCh38
rs7959371 Chr.12: 64194741-64194741 on Build GRCh38
rs907375660 Chr.12: 64194833-64194833 on Build GRCh38
rs540076332 Chr.12: 64194758-64194758 on Build GRCh38
rs772154676 Chr.12: 64194524-64194524 on Build GRCh38
rs540855665 Chr.12: 64194738-64194738 on Build GRCh38
rs753892112 Chr.12: 64194505-64194505 on Build GRCh38
rs763858895 Chr.12: 64194499-64194499 on Build GRCh38
rs770099991 Chr.12: 64194598-64194598 on Build GRCh38
rs961434069 Chr.12: 64194736-64194736 on Build GRCh38
rs748609929 Chr.12: 64194682-64194682 on Build GRCh38
rs544741033 Chr.12: 64194867-64194867 on Build GRCh38
rs566655147 Chr.12: 64194480-64194480 on Build GRCh38
rs185163970 Chr.12: 64194523-64194523 on Build GRCh38
rs753428827 Chr.12: 64194640-64194640 on Build GRCh38
rs777182409 Chr.12: 64194415-64194415 on Build GRCh38
rs938999601 Chr.12: 64194862-64194862 on Build GRCh38
rs755696160 Chr.12: 64194479-64194479 on Build GRCh38
rs144407399 Chr.12: 64194722-64194722 on Build GRCh38
rs767052371 Chr.12: 64194604-64194604 on Build GRCh38
rs760849526 Chr.12: 64194396-64194396 on Build GRCh38
rs767009173 Chr.12: 64194448-64194448 on Build GRCh38
rs757282208 Chr.12: 64194624-64194624 on Build GRCh38

View Full Product Details