Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102959230-102959814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL1
Entrez Gene ID
429
Gene Name
achaete-scute family bHLH transcription factor 1
Gene Aliases
ASH1, HASH1, MASH1, bHLHa46
UniGene
Hs.703025
Gene Chromosome Location
Chr.12: 102957674-102960516 on Build GRCh38
Transcripts
NM_004316.3

SNP Detail

SNP ID Chromosome Location
rs548773203 Chr.12: 102959344-102959344 on Build GRCh38
rs944457002 Chr.12: 102959394-102959394 on Build GRCh38
rs757127489 Chr.12: 102959550-102959550 on Build GRCh38
rs796751165 Chr.12: 102959621-102959621 on Build GRCh38
rs893031985 Chr.12: 102959746-102959746 on Build GRCh38
rs1391682 Chr.12: 102959747-102959747 on Build GRCh38
rs778727430 Chr.12: 102959349-102959349 on Build GRCh38
rs148067996 Chr.12: 102959509-102959509 on Build GRCh38
rs534402144 Chr.12: 102959438-102959438 on Build GRCh38
rs753747091 Chr.12: 102959265-102959265 on Build GRCh38
rs562200721 Chr.12: 102959609-102959609 on Build GRCh38
rs777890821 Chr.12: 102959455-102959456 on Build GRCh38
rs995766540 Chr.12: 102959421-102959421 on Build GRCh38
rs398076918 Chr.12: 102959439-102959439 on Build GRCh38
rs192574985 Chr.12: 102959730-102959730 on Build GRCh38
rs1040506992 Chr.12: 102959402-102959402 on Build GRCh38
rs66942154 Chr.12: 102959431-102959431 on Build GRCh38
rs759674014 Chr.12: 102959285-102959285 on Build GRCh38
rs777399679 Chr.12: 102959576-102959576 on Build GRCh38
rs991082628 Chr.12: 102959327-102959327 on Build GRCh38
rs541702840 Chr.12: 102959528-102959528 on Build GRCh38
rs940966021 Chr.12: 102959288-102959288 on Build GRCh38
rs758248657 Chr.12: 102959560-102959560 on Build GRCh38
rs921648512 Chr.12: 102959744-102959744 on Build GRCh38
rs576482822 Chr.12: 102959590-102959590 on Build GRCh38
rs76185146 Chr.12: 102959526-102959526 on Build GRCh38
rs983621689 Chr.12: 102959499-102959499 on Build GRCh38
rs78780516 Chr.12: 102959431-102959431 on Build GRCh38
rs1045526204 Chr.12: 102959699-102959699 on Build GRCh38
rs183912670 Chr.12: 102959761-102959761 on Build GRCh38
rs574386152 Chr.12: 102959471-102959471 on Build GRCh38
rs2291854 Chr.12: 102959760-102959760 on Build GRCh38
rs554073847 Chr.12: 102959453-102959453 on Build GRCh38
rs948460039 Chr.12: 102959672-102959672 on Build GRCh38
rs1035073778 Chr.12: 102959530-102959530 on Build GRCh38
rs900192941 Chr.12: 102959404-102959404 on Build GRCh38
rs968318334 Chr.12: 102959596-102959596 on Build GRCh38
rs386765865 Chr.12: 102959440-102959458 on Build GRCh38
rs886771555 Chr.12: 102959273-102959273 on Build GRCh38
rs940600334 Chr.12: 102959787-102959787 on Build GRCh38
rs1014746747 Chr.12: 102959474-102959474 on Build GRCh38
rs990081329 Chr.12: 102959438-102959439 on Build GRCh38
rs1030376150 Chr.12: 102959772-102959772 on Build GRCh38
rs1017546756 Chr.12: 102959757-102959757 on Build GRCh38
rs1022591034 Chr.12: 102959583-102959583 on Build GRCh38
rs577107256 Chr.12: 102959454-102959454 on Build GRCh38
rs915460814 Chr.12: 102959339-102959339 on Build GRCh38
rs143596223 Chr.12: 102959347-102959347 on Build GRCh38
rs944843330 Chr.12: 102959677-102959677 on Build GRCh38
rs540405976 Chr.12: 102959740-102959740 on Build GRCh38
rs1012684482 Chr.12: 102959338-102959338 on Build GRCh38
rs975866988 Chr.12: 102959713-102959713 on Build GRCh38
rs887724868 Chr.12: 102959786-102959786 on Build GRCh38
rs544068699 Chr.12: 102959368-102959368 on Build GRCh38
rs752838645 Chr.12: 102959366-102959366 on Build GRCh38
rs907071051 Chr.12: 102959376-102959376 on Build GRCh38
rs201782228 Chr.12: 102959430-102959430 on Build GRCh38
rs756197515 Chr.12: 102959451-102959455 on Build GRCh38
rs746670132 Chr.12: 102959733-102959733 on Build GRCh38
rs114008066 Chr.12: 102959305-102959305 on Build GRCh38
rs976222597 Chr.12: 102959468-102959468 on Build GRCh38
rs982550041 Chr.12: 102959281-102959281 on Build GRCh38
rs998614319 Chr.12: 102959732-102959732 on Build GRCh38
rs531723774 Chr.12: 102959606-102959606 on Build GRCh38
rs532413844 Chr.12: 102959748-102959748 on Build GRCh38
rs897255235 Chr.12: 102959515-102959515 on Build GRCh38
rs1005158382 Chr.12: 102959467-102959467 on Build GRCh38
rs1048970521 Chr.12: 102959434-102959434 on Build GRCh38
rs990778484 Chr.12: 102959569-102959569 on Build GRCh38
rs375372141 Chr.12: 102959295-102959295 on Build GRCh38
rs398020887 Chr.12: 102959440-102959440 on Build GRCh38

View Full Product Details