Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23874387-23874981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNR2
Entrez Gene ID
1269
Gene Name
cannabinoid receptor 2
Gene Aliases
CB-2, CB2, CX5
UniGene
Hs.73037
Gene Chromosome Location
Chr.1: 23870526-23913327 on Build GRCh38
Transcripts
NM_001841.2 XM_011540629.2 XM_017000261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1296085 c.676C>T p.H226Y Chr.1: 23874942-23874942 on Build GRCh38
7076390 c.1027G>A p.D343N Chr.1: 23874591-23874591 on Build GRCh38
7100284 c.1004C>A p.S335Y Chr.1: 23874614-23874614 on Build GRCh38
5528844 c.686G>A p.R229K Chr.1: 23874932-23874932 on Build GRCh38
7100286 c.934C>G p.H312D Chr.1: 23874684-23874684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764328610 Chr.1: 23874957-23874957 on Build GRCh38
rs1016615550 Chr.1: 23874921-23874921 on Build GRCh38
rs2229582 Chr.1: 23874512-23874512 on Build GRCh38
rs761834077 Chr.1: 23874661-23874661 on Build GRCh38
rs191065191 Chr.1: 23874713-23874713 on Build GRCh38
rs780744054 Chr.1: 23874794-23874794 on Build GRCh38
rs780999060 Chr.1: 23874892-23874892 on Build GRCh38
rs923397516 Chr.1: 23874467-23874467 on Build GRCh38
rs148656213 Chr.1: 23874698-23874698 on Build GRCh38
rs747511287 Chr.1: 23874781-23874783 on Build GRCh38
rs2502993 Chr.1: 23874772-23874772 on Build GRCh38
rs201819496 Chr.1: 23874636-23874636 on Build GRCh38
rs776045128 Chr.1: 23874695-23874695 on Build GRCh38
rs143960267 Chr.1: 23874422-23874422 on Build GRCh38
rs2229586 Chr.1: 23874413-23874413 on Build GRCh38
rs887069207 Chr.1: 23874873-23874873 on Build GRCh38
rs148142264 Chr.1: 23874549-23874549 on Build GRCh38
rs201178693 Chr.1: 23874949-23874949 on Build GRCh38
rs750919886 Chr.1: 23874527-23874527 on Build GRCh38
rs758916991 Chr.1: 23874485-23874485 on Build GRCh38
rs191952641 Chr.1: 23874563-23874563 on Build GRCh38
rs201582730 Chr.1: 23874507-23874507 on Build GRCh38
rs201829495 Chr.1: 23874887-23874887 on Build GRCh38
rs778618350 Chr.1: 23874594-23874594 on Build GRCh38
rs142029962 Chr.1: 23874834-23874834 on Build GRCh38
rs201166922 Chr.1: 23874733-23874733 on Build GRCh38
rs754775722 Chr.1: 23874850-23874851 on Build GRCh38
rs772561621 Chr.1: 23874931-23874931 on Build GRCh38
rs751574405 Chr.1: 23874635-23874635 on Build GRCh38
rs765615838 Chr.1: 23874525-23874525 on Build GRCh38
rs202124598 Chr.1: 23874719-23874719 on Build GRCh38
rs760468108 Chr.1: 23874488-23874488 on Build GRCh38
rs201633175 Chr.1: 23874620-23874620 on Build GRCh38
rs199599665 Chr.1: 23874558-23874558 on Build GRCh38
rs764050301 Chr.1: 23874935-23874935 on Build GRCh38
rs988922624 Chr.1: 23874647-23874647 on Build GRCh38
rs200710174 Chr.1: 23874575-23874575 on Build GRCh38
rs199612016 Chr.1: 23874605-23874605 on Build GRCh38
rs781339227 Chr.1: 23874644-23874644 on Build GRCh38
rs563535687 Chr.1: 23874697-23874697 on Build GRCh38
rs367701893 Chr.1: 23874528-23874528 on Build GRCh38
rs201916611 Chr.1: 23874858-23874858 on Build GRCh38
rs920970865 Chr.1: 23874432-23874432 on Build GRCh38
rs201612710 Chr.1: 23874932-23874932 on Build GRCh38
rs985148888 Chr.1: 23874637-23874637 on Build GRCh38
rs766834385 Chr.1: 23874702-23874702 on Build GRCh38
rs200997121 Chr.1: 23874876-23874876 on Build GRCh38
rs771259098 Chr.1: 23874809-23874809 on Build GRCh38
rs2229580 Chr.1: 23874619-23874619 on Build GRCh38
rs201974899 Chr.1: 23874659-23874659 on Build GRCh38
rs760399323 Chr.1: 23874495-23874495 on Build GRCh38
rs201487654 Chr.1: 23874555-23874555 on Build GRCh38
rs200225926 Chr.1: 23874574-23874574 on Build GRCh38
rs754195729 Chr.1: 23874504-23874504 on Build GRCh38
rs764336476 Chr.1: 23874870-23874870 on Build GRCh38
rs747652812 Chr.1: 23874903-23874903 on Build GRCh38
rs111751130 Chr.1: 23874856-23874856 on Build GRCh38
rs201940336 Chr.1: 23874457-23874457 on Build GRCh38
rs368692272 Chr.1: 23874628-23874628 on Build GRCh38
rs895889557 Chr.1: 23874831-23874831 on Build GRCh38
rs375133736 Chr.1: 23874674-23874674 on Build GRCh38
rs201016530 Chr.1: 23874419-23874419 on Build GRCh38
rs201546828 Chr.1: 23874587-23874587 on Build GRCh38
rs200979413 Chr.1: 23874926-23874926 on Build GRCh38
rs923029792 Chr.1: 23874689-23874689 on Build GRCh38
rs778764026 Chr.1: 23874660-23874660 on Build GRCh38
rs879246554 Chr.1: 23874421-23874421 on Build GRCh38
rs962859442 Chr.1: 23874650-23874650 on Build GRCh38
rs201049279 Chr.1: 23874803-23874803 on Build GRCh38
rs183583549 Chr.1: 23874585-23874585 on Build GRCh38
rs762114792 Chr.1: 23874952-23874952 on Build GRCh38
rs2229584 Chr.1: 23874479-23874479 on Build GRCh38
rs756345139 Chr.1: 23874748-23874748 on Build GRCh38
rs745782815 Chr.1: 23874595-23874595 on Build GRCh38
rs578041043 Chr.1: 23874577-23874577 on Build GRCh38
rs144980121 Chr.1: 23874729-23874729 on Build GRCh38
rs199818268 Chr.1: 23874824-23874824 on Build GRCh38
rs773572627 Chr.1: 23874699-23874699 on Build GRCh38
rs763896316 Chr.1: 23874496-23874496 on Build GRCh38
rs764236604 Chr.1: 23874750-23874750 on Build GRCh38
rs746912351 Chr.1: 23874784-23874784 on Build GRCh38
rs770633754 Chr.1: 23874920-23874920 on Build GRCh38
rs201941905 Chr.1: 23874764-23874764 on Build GRCh38
rs2229581 Chr.1: 23874604-23874604 on Build GRCh38
rs2229585 Chr.1: 23874431-23874431 on Build GRCh38
rs752079780 Chr.1: 23874705-23874705 on Build GRCh38
rs551688994 Chr.1: 23874472-23874472 on Build GRCh38
rs1050831547 Chr.1: 23874487-23874487 on Build GRCh38
rs200407932 Chr.1: 23874469-23874469 on Build GRCh38
rs774682084 Chr.1: 23874925-23874925 on Build GRCh38
rs371186526 Chr.1: 23874807-23874807 on Build GRCh38
rs372132839 Chr.1: 23874630-23874630 on Build GRCh38
rs201210941 Chr.1: 23874679-23874679 on Build GRCh38
rs779746814 Chr.1: 23874595-23874596 on Build GRCh38
rs774735405 Chr.1: 23874822-23874822 on Build GRCh38
rs758031503 Chr.1: 23874538-23874538 on Build GRCh38
rs752937607 Chr.1: 23874645-23874645 on Build GRCh38
rs567833720 Chr.1: 23874911-23874911 on Build GRCh38
rs199945476 Chr.1: 23874912-23874912 on Build GRCh38
rs1000489968 Chr.1: 23874539-23874539 on Build GRCh38
rs754091476 Chr.1: 23874875-23874875 on Build GRCh38
rs760083242 Chr.1: 23874714-23874714 on Build GRCh38
rs187686608 Chr.1: 23874625-23874625 on Build GRCh38
rs200157579 Chr.1: 23874624-23874624 on Build GRCh38
rs4649124 Chr.1: 23874867-23874867 on Build GRCh38
rs201160547 Chr.1: 23874446-23874446 on Build GRCh38
rs541088933 Chr.1: 23874603-23874603 on Build GRCh38
rs781130624 Chr.1: 23874852-23874855 on Build GRCh38
rs201088433 Chr.1: 23874658-23874658 on Build GRCh38
rs954473721 Chr.1: 23874690-23874690 on Build GRCh38
rs760759238 Chr.1: 23874934-23874934 on Build GRCh38
rs992946797 Chr.1: 23874414-23874414 on Build GRCh38
rs550621200 Chr.1: 23874680-23874680 on Build GRCh38
rs201461373 Chr.1: 23874470-23874470 on Build GRCh38
rs202216673 Chr.1: 23874420-23874420 on Build GRCh38
rs200162708 Chr.1: 23874936-23874936 on Build GRCh38
rs376013451 Chr.1: 23874874-23874874 on Build GRCh38
rs747658837 Chr.1: 23874893-23874893 on Build GRCh38
rs3003336 Chr.1: 23874958-23874958 on Build GRCh38
rs200211130 Chr.1: 23874802-23874802 on Build GRCh38
rs766274704 Chr.1: 23874886-23874886 on Build GRCh38
rs2229583 Chr.1: 23874493-23874493 on Build GRCh38
rs146922938 Chr.1: 23874779-23874779 on Build GRCh38
rs761421274 Chr.1: 23874614-23874614 on Build GRCh38
rs2229579 Chr.1: 23874672-23874672 on Build GRCh38
rs199680710 Chr.1: 23874793-23874793 on Build GRCh38

View Full Product Details